Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 107866535-107867027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM155A
Entrez Gene ID
728215
Gene Name
family with sequence similarity 155 member A
Gene Aliases
-
UniGene
Hs.535394
Gene Chromosome Location
Chr.13: 107168531-107867255 on Build GRCh38
Transcripts
NM_001080396.2 XM_011521109.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122984 c.3G>C p.M1I Chr.13: 107866594-107866594 on Build GRCh38
6658683 c.61G>A p.A21T Chr.13: 107866536-107866536 on Build GRCh38
5435794 c.26G>A p.R9Q Chr.13: 107866571-107866571 on Build GRCh38
6658666 c.5C>T p.T2I Chr.13: 107866592-107866592 on Build GRCh38
6658674 c.39C>T p.D13D Chr.13: 107866558-107866558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976746418 Chr.13: 107866738-107866738 on Build GRCh38
rs560324592 Chr.13: 107866848-107866848 on Build GRCh38
rs914516345 Chr.13: 107866757-107866757 on Build GRCh38
rs780045784 Chr.13: 107866598-107866598 on Build GRCh38
rs575855777 Chr.13: 107866633-107866633 on Build GRCh38
rs35939139 Chr.13: 107866900-107866901 on Build GRCh38
rs966049437 Chr.13: 107866704-107866704 on Build GRCh38
rs778929421 Chr.13: 107866874-107866874 on Build GRCh38
rs753292869 Chr.13: 107866572-107866572 on Build GRCh38
rs562648417 Chr.13: 107866926-107866926 on Build GRCh38
rs1055635599 Chr.13: 107866756-107866759 on Build GRCh38
rs947273650 Chr.13: 107866766-107866771 on Build GRCh38
rs1040833664 Chr.13: 107866813-107866813 on Build GRCh38
rs1033811221 Chr.13: 107866901-107866901 on Build GRCh38
rs183766307 Chr.13: 107866834-107866834 on Build GRCh38
rs35028871 Chr.13: 107866719-107866722 on Build GRCh38
rs901385772 Chr.13: 107867001-107867001 on Build GRCh38
rs982222471 Chr.13: 107866755-107866756 on Build GRCh38
rs746921336 Chr.13: 107866631-107866631 on Build GRCh38
rs899609528 Chr.13: 107866967-107866967 on Build GRCh38
rs549127147 Chr.13: 107866918-107866918 on Build GRCh38
rs201183573 Chr.13: 107866791-107866791 on Build GRCh38
rs766769005 Chr.13: 107866560-107866560 on Build GRCh38
rs1048108028 Chr.13: 107866992-107866992 on Build GRCh38
rs1008274825 Chr.13: 107866812-107866812 on Build GRCh38
rs1027233256 Chr.13: 107866728-107866728 on Build GRCh38
rs369812541 Chr.13: 107866890-107866890 on Build GRCh38
rs894829003 Chr.13: 107866892-107866892 on Build GRCh38
rs997415246 Chr.13: 107866727-107866727 on Build GRCh38
rs765281952 Chr.13: 107866902-107866903 on Build GRCh38
rs760481020 Chr.13: 107866721-107866722 on Build GRCh38
rs954075901 Chr.13: 107866760-107866760 on Build GRCh38
rs1027083334 Chr.13: 107866648-107866648 on Build GRCh38
rs758238905 Chr.13: 107866592-107866592 on Build GRCh38
rs998446632 Chr.13: 107867005-107867005 on Build GRCh38
rs997049019 Chr.13: 107866827-107866827 on Build GRCh38
rs1015834158 Chr.13: 107866695-107866695 on Build GRCh38
rs1016216251 Chr.13: 107866963-107866963 on Build GRCh38
rs778699110 Chr.13: 107866587-107866587 on Build GRCh38
rs147671709 Chr.13: 107866561-107866561 on Build GRCh38
rs952851175 Chr.13: 107866852-107866852 on Build GRCh38
rs201040672 Chr.13: 107866725-107866725 on Build GRCh38
rs902869787 Chr.13: 107866788-107866791 on Build GRCh38
rs989576780 Chr.13: 107866765-107866765 on Build GRCh38
rs143934150 Chr.13: 107866756-107866757 on Build GRCh38
rs902964547 Chr.13: 107866667-107866667 on Build GRCh38
rs982514349 Chr.13: 107866900-107866909 on Build GRCh38
rs369867004 Chr.13: 107866886-107866886 on Build GRCh38
rs776549432 Chr.13: 107866757-107866758 on Build GRCh38
rs894861740 Chr.13: 107866586-107866586 on Build GRCh38
rs775530329 Chr.13: 107866927-107866938 on Build GRCh38
rs930373237 Chr.13: 107866898-107866898 on Build GRCh38
rs914423729 Chr.13: 107866920-107866920 on Build GRCh38
rs952945308 Chr.13: 107866729-107866729 on Build GRCh38
rs959705754 Chr.13: 107866914-107866915 on Build GRCh38
rs924513481 Chr.13: 107866931-107866931 on Build GRCh38
rs977342806 Chr.13: 107866930-107866930 on Build GRCh38
rs1041628945 Chr.13: 107866788-107866789 on Build GRCh38
rs982773381 Chr.13: 107866733-107866742 on Build GRCh38
rs533102222 Chr.13: 107866782-107866782 on Build GRCh38
rs777298462 Chr.13: 107866563-107866563 on Build GRCh38
rs1054761362 Chr.13: 107866574-107866574 on Build GRCh38
rs796301951 Chr.13: 107866920-107866921 on Build GRCh38
rs921268787 Chr.13: 107866742-107866742 on Build GRCh38
rs1004813961 Chr.13: 107866682-107866682 on Build GRCh38
rs139671494 Chr.13: 107866775-107866775 on Build GRCh38
rs555804749 Chr.13: 107866589-107866589 on Build GRCh38
rs899577685 Chr.13: 107866816-107866816 on Build GRCh38
rs763303628 Chr.13: 107866557-107866557 on Build GRCh38
rs994556553 Chr.13: 107866619-107866619 on Build GRCh38
rs544232672 Chr.13: 107866766-107866766 on Build GRCh38
rs932415743 Chr.13: 107866975-107866975 on Build GRCh38
rs1004376877 Chr.13: 107866942-107866942 on Build GRCh38
rs1054687347 Chr.13: 107866885-107866885 on Build GRCh38
rs1019741036 Chr.13: 107866677-107866677 on Build GRCh38
rs76723856 Chr.13: 107866761-107866761 on Build GRCh38
rs952929352 Chr.13: 107866673-107866673 on Build GRCh38
rs372260922 Chr.13: 107866725-107866728 on Build GRCh38
rs187089646 Chr.13: 107866871-107866871 on Build GRCh38
rs947181807 Chr.13: 107866928-107866928 on Build GRCh38
rs756877201 Chr.13: 107866584-107866584 on Build GRCh38
rs915033385 Chr.13: 107866768-107866768 on Build GRCh38
rs748120698 Chr.13: 107866900-107866903 on Build GRCh38
rs74114313 Chr.13: 107866938-107866938 on Build GRCh38

View Full Product Details