Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110424566-110425092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A2
Entrez Gene ID
1284
Gene Name
collagen type IV alpha 2
Gene Aliases
ICH, POREN2
UniGene
Hs.508716
Gene Chromosome Location
Chr.13: 110307284-110513027 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6633188 c.348C>T p.A116A Chr.13: 110424985-110424985 on Build GRCh38
6021240 c.349G>C p.D117H Chr.13: 110424986-110424986 on Build GRCh38
5368864 c.301C>T p.P101S Chr.13: 110424854-110424854 on Build GRCh38
5788722 c.201C>A p.G67G Chr.13: 110424754-110424754 on Build GRCh38
4212973 c.295A>G p.T99A Chr.13: 110424848-110424848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898466541 Chr.13: 110424941-110424941 on Build GRCh38
rs1038417919 Chr.13: 110424932-110424932 on Build GRCh38
rs924955807 Chr.13: 110424687-110424687 on Build GRCh38
rs78713113 Chr.13: 110424882-110424882 on Build GRCh38
rs979079547 Chr.13: 110424679-110424679 on Build GRCh38
rs368648342 Chr.13: 110424902-110424902 on Build GRCh38
rs376916470 Chr.13: 110424747-110424747 on Build GRCh38
rs756286769 Chr.13: 110424873-110424874 on Build GRCh38
rs748175830 Chr.13: 110424712-110424712 on Build GRCh38
rs772419497 Chr.13: 110424945-110424945 on Build GRCh38
rs764272243 Chr.13: 110424896-110424896 on Build GRCh38
rs533541153 Chr.13: 110424911-110424911 on Build GRCh38
rs750061237 Chr.13: 110424795-110424795 on Build GRCh38
rs754810893 Chr.13: 110424900-110424900 on Build GRCh38
rs774623979 Chr.13: 110424745-110424745 on Build GRCh38
rs748157227 Chr.13: 110424840-110424840 on Build GRCh38
rs138453725 Chr.13: 110424919-110424919 on Build GRCh38
rs201777404 Chr.13: 110424862-110424862 on Build GRCh38
rs574404759 Chr.13: 110424616-110424616 on Build GRCh38
rs952733326 Chr.13: 110424642-110424643 on Build GRCh38
rs760573183 Chr.13: 110425035-110425035 on Build GRCh38
rs955569549 Chr.13: 110424594-110424594 on Build GRCh38
rs374976511 Chr.13: 110424986-110424986 on Build GRCh38
rs900964938 Chr.13: 110424632-110424632 on Build GRCh38
rs781221810 Chr.13: 110424838-110424838 on Build GRCh38
rs778470478 Chr.13: 110424689-110424689 on Build GRCh38
rs886049967 Chr.13: 110424948-110424948 on Build GRCh38
rs752982775 Chr.13: 110424693-110424694 on Build GRCh38
rs898519017 Chr.13: 110424725-110424725 on Build GRCh38
rs558384236 Chr.13: 110424910-110424910 on Build GRCh38
rs188399191 Chr.13: 110424695-110424695 on Build GRCh38
rs766078584 Chr.13: 110424801-110424801 on Build GRCh38
rs145900757 Chr.13: 110424872-110424872 on Build GRCh38
rs372424459 Chr.13: 110424641-110424642 on Build GRCh38
rs761236162 Chr.13: 110424877-110424877 on Build GRCh38
rs78090805 Chr.13: 110424638-110424639 on Build GRCh38
rs373525574 Chr.13: 110424708-110424708 on Build GRCh38
rs746943779 Chr.13: 110424696-110424696 on Build GRCh38
rs756791073 Chr.13: 110424688-110424688 on Build GRCh38
rs369171544 Chr.13: 110424703-110424703 on Build GRCh38
rs4238272 Chr.13: 110424850-110424850 on Build GRCh38
rs370490372 Chr.13: 110424841-110424841 on Build GRCh38
rs199799742 Chr.13: 110424949-110424949 on Build GRCh38
rs201980714 Chr.13: 110424935-110424936 on Build GRCh38
rs761254454 Chr.13: 110424777-110424777 on Build GRCh38
rs764706888 Chr.13: 110424878-110424878 on Build GRCh38
rs994273692 Chr.13: 110424635-110424636 on Build GRCh38
rs1005533400 Chr.13: 110424645-110424646 on Build GRCh38
rs778943484 Chr.13: 110424930-110424930 on Build GRCh38
rs749501904 Chr.13: 110424843-110424843 on Build GRCh38
rs779952797 Chr.13: 110424634-110424635 on Build GRCh38
rs932645606 Chr.13: 110424957-110424957 on Build GRCh38
rs200350705 Chr.13: 110424640-110424640 on Build GRCh38
rs4771678 Chr.13: 110424593-110424593 on Build GRCh38
rs751272301 Chr.13: 110424810-110424810 on Build GRCh38
rs376081023 Chr.13: 110424863-110424863 on Build GRCh38
rs370926553 Chr.13: 110424718-110424718 on Build GRCh38
rs115817425 Chr.13: 110424671-110424671 on Build GRCh38
rs758036875 Chr.13: 110424796-110424796 on Build GRCh38
rs1021564488 Chr.13: 110424816-110424816 on Build GRCh38
rs371144381 Chr.13: 110424704-110424704 on Build GRCh38
rs115760431 Chr.13: 110424898-110424898 on Build GRCh38
rs772439696 Chr.13: 110424866-110424866 on Build GRCh38
rs775980504 Chr.13: 110424749-110424749 on Build GRCh38
rs769112358 Chr.13: 110424968-110424968 on Build GRCh38
rs749283036 Chr.13: 110424913-110424913 on Build GRCh38
rs376642856 Chr.13: 110425034-110425034 on Build GRCh38
rs765881647 Chr.13: 110424897-110424897 on Build GRCh38
rs796159244 Chr.13: 110425057-110425057 on Build GRCh38
rs986921498 Chr.13: 110424619-110424619 on Build GRCh38
rs1050228895 Chr.13: 110424961-110424961 on Build GRCh38
rs764782733 Chr.13: 110424793-110424793 on Build GRCh38
rs74967960 Chr.13: 110424887-110424887 on Build GRCh38
rs910889605 Chr.13: 110424760-110424760 on Build GRCh38
rs200140203 Chr.13: 110424716-110424716 on Build GRCh38
rs568065959 Chr.13: 110425057-110425057 on Build GRCh38
rs763868177 Chr.13: 110425036-110425036 on Build GRCh38
rs553654506 Chr.13: 110424693-110424693 on Build GRCh38
rs759305120 Chr.13: 110424998-110424998 on Build GRCh38
rs774660355 Chr.13: 110424855-110424855 on Build GRCh38
rs767000579 Chr.13: 110424640-110424641 on Build GRCh38
rs997013619 Chr.13: 110424781-110424781 on Build GRCh38
rs754829210 Chr.13: 110424837-110424837 on Build GRCh38
rs377651489 Chr.13: 110424713-110424713 on Build GRCh38
rs746144961 Chr.13: 110424938-110424938 on Build GRCh38
rs773874575 Chr.13: 110424996-110424996 on Build GRCh38
rs1042770282 Chr.13: 110424757-110424757 on Build GRCh38
rs375136568 Chr.13: 110425054-110425054 on Build GRCh38
rs537746077 Chr.13: 110424965-110424965 on Build GRCh38
rs1023580071 Chr.13: 110424670-110424670 on Build GRCh38
rs531542947 Chr.13: 110424842-110424842 on Build GRCh38
rs58151684 Chr.13: 110425054-110425054 on Build GRCh38
rs572190601 Chr.13: 110424694-110424694 on Build GRCh38
rs71127938 Chr.13: 110424651-110424652 on Build GRCh38
rs749537372 Chr.13: 110424717-110424717 on Build GRCh38
rs777200276 Chr.13: 110424984-110424984 on Build GRCh38
rs913642254 Chr.13: 110424620-110424620 on Build GRCh38
rs555859760 Chr.13: 110425009-110425009 on Build GRCh38
rs530384029 Chr.13: 110424636-110424637 on Build GRCh38

View Full Product Details