Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110429766-110430300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A2
Entrez Gene ID
1284
Gene Name
collagen type IV alpha 2
Gene Aliases
ICH, POREN2
UniGene
Hs.508716
Gene Chromosome Location
Chr.13: 110307284-110513027 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
945357 c.517G>A p.A173T Chr.13: 110429924-110429924 on Build GRCh38
1196514 c.491C>A p.P164Q Chr.13: 110429898-110429898 on Build GRCh38
6463069 c.478-4T>A p.? Chr.13: 110429881-110429881 on Build GRCh38
6073994 c.480G>T p.G160G Chr.13: 110429887-110429887 on Build GRCh38
5376373 c.483C>T p.P161P Chr.13: 110429890-110429890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74124318 Chr.13: 110429971-110429971 on Build GRCh38
rs555323461 Chr.13: 110430089-110430089 on Build GRCh38
rs886049969 Chr.13: 110429961-110429961 on Build GRCh38
rs938302122 Chr.13: 110430205-110430205 on Build GRCh38
rs986726360 Chr.13: 110429801-110429801 on Build GRCh38
rs986505894 Chr.13: 110430058-110430058 on Build GRCh38
rs889901483 Chr.13: 110430055-110430055 on Build GRCh38
rs772399688 Chr.13: 110430014-110430014 on Build GRCh38
rs566420092 Chr.13: 110429972-110429972 on Build GRCh38
rs371112780 Chr.13: 110430006-110430006 on Build GRCh38
rs541707310 Chr.13: 110430122-110430122 on Build GRCh38
rs774443925 Chr.13: 110429863-110429863 on Build GRCh38
rs994397608 Chr.13: 110430083-110430083 on Build GRCh38
rs925127302 Chr.13: 110430115-110430115 on Build GRCh38
rs769617207 Chr.13: 110429852-110429853 on Build GRCh38
rs576966286 Chr.13: 110430026-110430026 on Build GRCh38
rs772097573 Chr.13: 110429979-110429979 on Build GRCh38
rs764529912 Chr.13: 110429997-110429997 on Build GRCh38
rs772984586 Chr.13: 110429989-110429989 on Build GRCh38
rs768315673 Chr.13: 110429838-110429838 on Build GRCh38
rs138498313 Chr.13: 110430022-110430022 on Build GRCh38
rs988455968 Chr.13: 110430223-110430223 on Build GRCh38
rs779584979 Chr.13: 110429928-110429928 on Build GRCh38
rs972920178 Chr.13: 110429833-110429833 on Build GRCh38
rs748675556 Chr.13: 110430012-110430012 on Build GRCh38
rs764619902 Chr.13: 110429897-110429897 on Build GRCh38
rs537243689 Chr.13: 110430062-110430063 on Build GRCh38
rs747118327 Chr.13: 110430040-110430040 on Build GRCh38
rs952981007 Chr.13: 110430109-110430109 on Build GRCh38
rs201457127 Chr.13: 110429954-110429954 on Build GRCh38
rs979817090 Chr.13: 110430159-110430159 on Build GRCh38
rs753463828 Chr.13: 110429867-110429867 on Build GRCh38
rs779627331 Chr.13: 110429841-110429841 on Build GRCh38
rs766426881 Chr.13: 110429859-110429859 on Build GRCh38
rs965639071 Chr.13: 110429814-110429814 on Build GRCh38
rs141408898 Chr.13: 110430059-110430059 on Build GRCh38
rs3929758 Chr.13: 110429810-110429810 on Build GRCh38
rs780520736 Chr.13: 110429844-110429845 on Build GRCh38
rs762922422 Chr.13: 110429849-110429849 on Build GRCh38
rs568655744 Chr.13: 110429945-110429945 on Build GRCh38
rs374251565 Chr.13: 110429955-110429955 on Build GRCh38
rs535492357 Chr.13: 110429953-110429953 on Build GRCh38
rs531839858 Chr.13: 110429795-110429795 on Build GRCh38
rs556283355 Chr.13: 110429794-110429794 on Build GRCh38
rs143043262 Chr.13: 110430080-110430080 on Build GRCh38
rs928300120 Chr.13: 110430172-110430172 on Build GRCh38
rs756484014 Chr.13: 110429888-110429888 on Build GRCh38
rs984287712 Chr.13: 110429846-110429846 on Build GRCh38
rs769636364 Chr.13: 110429845-110429845 on Build GRCh38
rs148199632 Chr.13: 110430108-110430108 on Build GRCh38
rs970216341 Chr.13: 110430152-110430152 on Build GRCh38
rs527590704 Chr.13: 110430184-110430184 on Build GRCh38
rs560006500 Chr.13: 110430163-110430163 on Build GRCh38
rs775774951 Chr.13: 110429983-110429983 on Build GRCh38
rs1043977516 Chr.13: 110429828-110429828 on Build GRCh38
rs371699757 Chr.13: 110430190-110430190 on Build GRCh38
rs755371182 Chr.13: 110429898-110429898 on Build GRCh38
rs776155180 Chr.13: 110429839-110429839 on Build GRCh38
rs752871655 Chr.13: 110429868-110429868 on Build GRCh38
rs201486366 Chr.13: 110429880-110429880 on Build GRCh38
rs759600188 Chr.13: 110429974-110429974 on Build GRCh38
rs908986004 Chr.13: 110429848-110429848 on Build GRCh38
rs746617802 Chr.13: 110429837-110429837 on Build GRCh38
rs754205667 Chr.13: 110430004-110430004 on Build GRCh38
rs757886951 Chr.13: 110429924-110429924 on Build GRCh38
rs747835784 Chr.13: 110429847-110429849 on Build GRCh38
rs913855477 Chr.13: 110429802-110429802 on Build GRCh38
rs761017209 Chr.13: 110429989-110429989 on Build GRCh38
rs866316171 Chr.13: 110430098-110430098 on Build GRCh38
rs950035585 Chr.13: 110430232-110430232 on Build GRCh38
rs1056275173 Chr.13: 110430125-110430125 on Build GRCh38
rs916033758 Chr.13: 110430144-110430144 on Build GRCh38
rs751015466 Chr.13: 110429942-110429942 on Build GRCh38
rs1007330318 Chr.13: 110429808-110429808 on Build GRCh38
rs759563517 Chr.13: 110429864-110429864 on Build GRCh38
rs773133213 Chr.13: 110429847-110429847 on Build GRCh38
rs913167447 Chr.13: 110430238-110430241 on Build GRCh38
rs1019575424 Chr.13: 110430067-110430067 on Build GRCh38
rs754461131 Chr.13: 110429944-110429944 on Build GRCh38

View Full Product Details