Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21070287-21070790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF40
Entrez Gene ID
55701
Gene Name
Rho guanine nucleotide exchange factor 40
Gene Aliases
SOLO
UniGene
Hs.35125
Gene Chromosome Location
Chr.14: 21061276-21090248 on Build GRCh38
Transcripts
NM_001278529.1 NM_001278530.1 NM_018071.4 XM_005267844.2 XM_011536937.2 XM_011536938.2 XM_017021434.1 XM_017021435.1 NM_001282211.1

Gene Symbol
NDRG2
Entrez Gene ID
57447
Gene Name
NDRG family member 2
Gene Aliases
SYLD
UniGene
Hs.525205
Gene Chromosome Location
Chr.14: 21016763-21070872 on Build GRCh38
Transcripts
NM_001278529.1 NM_001278530.1 NM_018071.4 XM_005267844.2 XM_011536937.2 XM_011536938.2 XM_017021434.1 XM_017021435.1 NM_001282211.1

SNP Detail

SNP ID Chromosome Location
rs759555308 Chr.14: 21070347-21070347 on Build GRCh38
rs959041992 Chr.14: 21070433-21070433 on Build GRCh38
rs112878437 Chr.14: 21070732-21070732 on Build GRCh38
rs1010594296 Chr.14: 21070340-21070340 on Build GRCh38
rs574697341 Chr.14: 21070587-21070587 on Build GRCh38
rs1034966690 Chr.14: 21070654-21070654 on Build GRCh38
rs1003350390 Chr.14: 21070647-21070647 on Build GRCh38
rs756607799 Chr.14: 21070374-21070374 on Build GRCh38
rs372003858 Chr.14: 21070683-21070684 on Build GRCh38
rs949917478 Chr.14: 21070601-21070601 on Build GRCh38
rs925611167 Chr.14: 21070540-21070540 on Build GRCh38
rs560871624 Chr.14: 21070660-21070660 on Build GRCh38
rs1045673493 Chr.14: 21070335-21070335 on Build GRCh38
rs185060561 Chr.14: 21070731-21070731 on Build GRCh38
rs968199763 Chr.14: 21070458-21070458 on Build GRCh38
rs950644581 Chr.14: 21070320-21070320 on Build GRCh38
rs758108745 Chr.14: 21070441-21070441 on Build GRCh38
rs770424666 Chr.14: 21070489-21070489 on Build GRCh38
rs1025811807 Chr.14: 21070446-21070446 on Build GRCh38
rs1025754658 Chr.14: 21070755-21070755 on Build GRCh38
rs1045989243 Chr.14: 21070618-21070618 on Build GRCh38
rs942733003 Chr.14: 21070539-21070539 on Build GRCh38
rs915022358 Chr.14: 21070450-21070450 on Build GRCh38
rs1052845231 Chr.14: 21070318-21070318 on Build GRCh38
rs767571298 Chr.14: 21070361-21070361 on Build GRCh38
rs964656411 Chr.14: 21070490-21070490 on Build GRCh38
rs754177738 Chr.14: 21070389-21070389 on Build GRCh38
rs1026561734 Chr.14: 21070310-21070310 on Build GRCh38
rs987438211 Chr.14: 21070486-21070486 on Build GRCh38
rs115796355 Chr.14: 21070509-21070509 on Build GRCh38
rs1001514789 Chr.14: 21070339-21070339 on Build GRCh38
rs988553861 Chr.14: 21070562-21070562 on Build GRCh38
rs892453357 Chr.14: 21070334-21070334 on Build GRCh38
rs905847407 Chr.14: 21070337-21070337 on Build GRCh38
rs898543995 Chr.14: 21070381-21070381 on Build GRCh38
rs1040749413 Chr.14: 21070330-21070330 on Build GRCh38
rs186973954 Chr.14: 21070385-21070385 on Build GRCh38
rs571291167 Chr.14: 21070679-21070679 on Build GRCh38
rs192490228 Chr.14: 21070497-21070497 on Build GRCh38
rs563223886 Chr.14: 21070521-21070521 on Build GRCh38
rs147829254 Chr.14: 21070528-21070528 on Build GRCh38
rs371932097 Chr.14: 21070498-21070498 on Build GRCh38
rs374778724 Chr.14: 21070673-21070674 on Build GRCh38
rs544958990 Chr.14: 21070681-21070681 on Build GRCh38
rs774590472 Chr.14: 21070346-21070346 on Build GRCh38
rs949968513 Chr.14: 21070327-21070327 on Build GRCh38
rs58394869 Chr.14: 21070671-21070671 on Build GRCh38
rs554705282 Chr.14: 21070545-21070545 on Build GRCh38
rs540513607 Chr.14: 21070643-21070643 on Build GRCh38
rs549612692 Chr.14: 21070618-21070618 on Build GRCh38
rs1003467019 Chr.14: 21070325-21070325 on Build GRCh38
rs753203762 Chr.14: 21070370-21070370 on Build GRCh38
rs1047106344 Chr.14: 21070610-21070610 on Build GRCh38
rs911444344 Chr.14: 21070521-21070521 on Build GRCh38
rs1051303171 Chr.14: 21070510-21070510 on Build GRCh38
rs1038756276 Chr.14: 21070543-21070543 on Build GRCh38
rs564741947 Chr.14: 21070728-21070728 on Build GRCh38
rs1022370861 Chr.14: 21070673-21070673 on Build GRCh38
rs773921131 Chr.14: 21070703-21070703 on Build GRCh38
rs886293980 Chr.14: 21070627-21070627 on Build GRCh38

View Full Product Details