Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21075508-21076008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF40
Entrez Gene ID
55701
Gene Name
Rho guanine nucleotide exchange factor 40
Gene Aliases
SOLO
UniGene
Hs.35125
Gene Chromosome Location
Chr.14: 21061276-21090248 on Build GRCh38
Transcripts
NM_001278529.1 NM_001278530.1 NM_018071.4 XM_005267844.2 XM_011536937.2 XM_011536938.2 XM_017021434.1 XM_017021435.1 XM_017021436.1 XM_017021437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6885147 c.1687C>T p.P563S Chr.14: 21075713-21075713 on Build GRCh38
6388454 c.1729T>C p.S577P Chr.14: 21075755-21075755 on Build GRCh38
2029518 c.1648C>T p.L550L Chr.14: 21075674-21075674 on Build GRCh38
6885148 c.1688C>T p.P563L Chr.14: 21075714-21075714 on Build GRCh38
7408956 c.1622G>T p.G541V Chr.14: 21075648-21075648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764706347 Chr.14: 21075647-21075647 on Build GRCh38
rs760876288 Chr.14: 21075846-21075846 on Build GRCh38
rs117657505 Chr.14: 21075574-21075574 on Build GRCh38
rs760901740 Chr.14: 21075723-21075723 on Build GRCh38
rs897697354 Chr.14: 21075937-21075937 on Build GRCh38
rs780896988 Chr.14: 21075772-21075772 on Build GRCh38
rs541008238 Chr.14: 21075820-21075820 on Build GRCh38
rs773309785 Chr.14: 21075585-21075585 on Build GRCh38
rs200069601 Chr.14: 21075536-21075536 on Build GRCh38
rs374202243 Chr.14: 21075548-21075548 on Build GRCh38
rs755823111 Chr.14: 21075796-21075796 on Build GRCh38
rs777007611 Chr.14: 21075560-21075560 on Build GRCh38
rs778056778 Chr.14: 21075803-21075803 on Build GRCh38
rs1034150311 Chr.14: 21075764-21075764 on Build GRCh38
rs1005983854 Chr.14: 21075857-21075857 on Build GRCh38
rs760036292 Chr.14: 21075715-21075715 on Build GRCh38
rs776747562 Chr.14: 21075723-21075724 on Build GRCh38
rs7140850 Chr.14: 21075897-21075897 on Build GRCh38
rs780242718 Chr.14: 21075693-21075693 on Build GRCh38
rs374757174 Chr.14: 21075711-21075711 on Build GRCh38
rs888844298 Chr.14: 21075980-21075980 on Build GRCh38
rs763416929 Chr.14: 21075587-21075587 on Build GRCh38
rs199597571 Chr.14: 21075771-21075771 on Build GRCh38
rs112166570 Chr.14: 21075871-21075871 on Build GRCh38
rs781447998 Chr.14: 21075830-21075832 on Build GRCh38
rs753505350 Chr.14: 21075621-21075621 on Build GRCh38
rs762080940 Chr.14: 21075564-21075564 on Build GRCh38
rs771068985 Chr.14: 21075813-21075813 on Build GRCh38
rs371182651 Chr.14: 21075676-21075676 on Build GRCh38
rs754878151 Chr.14: 21075682-21075682 on Build GRCh38
rs764973823 Chr.14: 21075726-21075726 on Build GRCh38
rs201075315 Chr.14: 21075613-21075613 on Build GRCh38
rs781036471 Chr.14: 21075688-21075688 on Build GRCh38
rs749979677 Chr.14: 21075727-21075727 on Build GRCh38
rs767913907 Chr.14: 21075716-21075716 on Build GRCh38
rs765974464 Chr.14: 21075749-21075749 on Build GRCh38
rs773255132 Chr.14: 21075692-21075692 on Build GRCh38
rs144572940 Chr.14: 21075720-21075720 on Build GRCh38
rs748356926 Chr.14: 21075690-21075690 on Build GRCh38
rs757944275 Chr.14: 21075673-21075673 on Build GRCh38
rs201983830 Chr.14: 21075600-21075600 on Build GRCh38
rs191691049 Chr.14: 21075810-21075810 on Build GRCh38
rs372104186 Chr.14: 21075599-21075599 on Build GRCh38
rs934903645 Chr.14: 21075763-21075763 on Build GRCh38
rs756905935 Chr.14: 21075622-21075622 on Build GRCh38
rs555231625 Chr.14: 21075710-21075710 on Build GRCh38
rs747994230 Chr.14: 21075790-21075790 on Build GRCh38
rs187300643 Chr.14: 21075551-21075551 on Build GRCh38
rs756975782 Chr.14: 21075629-21075629 on Build GRCh38
rs986107515 Chr.14: 21075637-21075637 on Build GRCh38
rs768499095 Chr.14: 21075553-21075553 on Build GRCh38
rs989921275 Chr.14: 21075797-21075797 on Build GRCh38
rs777484207 Chr.14: 21075555-21075555 on Build GRCh38
rs181747941 Chr.14: 21075834-21075834 on Build GRCh38
rs543869311 Chr.14: 21075974-21075974 on Build GRCh38
rs962236326 Chr.14: 21075554-21075554 on Build GRCh38
rs757995602 Chr.14: 21075744-21075744 on Build GRCh38
rs751612755 Chr.14: 21075770-21075770 on Build GRCh38
rs113362065 Chr.14: 21075620-21075620 on Build GRCh38
rs368329545 Chr.14: 21075550-21075550 on Build GRCh38
rs140826804 Chr.14: 21075666-21075666 on Build GRCh38
rs768953756 Chr.14: 21075789-21075789 on Build GRCh38
rs768328233 Chr.14: 21075707-21075707 on Build GRCh38
rs977274146 Chr.14: 21075864-21075864 on Build GRCh38
rs993533844 Chr.14: 21075658-21075658 on Build GRCh38
rs1050557644 Chr.14: 21075581-21075581 on Build GRCh38
rs774388173 Chr.14: 21075814-21075814 on Build GRCh38
rs772078806 Chr.14: 21075917-21075917 on Build GRCh38
rs770033714 Chr.14: 21075691-21075691 on Build GRCh38
rs549442706 Chr.14: 21075577-21075577 on Build GRCh38

View Full Product Details