Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21392372-21392886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD8
Entrez Gene ID
57680
Gene Name
chromodomain helicase DNA binding protein 8
Gene Aliases
AUTS18, HELSNF1
UniGene
Hs.530698
Gene Chromosome Location
Chr.14: 21385194-21437298 on Build GRCh38
Transcripts
NM_001170629.1 NM_020920.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628185 c.6728G>A p.R2243H Chr.14: 21392550-21392550 on Build GRCh38
182860 c.6499G>A p.V2167I Chr.14: 21392779-21392779 on Build GRCh38
6417845 c.5729G>A p.G1910E Chr.14: 21392712-21392712 on Build GRCh38
2029931 c.5868G>A p.A1956A Chr.14: 21392573-21392573 on Build GRCh38
4622915 c.6686C>T p.A2229V Chr.14: 21392592-21392592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759593522 Chr.14: 21392470-21392470 on Build GRCh38
rs775961607 Chr.14: 21392537-21392537 on Build GRCh38
rs901063076 Chr.14: 21392429-21392429 on Build GRCh38
rs762678508 Chr.14: 21392738-21392738 on Build GRCh38
rs767339333 Chr.14: 21392579-21392579 on Build GRCh38
rs928363428 Chr.14: 21392780-21392780 on Build GRCh38
rs894725097 Chr.14: 21392499-21392499 on Build GRCh38
rs775420723 Chr.14: 21392831-21392831 on Build GRCh38
rs748929440 Chr.14: 21392765-21392765 on Build GRCh38
rs761993643 Chr.14: 21392859-21392859 on Build GRCh38
rs760613512 Chr.14: 21392842-21392842 on Build GRCh38
rs759519178 Chr.14: 21392700-21392700 on Build GRCh38
rs528376589 Chr.14: 21392675-21392675 on Build GRCh38
rs747458076 Chr.14: 21392747-21392747 on Build GRCh38
rs947536597 Chr.14: 21392618-21392618 on Build GRCh38
rs770593901 Chr.14: 21392779-21392779 on Build GRCh38
rs752553650 Chr.14: 21392703-21392703 on Build GRCh38
rs769558618 Chr.14: 21392656-21392656 on Build GRCh38
rs780618366 Chr.14: 21392638-21392638 on Build GRCh38
rs373651476 Chr.14: 21392740-21392740 on Build GRCh38
rs756384802 Chr.14: 21392586-21392586 on Build GRCh38
rs777956601 Chr.14: 21392465-21392465 on Build GRCh38
rs745375504 Chr.14: 21392805-21392805 on Build GRCh38
rs201511497 Chr.14: 21392509-21392509 on Build GRCh38
rs763827337 Chr.14: 21392843-21392843 on Build GRCh38
rs144602267 Chr.14: 21392862-21392862 on Build GRCh38
rs61756312 Chr.14: 21392741-21392741 on Build GRCh38
rs939781466 Chr.14: 21392830-21392830 on Build GRCh38
rs765854470 Chr.14: 21392549-21392549 on Build GRCh38
rs780706133 Chr.14: 21392745-21392745 on Build GRCh38
rs371670547 Chr.14: 21392717-21392717 on Build GRCh38
rs996788249 Chr.14: 21392452-21392452 on Build GRCh38
rs769311687 Chr.14: 21392545-21392545 on Build GRCh38
rs763741439 Chr.14: 21392756-21392756 on Build GRCh38
rs781575717 Chr.14: 21392760-21392760 on Build GRCh38
rs149307240 Chr.14: 21392628-21392628 on Build GRCh38
rs778904376 Chr.14: 21392739-21392739 on Build GRCh38
rs773816387 Chr.14: 21392691-21392691 on Build GRCh38
rs747368212 Chr.14: 21392644-21392644 on Build GRCh38
rs754027111 Chr.14: 21392620-21392620 on Build GRCh38
rs982580250 Chr.14: 21392771-21392771 on Build GRCh38
rs773908554 Chr.14: 21392782-21392782 on Build GRCh38
rs751516489 Chr.14: 21392550-21392550 on Build GRCh38
rs772017728 Chr.14: 21392618-21392620 on Build GRCh38
rs761601600 Chr.14: 21392541-21392541 on Build GRCh38
rs753480258 Chr.14: 21392819-21392822 on Build GRCh38
rs377559805 Chr.14: 21392574-21392574 on Build GRCh38
rs762371692 Chr.14: 21392547-21392547 on Build GRCh38
rs772679723 Chr.14: 21392635-21392635 on Build GRCh38
rs752465998 Chr.14: 21392584-21392584 on Build GRCh38
rs770376299 Chr.14: 21392685-21392685 on Build GRCh38
rs772750664 Chr.14: 21392671-21392671 on Build GRCh38
rs767428909 Chr.14: 21392701-21392701 on Build GRCh38
rs775741325 Chr.14: 21392827-21392827 on Build GRCh38
rs777851125 Chr.14: 21392608-21392608 on Build GRCh38
rs749313449 Chr.14: 21392477-21392477 on Build GRCh38
rs757299407 Chr.14: 21392621-21392621 on Build GRCh38
rs768922999 Chr.14: 21392748-21392748 on Build GRCh38
rs779003135 Chr.14: 21392622-21392622 on Build GRCh38
rs371915075 Chr.14: 21392806-21392806 on Build GRCh38
rs966821898 Chr.14: 21392686-21392686 on Build GRCh38
rs776456494 Chr.14: 21392852-21392852 on Build GRCh38
rs61741241 Chr.14: 21392707-21392707 on Build GRCh38
rs991113852 Chr.14: 21392844-21392844 on Build GRCh38
rs771537778 Chr.14: 21392478-21392478 on Build GRCh38
rs893621541 Chr.14: 21392587-21392587 on Build GRCh38
rs774726221 Chr.14: 21392487-21392487 on Build GRCh38
rs1040689640 Chr.14: 21392406-21392406 on Build GRCh38
rs772587635 Chr.14: 21392522-21392522 on Build GRCh38
rs367894618 Chr.14: 21392713-21392713 on Build GRCh38

View Full Product Details