Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23064022-23064523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACIN1
Entrez Gene ID
22985
Gene Name
apoptotic chromatin condensation inducer 1
Gene Aliases
ACINUS, ACN, fSAP152
UniGene
Hs.124490
Gene Chromosome Location
Chr.14: 23058564-23095614 on Build GRCh38
Transcripts
NM_001164814.1 NM_001164815.1 NM_001164816.1 NM_001164817.1 NM_014977.3 XM_005267415.3 XM_005267416.3 XM_005267418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7299517 c.436C>T p.R146C Chr.14: 23064164-23064164 on Build GRCh38
2031113 c.2687G>A p.R896H Chr.14: 23064187-23064187 on Build GRCh38
6592062 c.2646G>A p.A882A Chr.14: 23064228-23064228 on Build GRCh38
4853253 c.529C>T p.R177C Chr.14: 23064164-23064164 on Build GRCh38
6592061 c.266G>A p.R89Q Chr.14: 23064431-23064431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898153238 Chr.14: 23064241-23064241 on Build GRCh38
rs760783145 Chr.14: 23064277-23064277 on Build GRCh38
rs559108629 Chr.14: 23064084-23064084 on Build GRCh38
rs375819825 Chr.14: 23064317-23064317 on Build GRCh38
rs756553368 Chr.14: 23064238-23064238 on Build GRCh38
rs148549658 Chr.14: 23064145-23064145 on Build GRCh38
rs752794146 Chr.14: 23064156-23064156 on Build GRCh38
rs770053332 Chr.14: 23064089-23064089 on Build GRCh38
rs773830224 Chr.14: 23064090-23064090 on Build GRCh38
rs768779890 Chr.14: 23064443-23064443 on Build GRCh38
rs952200881 Chr.14: 23064191-23064191 on Build GRCh38
rs755755592 Chr.14: 23064343-23064343 on Build GRCh38
rs367925618 Chr.14: 23064078-23064078 on Build GRCh38
rs754578279 Chr.14: 23064316-23064316 on Build GRCh38
rs201045976 Chr.14: 23064066-23064066 on Build GRCh38
rs974878808 Chr.14: 23064170-23064170 on Build GRCh38
rs779630005 Chr.14: 23064381-23064381 on Build GRCh38
rs905675829 Chr.14: 23064364-23064364 on Build GRCh38
rs905730261 Chr.14: 23064080-23064080 on Build GRCh38
rs757627793 Chr.14: 23064379-23064379 on Build GRCh38
rs202214497 Chr.14: 23064345-23064345 on Build GRCh38
rs766797052 Chr.14: 23064079-23064080 on Build GRCh38
rs750420657 Chr.14: 23064163-23064163 on Build GRCh38
rs772241048 Chr.14: 23064415-23064415 on Build GRCh38
rs748554203 Chr.14: 23064087-23064087 on Build GRCh38
rs746698786 Chr.14: 23064065-23064065 on Build GRCh38
rs746362549 Chr.14: 23064398-23064398 on Build GRCh38
rs956006612 Chr.14: 23064118-23064118 on Build GRCh38
rs751498032 Chr.14: 23064307-23064307 on Build GRCh38
rs763534511 Chr.14: 23064134-23064134 on Build GRCh38
rs1018979030 Chr.14: 23064141-23064141 on Build GRCh38
rs116199445 Chr.14: 23064467-23064467 on Build GRCh38
rs1037099136 Chr.14: 23064287-23064287 on Build GRCh38
rs371720632 Chr.14: 23064275-23064275 on Build GRCh38
rs766312604 Chr.14: 23064300-23064300 on Build GRCh38
rs770392805 Chr.14: 23064496-23064496 on Build GRCh38
rs566549950 Chr.14: 23064055-23064055 on Build GRCh38
rs774605825 Chr.14: 23064149-23064149 on Build GRCh38
rs766152645 Chr.14: 23064448-23064448 on Build GRCh38
rs752164185 Chr.14: 23064263-23064264 on Build GRCh38
rs762617835 Chr.14: 23064109-23064109 on Build GRCh38
rs1054262808 Chr.14: 23064438-23064438 on Build GRCh38
rs370843926 Chr.14: 23064071-23064071 on Build GRCh38
rs776771355 Chr.14: 23064445-23064445 on Build GRCh38
rs749631036 Chr.14: 23064246-23064246 on Build GRCh38
rs767793819 Chr.14: 23064155-23064155 on Build GRCh38
rs759363375 Chr.14: 23064479-23064479 on Build GRCh38
rs780968847 Chr.14: 23064229-23064229 on Build GRCh38
rs750851197 Chr.14: 23064071-23064071 on Build GRCh38
rs774324918 Chr.14: 23064469-23064469 on Build GRCh38
rs1052671 Chr.14: 23064440-23064440 on Build GRCh38
rs540621170 Chr.14: 23064434-23064434 on Build GRCh38
rs777970125 Chr.14: 23064245-23064245 on Build GRCh38
rs1032906053 Chr.14: 23064113-23064113 on Build GRCh38
rs751087343 Chr.14: 23064193-23064193 on Build GRCh38
rs764925330 Chr.14: 23064160-23064160 on Build GRCh38
rs544937300 Chr.14: 23064256-23064256 on Build GRCh38
rs767450696 Chr.14: 23064498-23064498 on Build GRCh38
rs760092837 Chr.14: 23064271-23064272 on Build GRCh38
rs759640351 Chr.14: 23064151-23064151 on Build GRCh38
rs758297526 Chr.14: 23064187-23064187 on Build GRCh38
rs377291390 Chr.14: 23064435-23064435 on Build GRCh38
rs774941229 Chr.14: 23064264-23064264 on Build GRCh38
rs1010650087 Chr.14: 23064127-23064127 on Build GRCh38
rs773018856 Chr.14: 23064282-23064282 on Build GRCh38
rs766261167 Chr.14: 23064190-23064190 on Build GRCh38
rs141625134 Chr.14: 23064318-23064318 on Build GRCh38
rs761733382 Chr.14: 23064159-23064159 on Build GRCh38
rs913461100 Chr.14: 23064437-23064437 on Build GRCh38
rs745946594 Chr.14: 23064272-23064272 on Build GRCh38
rs771587723 Chr.14: 23064252-23064252 on Build GRCh38
rs749684082 Chr.14: 23064366-23064366 on Build GRCh38
rs754422933 Chr.14: 23064213-23064213 on Build GRCh38
rs999995446 Chr.14: 23064388-23064388 on Build GRCh38
rs1000046308 Chr.14: 23064102-23064102 on Build GRCh38
rs762806045 Chr.14: 23064447-23064447 on Build GRCh38
rs762682982 Chr.14: 23064298-23064298 on Build GRCh38

View Full Product Details