Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23098617-23099148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf119
Entrez Gene ID
55017
Gene Name
chromosome 14 open reading frame 119
Gene Aliases
-
UniGene
Hs.525238
Gene Chromosome Location
Chr.14: 23095474-23100456 on Build GRCh38
Transcripts
NM_017924.3

SNP Detail

SNP ID Chromosome Location
rs1015909651 Chr.14: 23098853-23098853 on Build GRCh38
rs930466214 Chr.14: 23098728-23098728 on Build GRCh38
rs1028532959 Chr.14: 23098802-23098802 on Build GRCh38
rs372032552 Chr.14: 23098766-23098766 on Build GRCh38
rs113252156 Chr.14: 23098673-23098673 on Build GRCh38
rs775621483 Chr.14: 23098753-23098753 on Build GRCh38
rs572482884 Chr.14: 23099043-23099043 on Build GRCh38
rs982001438 Chr.14: 23098897-23098897 on Build GRCh38
rs539600967 Chr.14: 23098739-23098739 on Build GRCh38
rs985771022 Chr.14: 23098820-23098820 on Build GRCh38
rs1021118937 Chr.14: 23098744-23098744 on Build GRCh38
rs573602277 Chr.14: 23098937-23098937 on Build GRCh38
rs983356366 Chr.14: 23099061-23099061 on Build GRCh38
rs893882173 Chr.14: 23098690-23098690 on Build GRCh38
rs894020879 Chr.14: 23098821-23098821 on Build GRCh38
rs796825933 Chr.14: 23098749-23098749 on Build GRCh38
rs1046354996 Chr.14: 23098653-23098653 on Build GRCh38
rs1042873778 Chr.14: 23099051-23099051 on Build GRCh38
rs945473081 Chr.14: 23099022-23099022 on Build GRCh38
rs937689566 Chr.14: 23098673-23098674 on Build GRCh38
rs761540716 Chr.14: 23098817-23098817 on Build GRCh38
rs369884234 Chr.14: 23099122-23099122 on Build GRCh38
rs959306371 Chr.14: 23098978-23098978 on Build GRCh38
rs2180396 Chr.14: 23098809-23098809 on Build GRCh38
rs183176351 Chr.14: 23098651-23098651 on Build GRCh38
rs201509412 Chr.14: 23098660-23098660 on Build GRCh38
rs747574127 Chr.14: 23099102-23099102 on Build GRCh38
rs1034767757 Chr.14: 23098952-23098952 on Build GRCh38
rs76084661 Chr.14: 23098971-23098971 on Build GRCh38
rs45465198 Chr.14: 23098782-23098782 on Build GRCh38
rs573142701 Chr.14: 23098769-23098769 on Build GRCh38
rs58045401 Chr.14: 23098673-23098674 on Build GRCh38
rs966409001 Chr.14: 23098650-23098650 on Build GRCh38
rs949244913 Chr.14: 23098893-23098893 on Build GRCh38
rs1028501661 Chr.14: 23099108-23099108 on Build GRCh38
rs189210067 Chr.14: 23098855-23098855 on Build GRCh38
rs1053842900 Chr.14: 23098674-23098674 on Build GRCh38
rs539600080 Chr.14: 23098649-23098649 on Build GRCh38
rs767331868 Chr.14: 23098901-23098901 on Build GRCh38
rs1021050359 Chr.14: 23098902-23098902 on Build GRCh38
rs920490856 Chr.14: 23098716-23098718 on Build GRCh38
rs753338353 Chr.14: 23099079-23099079 on Build GRCh38
rs1053768727 Chr.14: 23098945-23098945 on Build GRCh38
rs775482738 Chr.14: 23098659-23098660 on Build GRCh38
rs879129704 Chr.14: 23098791-23098791 on Build GRCh38
rs79381626 Chr.14: 23098654-23098654 on Build GRCh38
rs966836134 Chr.14: 23098903-23098903 on Build GRCh38
rs3742485 Chr.14: 23099077-23099077 on Build GRCh38
rs570709395 Chr.14: 23098729-23098729 on Build GRCh38
rs981813519 Chr.14: 23098652-23098652 on Build GRCh38
rs772807690 Chr.14: 23099010-23099010 on Build GRCh38
rs532349263 Chr.14: 23099089-23099089 on Build GRCh38
rs192891128 Chr.14: 23098926-23098926 on Build GRCh38
rs972259720 Chr.14: 23098865-23098865 on Build GRCh38
rs901206540 Chr.14: 23098814-23098814 on Build GRCh38
rs926541605 Chr.14: 23098907-23098907 on Build GRCh38
rs541526802 Chr.14: 23099058-23099058 on Build GRCh38
rs544286111 Chr.14: 23099046-23099046 on Build GRCh38
rs893837271 Chr.14: 23099006-23099006 on Build GRCh38
rs919064908 Chr.14: 23098876-23098876 on Build GRCh38

View Full Product Details