Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23100344-23100840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf119
Entrez Gene ID
55017
Gene Name
chromosome 14 open reading frame 119
Gene Aliases
-
UniGene
Hs.525238
Gene Chromosome Location
Chr.14: 23095474-23100456 on Build GRCh38
Transcripts
NM_017924.3

Gene Symbol
LOC100128908
Entrez Gene ID
100128908
Gene Name
leishmanolysin homolog
Gene Aliases
-
UniGene
Hs.578287
Gene Chromosome Location
Chr.14: 23100212-23103308 on Build GRCh38
Transcripts
NM_017924.3

SNP Detail

SNP ID Chromosome Location
rs951475732 Chr.14: 23100551-23100551 on Build GRCh38
rs746749428 Chr.14: 23100532-23100532 on Build GRCh38
rs1050164231 Chr.14: 23100791-23100791 on Build GRCh38
rs1047053776 Chr.14: 23100777-23100777 on Build GRCh38
rs898399266 Chr.14: 23100746-23100746 on Build GRCh38
rs979102961 Chr.14: 23100691-23100691 on Build GRCh38
rs935099653 Chr.14: 23100456-23100456 on Build GRCh38
rs149909916 Chr.14: 23100752-23100752 on Build GRCh38
rs1035851752 Chr.14: 23100568-23100568 on Build GRCh38
rs942554866 Chr.14: 23100744-23100744 on Build GRCh38
rs543814502 Chr.14: 23100550-23100550 on Build GRCh38
rs1031090131 Chr.14: 23100604-23100604 on Build GRCh38
rs909405676 Chr.14: 23100429-23100430 on Build GRCh38
rs374592849 Chr.14: 23100700-23100700 on Build GRCh38
rs995850711 Chr.14: 23100762-23100762 on Build GRCh38
rs898466071 Chr.14: 23100499-23100499 on Build GRCh38
rs1043839363 Chr.14: 23100369-23100369 on Build GRCh38
rs923641854 Chr.14: 23100713-23100713 on Build GRCh38
rs1039598920 Chr.14: 23100496-23100496 on Build GRCh38
rs777458525 Chr.14: 23100486-23100486 on Build GRCh38
rs912355137 Chr.14: 23100740-23100740 on Build GRCh38
rs935074758 Chr.14: 23100715-23100715 on Build GRCh38
rs879356106 Chr.14: 23100500-23100500 on Build GRCh38
rs902249933 Chr.14: 23100411-23100411 on Build GRCh38
rs187174627 Chr.14: 23100669-23100669 on Build GRCh38
rs985029651 Chr.14: 23100396-23100396 on Build GRCh38
rs1024087250 Chr.14: 23100670-23100670 on Build GRCh38
rs923058269 Chr.14: 23100779-23100779 on Build GRCh38
rs1050810630 Chr.14: 23100731-23100731 on Build GRCh38
rs566397616 Chr.14: 23100804-23100804 on Build GRCh38
rs74701584 Chr.14: 23100556-23100556 on Build GRCh38
rs1050484976 Chr.14: 23100494-23100494 on Build GRCh38
rs771995866 Chr.14: 23100484-23100484 on Build GRCh38
rs867993387 Chr.14: 23100780-23100780 on Build GRCh38
rs940837070 Chr.14: 23100460-23100460 on Build GRCh38
rs958865333 Chr.14: 23100581-23100581 on Build GRCh38
rs969360600 Chr.14: 23100689-23100689 on Build GRCh38
rs962213013 Chr.14: 23100749-23100749 on Build GRCh38
rs757530732 Chr.14: 23100475-23100479 on Build GRCh38
rs748131832 Chr.14: 23100459-23100459 on Build GRCh38
rs933019317 Chr.14: 23100493-23100493 on Build GRCh38
rs529498249 Chr.14: 23100745-23100745 on Build GRCh38
rs995533455 Chr.14: 23100537-23100537 on Build GRCh38
rs991719384 Chr.14: 23100614-23100614 on Build GRCh38
rs549846583 Chr.14: 23100635-23100635 on Build GRCh38
rs59384505 Chr.14: 23100784-23100784 on Build GRCh38
rs961053934 Chr.14: 23100637-23100637 on Build GRCh38

View Full Product Details