Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23310607-23311102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L2
Entrez Gene ID
599
Gene Name
BCL2 like 2
Gene Aliases
BCL-W, BCL2-L-2, BCLW, PPP1R51
UniGene
Hs.410026
Gene Chromosome Location
Chr.14: 23306762-23311759 on Build GRCh38
Transcripts
NM_001199839.1 NM_004050.4 NM_001199864.1

Gene Symbol
BCL2L2-PABPN1
Entrez Gene ID
100529063
Gene Name
BCL2L2-PABPN1 readthrough
Gene Aliases
-
UniGene
Hs.707712
Gene Chromosome Location
Chr.14: 23306762-23326185 on Build GRCh38
Transcripts
NM_001199839.1 NM_004050.4 NM_001199864.1

SNP Detail

SNP ID Chromosome Location
rs10149339 Chr.14: 23311013-23311013 on Build GRCh38
rs767687695 Chr.14: 23310984-23310984 on Build GRCh38
rs758104312 Chr.14: 23310908-23310908 on Build GRCh38
rs112933062 Chr.14: 23310764-23310764 on Build GRCh38
rs1025178448 Chr.14: 23310742-23310742 on Build GRCh38
rs891539470 Chr.14: 23310639-23310639 on Build GRCh38
rs755229529 Chr.14: 23310831-23310831 on Build GRCh38
rs527518745 Chr.14: 23310967-23310967 on Build GRCh38
rs187184653 Chr.14: 23310923-23310923 on Build GRCh38
rs753786499 Chr.14: 23310745-23310745 on Build GRCh38
rs78478800 Chr.14: 23310731-23310731 on Build GRCh38
rs1003454773 Chr.14: 23310762-23310762 on Build GRCh38
rs759933515 Chr.14: 23310969-23310969 on Build GRCh38
rs183445900 Chr.14: 23310822-23310822 on Build GRCh38
rs1014888578 Chr.14: 23310757-23310757 on Build GRCh38
rs969584249 Chr.14: 23310876-23310876 on Build GRCh38
rs191138928 Chr.14: 23310663-23310663 on Build GRCh38
rs974044053 Chr.14: 23310775-23310775 on Build GRCh38
rs570494160 Chr.14: 23311015-23311015 on Build GRCh38
rs959224263 Chr.14: 23310772-23310772 on Build GRCh38
rs1040625755 Chr.14: 23310681-23310681 on Build GRCh38
rs899756009 Chr.14: 23310670-23310670 on Build GRCh38
rs980927271 Chr.14: 23310906-23310906 on Build GRCh38
rs995949896 Chr.14: 23310690-23310690 on Build GRCh38
rs982864099 Chr.14: 23310815-23310815 on Build GRCh38
rs568264910 Chr.14: 23310944-23310944 on Build GRCh38
rs945692694 Chr.14: 23310997-23310997 on Build GRCh38
rs559834241 Chr.14: 23310688-23310688 on Build GRCh38
rs764971735 Chr.14: 23310803-23310803 on Build GRCh38
rs542856490 Chr.14: 23310709-23310709 on Build GRCh38
rs756829010 Chr.14: 23311081-23311081 on Build GRCh38
rs774993450 Chr.14: 23310947-23310947 on Build GRCh38
rs540082087 Chr.14: 23311031-23311031 on Build GRCh38
rs925531717 Chr.14: 23310914-23310914 on Build GRCh38
rs912866438 Chr.14: 23310939-23310939 on Build GRCh38
rs562728241 Chr.14: 23310839-23310839 on Build GRCh38
rs749607151 Chr.14: 23310786-23310786 on Build GRCh38
rs936573375 Chr.14: 23310975-23310975 on Build GRCh38
rs112390836 Chr.14: 23310938-23310938 on Build GRCh38
rs1009226627 Chr.14: 23311011-23311011 on Build GRCh38
rs527487933 Chr.14: 23310697-23310697 on Build GRCh38
rs3210043 Chr.14: 23310668-23310668 on Build GRCh38
rs931852904 Chr.14: 23311035-23311035 on Build GRCh38

View Full Product Details