Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23310990-23311483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L2
Entrez Gene ID
599
Gene Name
BCL2 like 2
Gene Aliases
BCL-W, BCL2-L-2, BCLW, PPP1R51
UniGene
Hs.410026
Gene Chromosome Location
Chr.14: 23306762-23311759 on Build GRCh38
Transcripts
NM_001199839.1 NM_004050.4 NM_001199864.1

Gene Symbol
BCL2L2-PABPN1
Entrez Gene ID
100529063
Gene Name
BCL2L2-PABPN1 readthrough
Gene Aliases
-
UniGene
Hs.707712
Gene Chromosome Location
Chr.14: 23306762-23326185 on Build GRCh38
Transcripts
NM_001199839.1 NM_004050.4 NM_001199864.1

SNP Detail

SNP ID Chromosome Location
rs10149339 Chr.14: 23311013-23311013 on Build GRCh38
rs758326674 Chr.14: 23311139-23311139 on Build GRCh38
rs780786244 Chr.14: 23311087-23311087 on Build GRCh38
rs569972619 Chr.14: 23311453-23311453 on Build GRCh38
rs988438405 Chr.14: 23311328-23311328 on Build GRCh38
rs45519738 Chr.14: 23311456-23311456 on Build GRCh38
rs142850799 Chr.14: 23311128-23311129 on Build GRCh38
rs374168548 Chr.14: 23311130-23311130 on Build GRCh38
rs951322363 Chr.14: 23311147-23311147 on Build GRCh38
rs550907793 Chr.14: 23311361-23311362 on Build GRCh38
rs897849456 Chr.14: 23311153-23311153 on Build GRCh38
rs945007756 Chr.14: 23311438-23311438 on Build GRCh38
rs749831306 Chr.14: 23311202-23311202 on Build GRCh38
rs965864297 Chr.14: 23311224-23311224 on Build GRCh38
rs958543847 Chr.14: 23311316-23311316 on Build GRCh38
rs754056685 Chr.14: 23311129-23311129 on Build GRCh38
rs1036785604 Chr.14: 23311141-23311141 on Build GRCh38
rs995752800 Chr.14: 23311103-23311103 on Build GRCh38
rs914122872 Chr.14: 23311337-23311337 on Build GRCh38
rs570494160 Chr.14: 23311015-23311015 on Build GRCh38
rs958881436 Chr.14: 23311089-23311089 on Build GRCh38
rs755553188 Chr.14: 23311273-23311273 on Build GRCh38
rs921084280 Chr.14: 23311261-23311261 on Build GRCh38
rs967112420 Chr.14: 23311409-23311409 on Build GRCh38
rs756829010 Chr.14: 23311081-23311081 on Build GRCh38
rs540082087 Chr.14: 23311031-23311031 on Build GRCh38
rs1002373263 Chr.14: 23311275-23311275 on Build GRCh38
rs1006114997 Chr.14: 23311126-23311126 on Build GRCh38
rs571235443 Chr.14: 23311260-23311260 on Build GRCh38
rs975548374 Chr.14: 23311239-23311239 on Build GRCh38
rs1025097969 Chr.14: 23311207-23311207 on Build GRCh38
rs931852904 Chr.14: 23311035-23311035 on Build GRCh38
rs555742144 Chr.14: 23311288-23311288 on Build GRCh38

View Full Product Details