Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23638749-23639248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS2
Entrez Gene ID
10202
Gene Name
dehydrogenase/reductase 2
Gene Aliases
HEP27, SDR25C1
UniGene
Hs.272499
Gene Chromosome Location
Chr.14: 23630115-23645639 on Build GRCh38
Transcripts
NM_001318835.1 NM_005794.3 NM_182908.4 XM_005267249.1 XM_006720001.3 XM_011536338.1 XM_011536339.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3814574 c.133A>C p.T45P Chr.14: 23638997-23638997 on Build GRCh38
1369253 c.96C>T p.G32G Chr.14: 23638960-23638960 on Build GRCh38
6347433 c.6G>T p.L2L Chr.14: 23638870-23638870 on Build GRCh38
7364284 c.190G>A p.V64I Chr.14: 23639228-23639228 on Build GRCh38
5024652 c.133A>G p.T45A Chr.14: 23638997-23638997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746472602 Chr.14: 23638968-23638968 on Build GRCh38
rs965645191 Chr.14: 23638822-23638822 on Build GRCh38
rs781260739 Chr.14: 23638841-23638841 on Build GRCh38
rs772119597 Chr.14: 23638816-23638817 on Build GRCh38
rs765528019 Chr.14: 23638871-23638871 on Build GRCh38
rs916654680 Chr.14: 23639185-23639185 on Build GRCh38
rs560690909 Chr.14: 23639204-23639204 on Build GRCh38
rs758178912 Chr.14: 23639147-23639147 on Build GRCh38
rs768679039 Chr.14: 23639225-23639225 on Build GRCh38
rs777814574 Chr.14: 23638894-23638894 on Build GRCh38
rs146690475 Chr.14: 23639215-23639215 on Build GRCh38
rs187810266 Chr.14: 23638790-23638790 on Build GRCh38
rs144700687 Chr.14: 23639079-23639079 on Build GRCh38
rs756989511 Chr.14: 23638961-23638961 on Build GRCh38
rs199596675 Chr.14: 23639198-23639198 on Build GRCh38
rs138205531 Chr.14: 23639194-23639194 on Build GRCh38
rs746685787 Chr.14: 23639020-23639020 on Build GRCh38
rs771136730 Chr.14: 23639172-23639172 on Build GRCh38
rs2295907 Chr.14: 23638903-23638903 on Build GRCh38
rs759045112 Chr.14: 23638854-23638854 on Build GRCh38
rs922989465 Chr.14: 23638885-23638885 on Build GRCh38
rs201870202 Chr.14: 23638904-23638904 on Build GRCh38
rs749085785 Chr.14: 23638860-23638860 on Build GRCh38
rs146713514 Chr.14: 23638981-23638981 on Build GRCh38
rs148435951 Chr.14: 23638938-23638938 on Build GRCh38
rs771739122 Chr.14: 23638965-23638965 on Build GRCh38
rs761091968 Chr.14: 23638859-23638859 on Build GRCh38
rs199867663 Chr.14: 23639197-23639197 on Build GRCh38
rs550003411 Chr.14: 23639016-23639016 on Build GRCh38
rs141084997 Chr.14: 23638957-23638957 on Build GRCh38
rs748048295 Chr.14: 23638844-23638844 on Build GRCh38
rs759092326 Chr.14: 23638816-23638816 on Build GRCh38
rs769564695 Chr.14: 23638846-23638846 on Build GRCh38
rs372558157 Chr.14: 23638990-23638990 on Build GRCh38
rs576958094 Chr.14: 23639182-23639182 on Build GRCh38
rs757692103 Chr.14: 23638868-23638868 on Build GRCh38
rs779919015 Chr.14: 23639085-23639085 on Build GRCh38
rs767075399 Chr.14: 23638855-23638855 on Build GRCh38
rs776711817 Chr.14: 23639227-23639227 on Build GRCh38
rs1009802405 Chr.14: 23639180-23639180 on Build GRCh38
rs775921464 Chr.14: 23638857-23638857 on Build GRCh38
rs772712044 Chr.14: 23638810-23638810 on Build GRCh38
rs142650346 Chr.14: 23638942-23638942 on Build GRCh38
rs45608041 Chr.14: 23639128-23639128 on Build GRCh38
rs931645520 Chr.14: 23639065-23639065 on Build GRCh38
rs756491932 Chr.14: 23638821-23638821 on Build GRCh38
rs766518719 Chr.14: 23638951-23638951 on Build GRCh38
rs570092444 Chr.14: 23639058-23639058 on Build GRCh38
rs74036809 Chr.14: 23639210-23639210 on Build GRCh38
rs766035204 Chr.14: 23639196-23639196 on Build GRCh38
rs780781180 Chr.14: 23639031-23639031 on Build GRCh38
rs769025294 Chr.14: 23638931-23638931 on Build GRCh38
rs908875787 Chr.14: 23639069-23639069 on Build GRCh38
rs368193807 Chr.14: 23638960-23638960 on Build GRCh38
rs770402015 Chr.14: 23639034-23639034 on Build GRCh38
rs748612949 Chr.14: 23639033-23639033 on Build GRCh38
rs1036677928 Chr.14: 23639071-23639071 on Build GRCh38
rs199641343 Chr.14: 23639222-23639222 on Build GRCh38
rs535835871 Chr.14: 23639066-23639066 on Build GRCh38
rs761199813 Chr.14: 23638920-23638920 on Build GRCh38
rs754678449 Chr.14: 23639159-23639159 on Build GRCh38
rs764383940 Chr.14: 23638823-23638823 on Build GRCh38
rs200598513 Chr.14: 23639183-23639183 on Build GRCh38
rs375928308 Chr.14: 23639169-23639169 on Build GRCh38
rs533750929 Chr.14: 23639153-23639153 on Build GRCh38
rs747248253 Chr.14: 23639224-23639224 on Build GRCh38
rs953032691 Chr.14: 23639064-23639064 on Build GRCh38
rs555965971 Chr.14: 23639200-23639200 on Build GRCh38
rs879040761 Chr.14: 23639068-23639068 on Build GRCh38
rs762765287 Chr.14: 23639162-23639162 on Build GRCh38
rs780298869 Chr.14: 23638820-23638820 on Build GRCh38
rs149593009 Chr.14: 23639202-23639202 on Build GRCh38
rs562008596 Chr.14: 23638974-23638974 on Build GRCh38
rs554920203 Chr.14: 23639121-23639121 on Build GRCh38
rs762976156 Chr.14: 23639195-23639195 on Build GRCh38
rs775569686 Chr.14: 23639056-23639056 on Build GRCh38
rs373176822 Chr.14: 23639168-23639168 on Build GRCh38
rs770464494 Chr.14: 23638850-23638850 on Build GRCh38
rs1002130306 Chr.14: 23638980-23638980 on Build GRCh38
rs747195917 Chr.14: 23638981-23638982 on Build GRCh38
rs760741798 Chr.14: 23639190-23639190 on Build GRCh38
rs757218249 Chr.14: 23638993-23638993 on Build GRCh38
rs2295908 Chr.14: 23638989-23638989 on Build GRCh38
rs749663802 Chr.14: 23639039-23639039 on Build GRCh38
rs140094732 Chr.14: 23638881-23638881 on Build GRCh38
rs919141891 Chr.14: 23639223-23639223 on Build GRCh38
rs772883472 Chr.14: 23639025-23639025 on Build GRCh38
rs1046487246 Chr.14: 23639097-23639097 on Build GRCh38
rs545494016 Chr.14: 23638901-23638901 on Build GRCh38
rs941961243 Chr.14: 23639070-23639070 on Build GRCh38
rs901702684 Chr.14: 23639026-23639026 on Build GRCh38
rs771504271 Chr.14: 23638908-23638908 on Build GRCh38
rs113010522 Chr.14: 23639129-23639129 on Build GRCh38
rs540396182 Chr.14: 23638835-23638835 on Build GRCh38
rs763296378 Chr.14: 23638947-23638947 on Build GRCh38
rs372933053 Chr.14: 23638830-23638830 on Build GRCh38
rs774898009 Chr.14: 23638912-23638912 on Build GRCh38
rs369887257 Chr.14: 23639023-23639023 on Build GRCh38
rs144884940 Chr.14: 23638973-23638973 on Build GRCh38
rs767193133 Chr.14: 23639199-23639199 on Build GRCh38
rs772908006 Chr.14: 23638847-23638847 on Build GRCh38
rs779338474 Chr.14: 23639211-23639211 on Build GRCh38
rs373559238 Chr.14: 23639044-23639044 on Build GRCh38
rs773729400 Chr.14: 23638978-23638978 on Build GRCh38
rs758616199 Chr.14: 23638836-23638836 on Build GRCh38
rs757267631 Chr.14: 23638943-23638943 on Build GRCh38
rs746029038 Chr.14: 23639179-23639179 on Build GRCh38
rs147497806 Chr.14: 23638874-23638874 on Build GRCh38
rs748918137 Chr.14: 23638849-23638849 on Build GRCh38
rs369492474 Chr.14: 23638982-23638982 on Build GRCh38
rs774811528 Chr.14: 23639177-23639177 on Build GRCh38
rs201170104 Chr.14: 23638991-23638991 on Build GRCh38
rs145512436 Chr.14: 23638884-23638884 on Build GRCh38
rs747629991 Chr.14: 23639161-23639161 on Build GRCh38
rs932901464 Chr.14: 23638848-23638848 on Build GRCh38
rs764483650 Chr.14: 23638861-23638861 on Build GRCh38
rs1002161241 Chr.14: 23639130-23639130 on Build GRCh38
rs759810128 Chr.14: 23639054-23639054 on Build GRCh38
rs533508844 Chr.14: 23639009-23639009 on Build GRCh38
rs770240421 Chr.14: 23638976-23638976 on Build GRCh38
rs570863916 Chr.14: 23639003-23639003 on Build GRCh38
rs11558612 Chr.14: 23638950-23638950 on Build GRCh38
rs372389341 Chr.14: 23638963-23638963 on Build GRCh38
rs767456927 Chr.14: 23639055-23639055 on Build GRCh38
rs777237355 Chr.14: 23638935-23638935 on Build GRCh38
rs773894762 Chr.14: 23639035-23639035 on Build GRCh38
rs375239898 Chr.14: 23639160-23639160 on Build GRCh38
rs377066697 Chr.14: 23638862-23638862 on Build GRCh38
rs779809021 Chr.14: 23638967-23638967 on Build GRCh38
rs781236473 Chr.14: 23638944-23638944 on Build GRCh38
rs199794920 Chr.14: 23639005-23639005 on Build GRCh38
rs750429816 Chr.14: 23639139-23639139 on Build GRCh38
rs1004785290 Chr.14: 23639012-23639012 on Build GRCh38
rs778634375 Chr.14: 23639001-23639001 on Build GRCh38
rs754025097 Chr.14: 23638829-23638829 on Build GRCh38
rs772388017 Chr.14: 23639216-23639216 on Build GRCh38
rs774030550 Chr.14: 23638851-23638851 on Build GRCh38
rs565401225 Chr.14: 23639003-23639003 on Build GRCh38
rs931620407 Chr.14: 23638939-23638939 on Build GRCh38
rs897682767 Chr.14: 23639074-23639074 on Build GRCh38
rs983134978 Chr.14: 23638801-23638801 on Build GRCh38
rs767981116 Chr.14: 23639022-23639022 on Build GRCh38
rs965913400 Chr.14: 23639024-23639024 on Build GRCh38
rs370661385 Chr.14: 23639166-23639166 on Build GRCh38
rs773869769 Chr.14: 23638814-23638814 on Build GRCh38
rs376761935 Chr.14: 23638969-23638969 on Build GRCh38
rs1012609169 Chr.14: 23638782-23638782 on Build GRCh38
rs951900386 Chr.14: 23638800-23638800 on Build GRCh38
rs774379922 Chr.14: 23639052-23639052 on Build GRCh38
rs150047105 Chr.14: 23638883-23638883 on Build GRCh38
rs568333361 Chr.14: 23639150-23639150 on Build GRCh38
rs756027955 Chr.14: 23638888-23638888 on Build GRCh38
rs150935741 Chr.14: 23638952-23638952 on Build GRCh38
rs905240868 Chr.14: 23639100-23639100 on Build GRCh38

View Full Product Details