Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23639087-23639586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS2
Entrez Gene ID
10202
Gene Name
dehydrogenase/reductase 2
Gene Aliases
HEP27, SDR25C1
UniGene
Hs.272499
Gene Chromosome Location
Chr.14: 23630115-23645639 on Build GRCh38
Transcripts
NM_001318835.1 NM_005794.3 NM_182908.4 XM_005267249.1 XM_006720001.3 XM_011536338.1 XM_011536339.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7364284 c.190G>A p.V64I Chr.14: 23639228-23639228 on Build GRCh38
6643334 c.303G>A p.E101E Chr.14: 23639341-23639341 on Build GRCh38
4486414 c.304C>T p.Q102* Chr.14: 23639342-23639342 on Build GRCh38
5847888 c.178G>A p.G60R Chr.14: 23639216-23639216 on Build GRCh38
6486996 c.223C>T p.R75W Chr.14: 23639261-23639261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014224440 Chr.14: 23639524-23639524 on Build GRCh38
rs755277092 Chr.14: 23639400-23639400 on Build GRCh38
rs1042788206 Chr.14: 23639473-23639473 on Build GRCh38
rs916654680 Chr.14: 23639185-23639185 on Build GRCh38
rs760436099 Chr.14: 23639251-23639251 on Build GRCh38
rs999752054 Chr.14: 23639253-23639253 on Build GRCh38
rs758178912 Chr.14: 23639147-23639147 on Build GRCh38
rs560690909 Chr.14: 23639204-23639204 on Build GRCh38
rs768679039 Chr.14: 23639225-23639225 on Build GRCh38
rs998872575 Chr.14: 23639490-23639490 on Build GRCh38
rs772572636 Chr.14: 23639387-23639387 on Build GRCh38
rs146690475 Chr.14: 23639215-23639215 on Build GRCh38
rs992735040 Chr.14: 23639273-23639273 on Build GRCh38
rs775450619 Chr.14: 23639317-23639317 on Build GRCh38
rs745770459 Chr.14: 23639362-23639362 on Build GRCh38
rs199596675 Chr.14: 23639198-23639198 on Build GRCh38
rs1024198654 Chr.14: 23639372-23639372 on Build GRCh38
rs138205531 Chr.14: 23639194-23639194 on Build GRCh38
rs770867443 Chr.14: 23639232-23639232 on Build GRCh38
rs771136730 Chr.14: 23639172-23639172 on Build GRCh38
rs111785187 Chr.14: 23639302-23639302 on Build GRCh38
rs758924656 Chr.14: 23639338-23639338 on Build GRCh38
rs4147558 Chr.14: 23639327-23639327 on Build GRCh38
rs199867663 Chr.14: 23639197-23639197 on Build GRCh38
rs761380169 Chr.14: 23639373-23639373 on Build GRCh38
rs576958094 Chr.14: 23639182-23639182 on Build GRCh38
rs776711817 Chr.14: 23639227-23639227 on Build GRCh38
rs1009802405 Chr.14: 23639180-23639180 on Build GRCh38
rs147534556 Chr.14: 23639262-23639262 on Build GRCh38
rs993565211 Chr.14: 23639557-23639557 on Build GRCh38
rs45608041 Chr.14: 23639128-23639128 on Build GRCh38
rs761844165 Chr.14: 23639231-23639231 on Build GRCh38
rs74036809 Chr.14: 23639210-23639210 on Build GRCh38
rs766035204 Chr.14: 23639196-23639196 on Build GRCh38
rs2164865 Chr.14: 23639558-23639558 on Build GRCh38
rs375920436 Chr.14: 23639246-23639246 on Build GRCh38
rs756150583 Chr.14: 23639359-23639359 on Build GRCh38
rs534138708 Chr.14: 23639416-23639416 on Build GRCh38
rs139039851 Chr.14: 23639241-23639241 on Build GRCh38
rs575683380 Chr.14: 23639437-23639437 on Build GRCh38
rs192649432 Chr.14: 23639425-23639425 on Build GRCh38
rs777744624 Chr.14: 23639361-23639361 on Build GRCh38
rs199641343 Chr.14: 23639222-23639222 on Build GRCh38
rs774284809 Chr.14: 23639303-23639303 on Build GRCh38
rs767775957 Chr.14: 23639405-23639405 on Build GRCh38
rs568373479 Chr.14: 23639378-23639378 on Build GRCh38
rs754678449 Chr.14: 23639159-23639159 on Build GRCh38
rs200598513 Chr.14: 23639183-23639183 on Build GRCh38
rs141370810 Chr.14: 23639321-23639321 on Build GRCh38
rs888979561 Chr.14: 23639326-23639326 on Build GRCh38
rs375928308 Chr.14: 23639169-23639169 on Build GRCh38
rs533750929 Chr.14: 23639153-23639153 on Build GRCh38
rs747248253 Chr.14: 23639224-23639224 on Build GRCh38
rs368578886 Chr.14: 23639263-23639263 on Build GRCh38
rs45599632 Chr.14: 23639288-23639288 on Build GRCh38
rs78136100 Chr.14: 23639526-23639526 on Build GRCh38
rs555965971 Chr.14: 23639200-23639200 on Build GRCh38
rs762765287 Chr.14: 23639162-23639162 on Build GRCh38
rs993256569 Chr.14: 23639546-23639546 on Build GRCh38
rs539243409 Chr.14: 23639430-23639430 on Build GRCh38
rs149593009 Chr.14: 23639202-23639202 on Build GRCh38
rs554920203 Chr.14: 23639121-23639121 on Build GRCh38
rs762976156 Chr.14: 23639195-23639195 on Build GRCh38
rs947214396 Chr.14: 23639468-23639468 on Build GRCh38
rs201278137 Chr.14: 23639240-23639240 on Build GRCh38
rs759167374 Chr.14: 23639293-23639294 on Build GRCh38
rs140244120 Chr.14: 23639270-23639270 on Build GRCh38
rs373176822 Chr.14: 23639168-23639168 on Build GRCh38
rs1023841039 Chr.14: 23639278-23639278 on Build GRCh38
rs752461530 Chr.14: 23639384-23639385 on Build GRCh38
rs372014670 Chr.14: 23639506-23639506 on Build GRCh38
rs201771941 Chr.14: 23639318-23639318 on Build GRCh38
rs760741798 Chr.14: 23639190-23639190 on Build GRCh38
rs919141891 Chr.14: 23639223-23639223 on Build GRCh38
rs1031266798 Chr.14: 23639258-23639258 on Build GRCh38
rs982461659 Chr.14: 23639559-23639559 on Build GRCh38
rs1057190056 Chr.14: 23639493-23639493 on Build GRCh38
rs781429061 Chr.14: 23639291-23639291 on Build GRCh38
rs1006340045 Chr.14: 23639547-23639547 on Build GRCh38
rs963084013 Chr.14: 23639527-23639527 on Build GRCh38
rs560554655 Chr.14: 23639432-23639432 on Build GRCh38
rs895499911 Chr.14: 23639499-23639499 on Build GRCh38
rs780585174 Chr.14: 23639339-23639339 on Build GRCh38
rs752118286 Chr.14: 23639399-23639399 on Build GRCh38
rs113010522 Chr.14: 23639129-23639129 on Build GRCh38
rs984805511 Chr.14: 23639415-23639415 on Build GRCh38
rs201882888 Chr.14: 23639272-23639272 on Build GRCh38
rs777096673 Chr.14: 23639371-23639371 on Build GRCh38
rs547375774 Chr.14: 23639418-23639418 on Build GRCh38
rs61745475 Chr.14: 23639314-23639314 on Build GRCh38
rs767193133 Chr.14: 23639199-23639199 on Build GRCh38
rs780425610 Chr.14: 23639279-23639279 on Build GRCh38
rs910349975 Chr.14: 23639458-23639458 on Build GRCh38
rs779338474 Chr.14: 23639211-23639211 on Build GRCh38
rs759930024 Chr.14: 23639463-23639463 on Build GRCh38
rs771011408 Chr.14: 23639256-23639267 on Build GRCh38
rs1016305207 Chr.14: 23639424-23639424 on Build GRCh38
rs746029038 Chr.14: 23639179-23639179 on Build GRCh38
rs143275916 Chr.14: 23639261-23639261 on Build GRCh38
rs545604056 Chr.14: 23639388-23639388 on Build GRCh38
rs774811528 Chr.14: 23639177-23639177 on Build GRCh38
rs747629991 Chr.14: 23639161-23639161 on Build GRCh38
rs759395931 Chr.14: 23639312-23639312 on Build GRCh38
rs1028944554 Chr.14: 23639300-23639300 on Build GRCh38
rs1002161241 Chr.14: 23639130-23639130 on Build GRCh38
rs766862965 Chr.14: 23639389-23639389 on Build GRCh38
rs1016418783 Chr.14: 23639553-23639553 on Build GRCh38
rs763650696 Chr.14: 23639254-23639254 on Build GRCh38
rs750878707 Chr.14: 23639337-23639337 on Build GRCh38
rs375787784 Chr.14: 23639370-23639370 on Build GRCh38
rs529188012 Chr.14: 23639361-23639361 on Build GRCh38
rs962087126 Chr.14: 23639554-23639554 on Build GRCh38
rs375239898 Chr.14: 23639160-23639160 on Build GRCh38
rs774405927 Chr.14: 23639257-23639283 on Build GRCh38
rs781461354 Chr.14: 23639355-23639355 on Build GRCh38
rs150243637 Chr.14: 23639500-23639500 on Build GRCh38
rs889978771 Chr.14: 23639511-23639511 on Build GRCh38
rs750429816 Chr.14: 23639139-23639139 on Build GRCh38
rs772388017 Chr.14: 23639216-23639216 on Build GRCh38
rs998446362 Chr.14: 23639296-23639296 on Build GRCh38
rs868502466 Chr.14: 23639491-23639491 on Build GRCh38
rs201826965 Chr.14: 23639356-23639356 on Build GRCh38
rs370661385 Chr.14: 23639166-23639166 on Build GRCh38
rs938128840 Chr.14: 23639255-23639255 on Build GRCh38
rs568333361 Chr.14: 23639150-23639150 on Build GRCh38
rs750953197 Chr.14: 23639264-23639264 on Build GRCh38
rs549350739 Chr.14: 23639336-23639336 on Build GRCh38
rs765585817 Chr.14: 23639520-23639520 on Build GRCh38
rs751886725 Chr.14: 23639346-23639346 on Build GRCh38
rs933216908 Chr.14: 23639435-23639435 on Build GRCh38
rs554982211 Chr.14: 23639516-23639516 on Build GRCh38
rs562246921 Chr.14: 23639301-23639301 on Build GRCh38
rs1026432420 Chr.14: 23639556-23639556 on Build GRCh38
rs902871296 Chr.14: 23639248-23639248 on Build GRCh38
rs755399377 Chr.14: 23639348-23639348 on Build GRCh38

View Full Product Details