Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23990158-23990668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS4L2
Entrez Gene ID
317749
Gene Name
dehydrogenase/reductase 4 like 2
Gene Aliases
SDR25C3
UniGene
Hs.743442
Gene Chromosome Location
Chr.14: 23969874-24006408 on Build GRCh38
Transcripts
NM_001193635.1 NM_001193636.1 NM_001193637.1 NM_198083.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3983845 c.230A>G p.E77G Chr.14: 23990289-23990289 on Build GRCh38
3931912 c.253G>A p.G85S Chr.14: 23990306-23990306 on Build GRCh38
4958201 c.221C>T p.A74V Chr.14: 23990274-23990274 on Build GRCh38
1748769 c.253G>A p.G85S Chr.14: 23990306-23990306 on Build GRCh38
4539627 c.270G>T p.V90V Chr.14: 23990323-23990323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764699848 Chr.14: 23990243-23990243 on Build GRCh38
rs768424309 Chr.14: 23990192-23990192 on Build GRCh38
rs60865462 Chr.14: 23990284-23990284 on Build GRCh38
rs759748439 Chr.14: 23990354-23990354 on Build GRCh38
rs529238562 Chr.14: 23990232-23990232 on Build GRCh38
rs895481214 Chr.14: 23990473-23990473 on Build GRCh38
rs111637036 Chr.14: 23990339-23990339 on Build GRCh38
rs768484737 Chr.14: 23990345-23990345 on Build GRCh38
rs771245455 Chr.14: 23990186-23990186 on Build GRCh38
rs771254883 Chr.14: 23990351-23990351 on Build GRCh38
rs371577743 Chr.14: 23990394-23990394 on Build GRCh38
rs776057464 Chr.14: 23990318-23990318 on Build GRCh38
rs146647822 Chr.14: 23990204-23990204 on Build GRCh38
rs1056682417 Chr.14: 23990456-23990456 on Build GRCh38
rs371338153 Chr.14: 23990185-23990185 on Build GRCh38
rs758455757 Chr.14: 23990371-23990371 on Build GRCh38
rs979641087 Chr.14: 23990426-23990426 on Build GRCh38
rs45583341 Chr.14: 23990311-23990311 on Build GRCh38
rs775071926 Chr.14: 23990276-23990276 on Build GRCh38
rs568460062 Chr.14: 23990191-23990191 on Build GRCh38
rs758295909 Chr.14: 23990342-23990342 on Build GRCh38
rs1052728921 Chr.14: 23990296-23990296 on Build GRCh38
rs777383608 Chr.14: 23990404-23990404 on Build GRCh38
rs761559403 Chr.14: 23990195-23990195 on Build GRCh38
rs769548882 Chr.14: 23990197-23990197 on Build GRCh38
rs144494372 Chr.14: 23990648-23990648 on Build GRCh38
rs142348998 Chr.14: 23990333-23990333 on Build GRCh38
rs747619043 Chr.14: 23990346-23990346 on Build GRCh38
rs766123637 Chr.14: 23990201-23990201 on Build GRCh38
rs145720846 Chr.14: 23990187-23990187 on Build GRCh38
rs368172750 Chr.14: 23990194-23990194 on Build GRCh38
rs1005770215 Chr.14: 23990528-23990528 on Build GRCh38
rs757199283 Chr.14: 23990301-23990301 on Build GRCh38
rs780097678 Chr.14: 23990258-23990258 on Build GRCh38
rs765615694 Chr.14: 23990286-23990286 on Build GRCh38
rs748058225 Chr.14: 23990228-23990228 on Build GRCh38
rs542907863 Chr.14: 23990332-23990332 on Build GRCh38
rs773562364 Chr.14: 23990328-23990328 on Build GRCh38
rs769783380 Chr.14: 23990229-23990229 on Build GRCh38
rs61732730 Chr.14: 23990198-23990198 on Build GRCh38
rs758645235 Chr.14: 23990291-23990291 on Build GRCh38
rs905631649 Chr.14: 23990501-23990501 on Build GRCh38
rs150079702 Chr.14: 23990246-23990246 on Build GRCh38
rs774755632 Chr.14: 23990352-23990352 on Build GRCh38
rs767636947 Chr.14: 23990357-23990357 on Build GRCh38
rs557426954 Chr.14: 23990304-23990304 on Build GRCh38
rs762582832 Chr.14: 23990200-23990200 on Build GRCh38
rs762271415 Chr.14: 23990285-23990285 on Build GRCh38
rs752311258 Chr.14: 23990240-23990240 on Build GRCh38
rs755074388 Chr.14: 23990297-23990297 on Build GRCh38
rs1044520353 Chr.14: 23990488-23990488 on Build GRCh38
rs750631907 Chr.14: 23990251-23990251 on Build GRCh38
rs758600997 Chr.14: 23990254-23990254 on Build GRCh38
rs61729875 Chr.14: 23990266-23990266 on Build GRCh38
rs549660919 Chr.14: 23990555-23990555 on Build GRCh38
rs747646699 Chr.14: 23990388-23990388 on Build GRCh38
rs754318476 Chr.14: 23990222-23990222 on Build GRCh38
rs762372561 Chr.14: 23990319-23990319 on Build GRCh38
rs145901014 Chr.14: 23990202-23990202 on Build GRCh38
rs184019002 Chr.14: 23990358-23990358 on Build GRCh38
rs897034095 Chr.14: 23990571-23990571 on Build GRCh38
rs773615402 Chr.14: 23990348-23990348 on Build GRCh38
rs139813039 Chr.14: 23990277-23990277 on Build GRCh38
rs770433652 Chr.14: 23990233-23990233 on Build GRCh38
rs61729874 Chr.14: 23990268-23990268 on Build GRCh38
rs745882324 Chr.14: 23990226-23990226 on Build GRCh38
rs1016783942 Chr.14: 23990562-23990562 on Build GRCh38
rs45596131 Chr.14: 23990305-23990305 on Build GRCh38
rs779986156 Chr.14: 23990376-23990376 on Build GRCh38
rs571000190 Chr.14: 23990205-23990205 on Build GRCh38
rs148736302 Chr.14: 23990331-23990331 on Build GRCh38
rs751582343 Chr.14: 23990294-23990294 on Build GRCh38
rs994083019 Chr.14: 23990581-23990581 on Build GRCh38
rs112853904 Chr.14: 23990287-23990287 on Build GRCh38
rs985500168 Chr.14: 23990630-23990630 on Build GRCh38
rs1014420446 Chr.14: 23990324-23990324 on Build GRCh38
rs376416180 Chr.14: 23990270-23990270 on Build GRCh38
rs924162909 Chr.14: 23990447-23990447 on Build GRCh38
rs955541087 Chr.14: 23990607-23990607 on Build GRCh38
rs761257997 Chr.14: 23990241-23990241 on Build GRCh38
rs147428238 Chr.14: 23990269-23990269 on Build GRCh38
rs754193813 Chr.14: 23990244-23990244 on Build GRCh38
rs1029616993 Chr.14: 23990599-23990599 on Build GRCh38
rs143357796 Chr.14: 23990224-23990224 on Build GRCh38
rs376916778 Chr.14: 23990363-23990363 on Build GRCh38
rs11556286 Chr.14: 23990227-23990227 on Build GRCh38
rs768245766 Chr.14: 23990314-23990314 on Build GRCh38
rs527674493 Chr.14: 23990338-23990338 on Build GRCh38
rs760690570 Chr.14: 23990361-23990361 on Build GRCh38
rs767570135 Chr.14: 23990335-23990335 on Build GRCh38
rs777648090 Chr.14: 23990299-23990299 on Build GRCh38
rs550075024 Chr.14: 23990383-23990383 on Build GRCh38
rs45579231 Chr.14: 23990234-23990234 on Build GRCh38
rs111455317 Chr.14: 23990288-23990288 on Build GRCh38
rs749227142 Chr.14: 23990300-23990300 on Build GRCh38
rs781071784 Chr.14: 23990222-23990223 on Build GRCh38
rs779206377 Chr.14: 23990225-23990225 on Build GRCh38
rs763265273 Chr.14: 23990350-23990350 on Build GRCh38
rs766594917 Chr.14: 23990293-23990293 on Build GRCh38
rs935590254 Chr.14: 23990454-23990454 on Build GRCh38
rs772988441 Chr.14: 23990231-23990231 on Build GRCh38
rs1044354435 Chr.14: 23990271-23990271 on Build GRCh38
rs746642424 Chr.14: 23990344-23990344 on Build GRCh38
rs780725091 Chr.14: 23990384-23990384 on Build GRCh38
rs371611892 Chr.14: 23990262-23990262 on Build GRCh38
rs949776148 Chr.14: 23990487-23990487 on Build GRCh38
rs747160452 Chr.14: 23990260-23990260 on Build GRCh38
rs535728514 Chr.14: 23990457-23990457 on Build GRCh38
rs370762608 Chr.14: 23990379-23990379 on Build GRCh38
rs45534140 Chr.14: 23990219-23990219 on Build GRCh38
rs563512511 Chr.14: 23990320-23990320 on Build GRCh38
rs756732860 Chr.14: 23990218-23990218 on Build GRCh38
rs750181122 Chr.14: 23990340-23990340 on Build GRCh38
rs764644383 Chr.14: 23990278-23990278 on Build GRCh38

View Full Product Details