Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24036163-24036704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS4L1
Entrez Gene ID
728635
Gene Name
dehydrogenase/reductase 4 like 1
Gene Aliases
SDR25C4
UniGene
Hs.647569
Gene Chromosome Location
Chr.14: 24007009-24051377 on Build GRCh38
Transcripts
NM_001277864.1 NR_102687.1 NR_102688.1 NR_102689.1 NR_102691.1 NR_102692.1 NR_102693.1 NR_102694.1 XM_006720246.3

SNP Detail

SNP ID Chromosome Location
rs770126429 Chr.14: 24036567-24036567 on Build GRCh38
rs766524653 Chr.14: 24036500-24036500 on Build GRCh38
rs11158517 Chr.14: 24036259-24036259 on Build GRCh38
rs772832397 Chr.14: 24036641-24036641 on Build GRCh38
rs748693863 Chr.14: 24036566-24036566 on Build GRCh38
rs752558831 Chr.14: 24036505-24036505 on Build GRCh38
rs746246711 Chr.14: 24036461-24036462 on Build GRCh38
rs761899832 Chr.14: 24036583-24036583 on Build GRCh38
rs757824244 Chr.14: 24036517-24036517 on Build GRCh38
rs746285146 Chr.14: 24036470-24036470 on Build GRCh38
rs1021296841 Chr.14: 24036399-24036399 on Build GRCh38
rs886743712 Chr.14: 24036364-24036364 on Build GRCh38
rs560113625 Chr.14: 24036343-24036343 on Build GRCh38
rs769865035 Chr.14: 24036639-24036639 on Build GRCh38
rs558127076 Chr.14: 24036468-24036468 on Build GRCh38
rs1009356169 Chr.14: 24036527-24036527 on Build GRCh38
rs4982848 Chr.14: 24036199-24036199 on Build GRCh38
rs528061527 Chr.14: 24036375-24036375 on Build GRCh38
rs2332131 Chr.14: 24036349-24036349 on Build GRCh38
rs957935354 Chr.14: 24036254-24036254 on Build GRCh38
rs536063095 Chr.14: 24036419-24036419 on Build GRCh38
rs772300051 Chr.14: 24036534-24036535 on Build GRCh38
rs557897654 Chr.14: 24036250-24036250 on Build GRCh38
rs746540852 Chr.14: 24036553-24036553 on Build GRCh38
rs771016169 Chr.14: 24036614-24036614 on Build GRCh38
rs778735858 Chr.14: 24036515-24036515 on Build GRCh38
rs3049729 Chr.14: 24036343-24036348 on Build GRCh38
rs982183180 Chr.14: 24036318-24036318 on Build GRCh38
rs777045662 Chr.14: 24036478-24036478 on Build GRCh38
rs781688104 Chr.14: 24036561-24036561 on Build GRCh38
rs1040776304 Chr.14: 24036367-24036367 on Build GRCh38
rs746121325 Chr.14: 24036621-24036621 on Build GRCh38
rs2332129 Chr.14: 24036274-24036274 on Build GRCh38
rs371755645 Chr.14: 24036593-24036593 on Build GRCh38
rs377708297 Chr.14: 24036494-24036494 on Build GRCh38
rs779363016 Chr.14: 24036552-24036552 on Build GRCh38
rs769964703 Chr.14: 24036529-24036529 on Build GRCh38
rs2332128 Chr.14: 24036272-24036272 on Build GRCh38
rs551341921 Chr.14: 24036376-24036376 on Build GRCh38
rs557180142 Chr.14: 24036518-24036518 on Build GRCh38
rs372557477 Chr.14: 24036580-24036580 on Build GRCh38
rs753193433 Chr.14: 24036606-24036606 on Build GRCh38
rs766129186 Chr.14: 24036644-24036644 on Build GRCh38
rs926702227 Chr.14: 24036319-24036319 on Build GRCh38
rs758135635 Chr.14: 24036549-24036549 on Build GRCh38
rs577988643 Chr.14: 24036510-24036510 on Build GRCh38
rs2332127 Chr.14: 24036184-24036184 on Build GRCh38
rs768888468 Chr.14: 24036476-24036476 on Build GRCh38
rs759144681 Chr.14: 24036339-24036339 on Build GRCh38
rs938165887 Chr.14: 24036321-24036321 on Build GRCh38
rs761775336 Chr.14: 24036638-24036638 on Build GRCh38
rs779161775 Chr.14: 24036615-24036615 on Build GRCh38
rs200625178 Chr.14: 24036526-24036526 on Build GRCh38
rs2332130 Chr.14: 24036328-24036328 on Build GRCh38
rs556461153 Chr.14: 24036531-24036531 on Build GRCh38
rs750086786 Chr.14: 24036545-24036545 on Build GRCh38
rs139393893 Chr.14: 24036431-24036431 on Build GRCh38
rs538571970 Chr.14: 24036455-24036455 on Build GRCh38
rs2332132 Chr.14: 24036358-24036358 on Build GRCh38
rs543261626 Chr.14: 24036539-24036539 on Build GRCh38
rs764112210 Chr.14: 24036540-24036540 on Build GRCh38
rs899627308 Chr.14: 24036237-24036237 on Build GRCh38
rs192081772 Chr.14: 24036356-24036356 on Build GRCh38
rs959704337 Chr.14: 24036420-24036420 on Build GRCh38
rs773628346 Chr.14: 24036572-24036572 on Build GRCh38
rs113048202 Chr.14: 24036676-24036676 on Build GRCh38
rs971168721 Chr.14: 24036300-24036300 on Build GRCh38
rs773227607 Chr.14: 24036488-24036488 on Build GRCh38
rs759690086 Chr.14: 24036579-24036579 on Build GRCh38
rs563895992 Chr.14: 24036280-24036280 on Build GRCh38
rs371416349 Chr.14: 24036508-24036508 on Build GRCh38
rs765168101 Chr.14: 24036543-24036543 on Build GRCh38
rs374626236 Chr.14: 24036542-24036542 on Build GRCh38
rs766359960 Chr.14: 24036535-24036535 on Build GRCh38
rs578027799 Chr.14: 24036257-24036257 on Build GRCh38
rs1009429484 Chr.14: 24036397-24036397 on Build GRCh38
rs943688483 Chr.14: 24036499-24036499 on Build GRCh38
rs1032190210 Chr.14: 24036253-24036253 on Build GRCh38
rs371481650 Chr.14: 24036410-24036410 on Build GRCh38
rs866741302 Chr.14: 24036434-24036434 on Build GRCh38
rs762839215 Chr.14: 24036643-24036643 on Build GRCh38
rs530628480 Chr.14: 24036400-24036400 on Build GRCh38
rs8006288 Chr.14: 24036405-24036405 on Build GRCh38
rs566915788 Chr.14: 24036680-24036680 on Build GRCh38
rs71405834 Chr.14: 24036355-24036355 on Build GRCh38
rs369681534 Chr.14: 24036658-24036658 on Build GRCh38
rs1020906593 Chr.14: 24036281-24036281 on Build GRCh38
rs374800603 Chr.14: 24036598-24036598 on Build GRCh38
rs756918584 Chr.14: 24036514-24036514 on Build GRCh38
rs762453573 Chr.14: 24036591-24036591 on Build GRCh38
rs776684254 Chr.14: 24036582-24036582 on Build GRCh38
rs368624982 Chr.14: 24036613-24036613 on Build GRCh38
rs564757951 Chr.14: 24036390-24036390 on Build GRCh38
rs573556548 Chr.14: 24036575-24036575 on Build GRCh38
rs997101831 Chr.14: 24036244-24036244 on Build GRCh38
rs763289020 Chr.14: 24036534-24036534 on Build GRCh38
rs745391003 Chr.14: 24036458-24036458 on Build GRCh38
rs8005834 Chr.14: 24036513-24036513 on Build GRCh38
rs375426884 Chr.14: 24036633-24036633 on Build GRCh38
rs943705445 Chr.14: 24036366-24036366 on Build GRCh38
rs748536295 Chr.14: 24036528-24036528 on Build GRCh38
rs774244551 Chr.14: 24036577-24036577 on Build GRCh38
rs769358913 Chr.14: 24036423-24036423 on Build GRCh38
rs759247088 Chr.14: 24036677-24036677 on Build GRCh38
rs1019791579 Chr.14: 24036201-24036201 on Build GRCh38
rs529522973 Chr.14: 24036282-24036282 on Build GRCh38
rs566410839 Chr.14: 24036432-24036432 on Build GRCh38
rs574260536 Chr.14: 24036520-24036520 on Build GRCh38
rs761955476 Chr.14: 24036483-24036483 on Build GRCh38
rs772588010 Chr.14: 24036460-24036460 on Build GRCh38
rs765460680 Chr.14: 24036485-24036485 on Build GRCh38
rs970712559 Chr.14: 24036403-24036403 on Build GRCh38
rs775826084 Chr.14: 24036463-24036463 on Build GRCh38
rs771190757 Chr.14: 24036576-24036576 on Build GRCh38
rs780730404 Chr.14: 24036600-24036600 on Build GRCh38
rs145675272 Chr.14: 24036555-24036555 on Build GRCh38
rs756685151 Chr.14: 24036607-24036607 on Build GRCh38
rs973556999 Chr.14: 24036322-24036322 on Build GRCh38
rs929544873 Chr.14: 24036341-24036341 on Build GRCh38
rs994888893 Chr.14: 24036620-24036620 on Build GRCh38
rs765118978 Chr.14: 24036586-24036586 on Build GRCh38
rs111872405 Chr.14: 24036427-24036427 on Build GRCh38
rs372588807 Chr.14: 24036296-24036297 on Build GRCh38
rs5002068 Chr.14: 24036214-24036214 on Build GRCh38
rs369829744 Chr.14: 24036669-24036669 on Build GRCh38
rs2277476 Chr.14: 24036627-24036627 on Build GRCh38
rs1003487358 Chr.14: 24036574-24036574 on Build GRCh38
rs774560123 Chr.14: 24036538-24036538 on Build GRCh38

View Full Product Details