Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24132512-24133015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FITM1
Entrez Gene ID
161247
Gene Name
fat storage inducing transmembrane protein 1
Gene Aliases
FIT1
UniGene
Hs.128060
Gene Chromosome Location
Chr.14: 24131466-24132849 on Build GRCh38
Transcripts
NM_203402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7146137 c.860C>T p.S287F Chr.14: 24132804-24132804 on Build GRCh38
1369292 c.870G>A p.K290K Chr.14: 24132814-24132814 on Build GRCh38
363256 c.793G>T p.G265C Chr.14: 24132737-24132737 on Build GRCh38
3700846 c.575T>G p.F192C Chr.14: 24132519-24132519 on Build GRCh38
7080182 c.776C>T p.A259V Chr.14: 24132720-24132720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910923207 Chr.14: 24132791-24132791 on Build GRCh38
rs909608371 Chr.14: 24132549-24132549 on Build GRCh38
rs748718994 Chr.14: 24132779-24132779 on Build GRCh38
rs768483415 Chr.14: 24132754-24132776 on Build GRCh38
rs755780956 Chr.14: 24132773-24132773 on Build GRCh38
rs752572871 Chr.14: 24132771-24132771 on Build GRCh38
rs144083663 Chr.14: 24132701-24132701 on Build GRCh38
rs769576186 Chr.14: 24132664-24132664 on Build GRCh38
rs770544820 Chr.14: 24132780-24132780 on Build GRCh38
rs776535061 Chr.14: 24132911-24132912 on Build GRCh38
rs142215470 Chr.14: 24132963-24132963 on Build GRCh38
rs779163531 Chr.14: 24132744-24132744 on Build GRCh38
rs772794305 Chr.14: 24132597-24132597 on Build GRCh38
rs570159546 Chr.14: 24132599-24132599 on Build GRCh38
rs917491823 Chr.14: 24132614-24132614 on Build GRCh38
rs768301865 Chr.14: 24132588-24132589 on Build GRCh38
rs150217465 Chr.14: 24132795-24132795 on Build GRCh38
rs917453068 Chr.14: 24132862-24132862 on Build GRCh38
rs746849731 Chr.14: 24132559-24132559 on Build GRCh38
rs759946490 Chr.14: 24132607-24132607 on Build GRCh38
rs760434431 Chr.14: 24132647-24132647 on Build GRCh38
rs745847677 Chr.14: 24132554-24132554 on Build GRCh38
rs768737140 Chr.14: 24132796-24132796 on Build GRCh38
rs575266836 Chr.14: 24132794-24132794 on Build GRCh38
rs765309579 Chr.14: 24132955-24132955 on Build GRCh38
rs1023632109 Chr.14: 24132906-24132906 on Build GRCh38
rs776951089 Chr.14: 24132801-24132801 on Build GRCh38
rs140970200 Chr.14: 24132764-24132764 on Build GRCh38
rs753811686 Chr.14: 24132706-24132706 on Build GRCh38
rs11574505 Chr.14: 24132611-24132611 on Build GRCh38
rs753839305 Chr.14: 24132735-24132735 on Build GRCh38
rs1054425285 Chr.14: 24132966-24132966 on Build GRCh38
rs745411899 Chr.14: 24132871-24132871 on Build GRCh38
rs771324675 Chr.14: 24132683-24132683 on Build GRCh38
rs746827990 Chr.14: 24132747-24132749 on Build GRCh38
rs763653375 Chr.14: 24132830-24132830 on Build GRCh38
rs748097132 Chr.14: 24132830-24132831 on Build GRCh38
rs371806629 Chr.14: 24132704-24132704 on Build GRCh38
rs140419269 Chr.14: 24132669-24132669 on Build GRCh38
rs373700463 Chr.14: 24132652-24132652 on Build GRCh38
rs148679814 Chr.14: 24132703-24132703 on Build GRCh38
rs546259053 Chr.14: 24132863-24132863 on Build GRCh38
rs965197578 Chr.14: 24132680-24132680 on Build GRCh38
rs769487798 Chr.14: 24132589-24132589 on Build GRCh38
rs370542722 Chr.14: 24132600-24132600 on Build GRCh38
rs146750862 Chr.14: 24132662-24132662 on Build GRCh38
rs775322986 Chr.14: 24132583-24132583 on Build GRCh38
rs776499263 Chr.14: 24132830-24132830 on Build GRCh38
rs374523882 Chr.14: 24132785-24132785 on Build GRCh38
rs747656626 Chr.14: 24132587-24132587 on Build GRCh38
rs897402807 Chr.14: 24132873-24132888 on Build GRCh38
rs752979100 Chr.14: 24132612-24132612 on Build GRCh38
rs761810676 Chr.14: 24132818-24132818 on Build GRCh38
rs950203157 Chr.14: 24132901-24132901 on Build GRCh38
rs760727118 Chr.14: 24132618-24132618 on Build GRCh38
rs541381673 Chr.14: 24132802-24132802 on Build GRCh38
rs972886438 Chr.14: 24132857-24132857 on Build GRCh38
rs752521570 Chr.14: 24132811-24132811 on Build GRCh38
rs758226855 Chr.14: 24132629-24132629 on Build GRCh38
rs780060685 Chr.14: 24132650-24132650 on Build GRCh38
rs143975707 Chr.14: 24132548-24132548 on Build GRCh38
rs774966849 Chr.14: 24132686-24132686 on Build GRCh38
rs923505200 Chr.14: 24132981-24132981 on Build GRCh38
rs1012210659 Chr.14: 24132903-24132903 on Build GRCh38
rs1044614802 Chr.14: 24132640-24132640 on Build GRCh38
rs531850176 Chr.14: 24132944-24132944 on Build GRCh38
rs980340097 Chr.14: 24132921-24132921 on Build GRCh38
rs776470418 Chr.14: 24132581-24132581 on Build GRCh38
rs766196340 Chr.14: 24132733-24132733 on Build GRCh38
rs772869419 Chr.14: 24132838-24132838 on Build GRCh38
rs369314331 Chr.14: 24132899-24132899 on Build GRCh38
rs535259645 Chr.14: 24132700-24132700 on Build GRCh38
rs117284518 Chr.14: 24132556-24132556 on Build GRCh38
rs777532782 Chr.14: 24132776-24132776 on Build GRCh38
rs562816366 Chr.14: 24132910-24132910 on Build GRCh38
rs765466271 Chr.14: 24132762-24132762 on Build GRCh38
rs759507646 Chr.14: 24132809-24132809 on Build GRCh38
rs577200461 Chr.14: 24132855-24132855 on Build GRCh38
rs755980705 Chr.14: 24132826-24132826 on Build GRCh38
rs142074658 Chr.14: 24132623-24132623 on Build GRCh38
rs199842859 Chr.14: 24132724-24132724 on Build GRCh38
rs781068396 Chr.14: 24132768-24132768 on Build GRCh38
rs762001311 Chr.14: 24132732-24132732 on Build GRCh38
rs773430809 Chr.14: 24132803-24132803 on Build GRCh38
rs999370240 Chr.14: 24132586-24132586 on Build GRCh38
rs962164629 Chr.14: 24132960-24132960 on Build GRCh38
rs187467245 Chr.14: 24132840-24132840 on Build GRCh38
rs759885341 Chr.14: 24132697-24132697 on Build GRCh38
rs753954858 Chr.14: 24132627-24132627 on Build GRCh38
rs757503988 Chr.14: 24132609-24132610 on Build GRCh38
rs768374310 Chr.14: 24132562-24132562 on Build GRCh38
rs556012970 Chr.14: 24132661-24132661 on Build GRCh38
rs902312537 Chr.14: 24132557-24132557 on Build GRCh38
rs367884290 Chr.14: 24132666-24132666 on Build GRCh38
rs769793809 Chr.14: 24132835-24132836 on Build GRCh38
rs554195528 Chr.14: 24132810-24132810 on Build GRCh38
rs751572758 Chr.14: 24132757-24132757 on Build GRCh38
rs774533864 Chr.14: 24132604-24132604 on Build GRCh38
rs754860068 Chr.14: 24132657-24132657 on Build GRCh38
rs1015397596 Chr.14: 24132934-24132934 on Build GRCh38
rs1000129518 Chr.14: 24132653-24132653 on Build GRCh38

View Full Product Details