Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24232166-24232672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPR2
Entrez Gene ID
51292
Gene Name
guanosine monophosphate reductase 2
Gene Aliases
GMPR 2
UniGene
Hs.368855
Gene Chromosome Location
Chr.14: 24232417-24239242 on Build GRCh38
Transcripts
NM_001002000.2 NM_001002001.2 NM_001283021.1 NR_104265.1 XM_005267740.3 XM_005267741.3 XM_017021356.1 XM_017021357.1 XM_017021358.1 NM_006156.2

Gene Symbol
NEDD8
Entrez Gene ID
4738
Gene Name
neural precursor cell expressed, developmentally down-regulated 8
Gene Aliases
NEDD-8
UniGene
Hs.531064
Gene Chromosome Location
Chr.14: 24216851-24232370 on Build GRCh38
Transcripts
NM_001002000.2 NM_001002001.2 NM_001283021.1 NR_104265.1 XM_005267740.3 XM_005267741.3 XM_017021356.1 XM_017021357.1 XM_017021358.1 NM_006156.2

Gene Symbol
NEDD8-MDP1
Entrez Gene ID
100528064
Gene Name
NEDD8-MDP1 readthrough
Gene Aliases
-
UniGene
Hs.220963
Gene Chromosome Location
Chr.14: 24213937-24232370 on Build GRCh38
Transcripts
NM_001002000.2 NM_001002001.2 NM_001283021.1 NR_104265.1 XM_005267740.3 XM_005267741.3 XM_017021356.1 XM_017021357.1 XM_017021358.1 NM_006156.2

SNP Detail

SNP ID Chromosome Location
rs936668451 Chr.14: 24232457-24232457 on Build GRCh38
rs547890052 Chr.14: 24232469-24232469 on Build GRCh38
rs765959352 Chr.14: 24232280-24232280 on Build GRCh38
rs777133918 Chr.14: 24232639-24232639 on Build GRCh38
rs1028809691 Chr.14: 24232608-24232609 on Build GRCh38
rs532687339 Chr.14: 24232393-24232393 on Build GRCh38
rs199870597 Chr.14: 24232395-24232396 on Build GRCh38
rs961733933 Chr.14: 24232407-24232407 on Build GRCh38
rs773548274 Chr.14: 24232346-24232346 on Build GRCh38
rs879115110 Chr.14: 24232409-24232409 on Build GRCh38
rs940877515 Chr.14: 24232191-24232191 on Build GRCh38
rs901793409 Chr.14: 24232317-24232317 on Build GRCh38
rs200996957 Chr.14: 24232222-24232222 on Build GRCh38
rs754131162 Chr.14: 24232306-24232306 on Build GRCh38
rs907372814 Chr.14: 24232443-24232443 on Build GRCh38
rs372033101 Chr.14: 24232228-24232228 on Build GRCh38
rs774265073 Chr.14: 24232208-24232208 on Build GRCh38
rs867256608 Chr.14: 24232399-24232399 on Build GRCh38
rs750981651 Chr.14: 24232285-24232285 on Build GRCh38
rs975012584 Chr.14: 24232605-24232605 on Build GRCh38
rs946148982 Chr.14: 24232413-24232413 on Build GRCh38
rs928674356 Chr.14: 24232647-24232647 on Build GRCh38
rs111750853 Chr.14: 24232396-24232396 on Build GRCh38
rs531311294 Chr.14: 24232450-24232450 on Build GRCh38
rs868245917 Chr.14: 24232394-24232394 on Build GRCh38
rs779074616 Chr.14: 24232313-24232313 on Build GRCh38
rs375017092 Chr.14: 24232303-24232303 on Build GRCh38
rs71426837 Chr.14: 24232393-24232393 on Build GRCh38
rs913990286 Chr.14: 24232496-24232496 on Build GRCh38
rs377449127 Chr.14: 24232598-24232598 on Build GRCh38
rs948300048 Chr.14: 24232211-24232211 on Build GRCh38
rs750787856 Chr.14: 24232224-24232224 on Build GRCh38
rs1004595459 Chr.14: 24232543-24232543 on Build GRCh38
rs1014636815 Chr.14: 24232406-24232406 on Build GRCh38
rs569456287 Chr.14: 24232408-24232408 on Build GRCh38
rs761636517 Chr.14: 24232239-24232239 on Build GRCh38
rs376943811 Chr.14: 24232595-24232595 on Build GRCh38
rs1019154406 Chr.14: 24232314-24232314 on Build GRCh38
rs200178610 Chr.14: 24232392-24232392 on Build GRCh38
rs553546204 Chr.14: 24232519-24232519 on Build GRCh38
rs769588288 Chr.14: 24232241-24232241 on Build GRCh38
rs757400523 Chr.14: 24232309-24232309 on Build GRCh38
rs796357963 Chr.14: 24232348-24232348 on Build GRCh38
rs201636009 Chr.14: 24232396-24232397 on Build GRCh38
rs921817774 Chr.14: 24232354-24232354 on Build GRCh38
rs34063134 Chr.14: 24232410-24232411 on Build GRCh38
rs769272318 Chr.14: 24232555-24232555 on Build GRCh38
rs111329140 Chr.14: 24232547-24232547 on Build GRCh38
rs957557096 Chr.14: 24232630-24232631 on Build GRCh38
rs974708608 Chr.14: 24232345-24232345 on Build GRCh38
rs1052401487 Chr.14: 24232476-24232476 on Build GRCh38
rs540808323 Chr.14: 24232353-24232353 on Build GRCh38
rs996001065 Chr.14: 24232637-24232637 on Build GRCh38
rs746589220 Chr.14: 24232297-24232297 on Build GRCh38
rs753072975 Chr.14: 24232298-24232298 on Build GRCh38
rs113067616 Chr.14: 24232397-24232397 on Build GRCh38
rs762477966 Chr.14: 24232276-24232276 on Build GRCh38
rs772979559 Chr.14: 24232246-24232246 on Build GRCh38
rs374692467 Chr.14: 24232323-24232323 on Build GRCh38
rs747187203 Chr.14: 24232391-24232391 on Build GRCh38
rs367915992 Chr.14: 24232189-24232189 on Build GRCh38
rs886096996 Chr.14: 24232539-24232539 on Build GRCh38
rs570352987 Chr.14: 24232523-24232523 on Build GRCh38
rs983160032 Chr.14: 24232321-24232321 on Build GRCh38
rs969694063 Chr.14: 24232442-24232442 on Build GRCh38
rs190461080 Chr.14: 24232352-24232352 on Build GRCh38
rs950492506 Chr.14: 24232320-24232320 on Build GRCh38
rs780130015 Chr.14: 24232267-24232269 on Build GRCh38
rs990077597 Chr.14: 24232633-24232633 on Build GRCh38
rs79349777 Chr.14: 24232507-24232507 on Build GRCh38
rs935922047 Chr.14: 24232382-24232382 on Build GRCh38
rs990580975 Chr.14: 24232403-24232403 on Build GRCh38
rs747017666 Chr.14: 24232221-24232221 on Build GRCh38
rs544283366 Chr.14: 24232296-24232296 on Build GRCh38
rs981411737 Chr.14: 24232444-24232444 on Build GRCh38
rs756528078 Chr.14: 24232301-24232301 on Build GRCh38
rs965865577 Chr.14: 24232604-24232604 on Build GRCh38
rs758652094 Chr.14: 24232461-24232461 on Build GRCh38
rs767944508 Chr.14: 24232294-24232294 on Build GRCh38
rs760116856 Chr.14: 24232293-24232293 on Build GRCh38
rs17794752 Chr.14: 24232499-24232499 on Build GRCh38
rs879241451 Chr.14: 24232404-24232404 on Build GRCh38
rs965815440 Chr.14: 24232337-24232337 on Build GRCh38
rs1056063511 Chr.14: 24232475-24232475 on Build GRCh38
rs868288558 Chr.14: 24232398-24232398 on Build GRCh38
rs984394636 Chr.14: 24232405-24232405 on Build GRCh38
rs552798131 Chr.14: 24232401-24232401 on Build GRCh38
rs918180518 Chr.14: 24232209-24232209 on Build GRCh38
rs34445880 Chr.14: 24232258-24232259 on Build GRCh38
rs769714663 Chr.14: 24232393-24232395 on Build GRCh38
rs779330503 Chr.14: 24232220-24232220 on Build GRCh38
rs901656388 Chr.14: 24232580-24232580 on Build GRCh38
rs560715392 Chr.14: 24232380-24232380 on Build GRCh38
rs561332417 Chr.14: 24232340-24232340 on Build GRCh38
rs562829341 Chr.14: 24232526-24232526 on Build GRCh38
rs1034974723 Chr.14: 24232219-24232219 on Build GRCh38
rs571626471 Chr.14: 24232559-24232559 on Build GRCh38

View Full Product Details