Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24290506-24291018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS1
Entrez Gene ID
115817
Gene Name
dehydrogenase/reductase 1
Gene Aliases
SDR19C1
UniGene
Hs.348350
Gene Chromosome Location
Chr.14: 24290598-24299833 on Build GRCh38
Transcripts
NM_001136050.2 NM_138452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6643297 c.929C>T p.T310I Chr.14: 24290872-24290872 on Build GRCh38
5045809 c.819A>T p.Q273H Chr.14: 24290982-24290982 on Build GRCh38
7364277 c.811C>T p.P271S Chr.14: 24290990-24290990 on Build GRCh38
1203515 c.844C>A p.L282I Chr.14: 24290957-24290957 on Build GRCh38
955011 c.898C>T p.R300C Chr.14: 24290903-24290903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574079507 Chr.14: 24290990-24290990 on Build GRCh38
rs765481094 Chr.14: 24290972-24290972 on Build GRCh38
rs891255766 Chr.14: 24290597-24290597 on Build GRCh38
rs1020898377 Chr.14: 24290922-24290922 on Build GRCh38
rs1000065403 Chr.14: 24290706-24290707 on Build GRCh38
rs4568 Chr.14: 24290803-24290803 on Build GRCh38
rs770444113 Chr.14: 24290850-24290850 on Build GRCh38
rs529685850 Chr.14: 24290705-24290705 on Build GRCh38
rs773895645 Chr.14: 24290948-24290948 on Build GRCh38
rs754152881 Chr.14: 24290965-24290965 on Build GRCh38
rs957977799 Chr.14: 24290856-24290856 on Build GRCh38
rs770734291 Chr.14: 24290846-24290846 on Build GRCh38
rs1018514276 Chr.14: 24290610-24290610 on Build GRCh38
rs958285807 Chr.14: 24290697-24290697 on Build GRCh38
rs1034868419 Chr.14: 24290719-24290719 on Build GRCh38
rs769461700 Chr.14: 24290937-24290937 on Build GRCh38
rs750452881 Chr.14: 24290984-24290984 on Build GRCh38
rs925193451 Chr.14: 24290696-24290696 on Build GRCh38
rs761050614 Chr.14: 24290954-24290954 on Build GRCh38
rs1035765209 Chr.14: 24290902-24290902 on Build GRCh38
rs147677428 Chr.14: 24290726-24290726 on Build GRCh38
rs374593072 Chr.14: 24290637-24290637 on Build GRCh38
rs777525623 Chr.14: 24290993-24290993 on Build GRCh38
rs754887846 Chr.14: 24290987-24290987 on Build GRCh38
rs750765506 Chr.14: 24290550-24290550 on Build GRCh38
rs550896096 Chr.14: 24290664-24290664 on Build GRCh38
rs137896489 Chr.14: 24290890-24290890 on Build GRCh38
rs150174829 Chr.14: 24290988-24290988 on Build GRCh38
rs903929500 Chr.14: 24290692-24290693 on Build GRCh38
rs754939277 Chr.14: 24290912-24290912 on Build GRCh38
rs753986930 Chr.14: 24290900-24290900 on Build GRCh38
rs758865544 Chr.14: 24290629-24290629 on Build GRCh38
rs748912270 Chr.14: 24290943-24290943 on Build GRCh38
rs142273312 Chr.14: 24290762-24290762 on Build GRCh38
rs1046758708 Chr.14: 24290772-24290772 on Build GRCh38
rs1025651307 Chr.14: 24290867-24290867 on Build GRCh38
rs112529214 Chr.14: 24290784-24290784 on Build GRCh38
rs779104077 Chr.14: 24290903-24290903 on Build GRCh38
rs767826944 Chr.14: 24290870-24290870 on Build GRCh38
rs772409680 Chr.14: 24290832-24290832 on Build GRCh38
rs755632621 Chr.14: 24290725-24290725 on Build GRCh38
rs565158722 Chr.14: 24290593-24290593 on Build GRCh38
rs113237099 Chr.14: 24290792-24290792 on Build GRCh38
rs372828362 Chr.14: 24290977-24290977 on Build GRCh38
rs114684041 Chr.14: 24290877-24290877 on Build GRCh38
rs951448499 Chr.14: 24290673-24290673 on Build GRCh38
rs1008099322 Chr.14: 24290659-24290659 on Build GRCh38
rs561260562 Chr.14: 24290929-24290929 on Build GRCh38
rs769423302 Chr.14: 24290824-24290824 on Build GRCh38
rs1032675788 Chr.14: 24290843-24290843 on Build GRCh38
rs1025402664 Chr.14: 24290646-24290646 on Build GRCh38
rs139818812 Chr.14: 24290654-24290654 on Build GRCh38
rs145199333 Chr.14: 24290887-24290887 on Build GRCh38
rs566519009 Chr.14: 24290736-24290736 on Build GRCh38
rs760809520 Chr.14: 24290881-24290881 on Build GRCh38
rs775954531 Chr.14: 24290865-24290865 on Build GRCh38
rs369654913 Chr.14: 24290910-24290910 on Build GRCh38
rs988118164 Chr.14: 24290698-24290698 on Build GRCh38
rs149470750 Chr.14: 24290962-24290962 on Build GRCh38
rs373524926 Chr.14: 24290949-24290949 on Build GRCh38
rs369086635 Chr.14: 24290960-24290960 on Build GRCh38
rs778570755 Chr.14: 24290942-24290942 on Build GRCh38
rs770482024 Chr.14: 24290946-24290946 on Build GRCh38
rs376681789 Chr.14: 24290826-24290826 on Build GRCh38
rs963055671 Chr.14: 24290631-24290631 on Build GRCh38
rs970810126 Chr.14: 24290925-24290925 on Build GRCh38
rs899376092 Chr.14: 24290751-24290751 on Build GRCh38
rs745599685 Chr.14: 24290971-24290971 on Build GRCh38
rs199688816 Chr.14: 24290872-24290872 on Build GRCh38
rs201053608 Chr.14: 24290838-24290838 on Build GRCh38
rs886887589 Chr.14: 24290779-24290779 on Build GRCh38
rs775730024 Chr.14: 24290951-24290951 on Build GRCh38
rs749270244 Chr.14: 24290817-24290817 on Build GRCh38
rs369840074 Chr.14: 24290598-24290598 on Build GRCh38
rs145354185 Chr.14: 24290686-24290686 on Build GRCh38
rs747920240 Chr.14: 24290932-24290932 on Build GRCh38
rs773959326 Chr.14: 24290910-24290911 on Build GRCh38
rs747824806 Chr.14: 24290808-24290808 on Build GRCh38
rs145558204 Chr.14: 24290992-24290992 on Build GRCh38
rs981484129 Chr.14: 24290706-24290706 on Build GRCh38
rs774882704 Chr.14: 24290842-24290842 on Build GRCh38
rs771463248 Chr.14: 24290839-24290839 on Build GRCh38

View Full Product Details