Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24290907-24291410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS1
Entrez Gene ID
115817
Gene Name
dehydrogenase/reductase 1
Gene Aliases
SDR19C1
UniGene
Hs.348350
Gene Chromosome Location
Chr.14: 24290598-24299833 on Build GRCh38
Transcripts
NM_001136050.2 NM_138452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5045809 c.819A>T p.Q273H Chr.14: 24290982-24290982 on Build GRCh38
3386498 c.781C>A p.R261S Chr.14: 24291163-24291163 on Build GRCh38
1203515 c.844C>A p.L282I Chr.14: 24290957-24290957 on Build GRCh38
4878245 c.805G>A p.G269S Chr.14: 24291139-24291139 on Build GRCh38
6876515 c.887C>T p.P296L Chr.14: 24290914-24290914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768752833 Chr.14: 24291215-24291215 on Build GRCh38
rs752733121 Chr.14: 24291151-24291151 on Build GRCh38
rs574079507 Chr.14: 24290990-24290990 on Build GRCh38
rs375287610 Chr.14: 24291307-24291307 on Build GRCh38
rs929438635 Chr.14: 24291282-24291282 on Build GRCh38
rs765481094 Chr.14: 24290972-24290972 on Build GRCh38
rs934971060 Chr.14: 24291126-24291126 on Build GRCh38
rs1024007654 Chr.14: 24290998-24290998 on Build GRCh38
rs761972766 Chr.14: 24291262-24291262 on Build GRCh38
rs773895645 Chr.14: 24290948-24290948 on Build GRCh38
rs754152881 Chr.14: 24290965-24290965 on Build GRCh38
rs1030856433 Chr.14: 24291342-24291342 on Build GRCh38
rs745400245 Chr.14: 24291173-24291173 on Build GRCh38
rs769461700 Chr.14: 24290937-24290937 on Build GRCh38
rs750452881 Chr.14: 24290984-24290984 on Build GRCh38
rs912111522 Chr.14: 24291299-24291299 on Build GRCh38
rs761050614 Chr.14: 24290954-24290954 on Build GRCh38
rs778287414 Chr.14: 24291022-24291022 on Build GRCh38
rs757965746 Chr.14: 24291174-24291174 on Build GRCh38
rs201880552 Chr.14: 24291104-24291104 on Build GRCh38
rs777525623 Chr.14: 24290993-24290993 on Build GRCh38
rs538086205 Chr.14: 24291274-24291274 on Build GRCh38
rs943686019 Chr.14: 24291385-24291385 on Build GRCh38
rs375216375 Chr.14: 24291270-24291270 on Build GRCh38
rs754887846 Chr.14: 24290987-24290987 on Build GRCh38
rs765513848 Chr.14: 24291288-24291288 on Build GRCh38
rs762131126 Chr.14: 24291130-24291130 on Build GRCh38
rs1023473056 Chr.14: 24291287-24291287 on Build GRCh38
rs758955966 Chr.14: 24291142-24291142 on Build GRCh38
rs956410588 Chr.14: 24291356-24291356 on Build GRCh38
rs150174829 Chr.14: 24290988-24290988 on Build GRCh38
rs137938287 Chr.14: 24291243-24291243 on Build GRCh38
rs200040073 Chr.14: 24291179-24291179 on Build GRCh38
rs747486685 Chr.14: 24291102-24291102 on Build GRCh38
rs59742678 Chr.14: 24291071-24291071 on Build GRCh38
rs773189484 Chr.14: 24291139-24291139 on Build GRCh38
rs756895208 Chr.14: 24291018-24291018 on Build GRCh38
rs748912270 Chr.14: 24290943-24290943 on Build GRCh38
rs148881510 Chr.14: 24291077-24291077 on Build GRCh38
rs763940229 Chr.14: 24291162-24291162 on Build GRCh38
rs200636329 Chr.14: 24291140-24291140 on Build GRCh38
rs370794166 Chr.14: 24291224-24291224 on Build GRCh38
rs372828362 Chr.14: 24290977-24290977 on Build GRCh38
rs201101817 Chr.14: 24291166-24291166 on Build GRCh38
rs973595499 Chr.14: 24291024-24291024 on Build GRCh38
rs561260562 Chr.14: 24290929-24290929 on Build GRCh38
rs997121611 Chr.14: 24291098-24291098 on Build GRCh38
rs1035816061 Chr.14: 24291257-24291257 on Build GRCh38
rs899539298 Chr.14: 24291093-24291093 on Build GRCh38
rs2295310 Chr.14: 24291049-24291049 on Build GRCh38
rs938181853 Chr.14: 24291023-24291023 on Build GRCh38
rs368276525 Chr.14: 24291223-24291223 on Build GRCh38
rs34586111 Chr.14: 24291155-24291155 on Build GRCh38
rs983720127 Chr.14: 24291284-24291284 on Build GRCh38
rs1000752062 Chr.14: 24291312-24291312 on Build GRCh38
rs575472175 Chr.14: 24291092-24291092 on Build GRCh38
rs545198591 Chr.14: 24291072-24291072 on Build GRCh38
rs148282342 Chr.14: 24291150-24291150 on Build GRCh38
rs559917499 Chr.14: 24291163-24291163 on Build GRCh38
rs775285045 Chr.14: 24291271-24291272 on Build GRCh38
rs769942715 Chr.14: 24291229-24291229 on Build GRCh38
rs149470750 Chr.14: 24290962-24290962 on Build GRCh38
rs2295309 Chr.14: 24291167-24291167 on Build GRCh38
rs373524926 Chr.14: 24290949-24290949 on Build GRCh38
rs763067122 Chr.14: 24291248-24291248 on Build GRCh38
rs770482024 Chr.14: 24290946-24290946 on Build GRCh38
rs369086635 Chr.14: 24290960-24290960 on Build GRCh38
rs778570755 Chr.14: 24290942-24290942 on Build GRCh38
rs771764036 Chr.14: 24291073-24291074 on Build GRCh38
rs974010205 Chr.14: 24291255-24291255 on Build GRCh38
rs774406520 Chr.14: 24291266-24291266 on Build GRCh38
rs765442917 Chr.14: 24291138-24291138 on Build GRCh38
rs745599685 Chr.14: 24290971-24290971 on Build GRCh38
rs200839029 Chr.14: 24291124-24291124 on Build GRCh38
rs772476732 Chr.14: 24291034-24291034 on Build GRCh38
rs775730024 Chr.14: 24290951-24290951 on Build GRCh38
rs756913740 Chr.14: 24291165-24291165 on Build GRCh38
rs745323010 Chr.14: 24291033-24291033 on Build GRCh38
rs777130594 Chr.14: 24291129-24291129 on Build GRCh38
rs771914758 Chr.14: 24291211-24291212 on Build GRCh38
rs747920240 Chr.14: 24290932-24290932 on Build GRCh38
rs375012309 Chr.14: 24291206-24291206 on Build GRCh38
rs370345152 Chr.14: 24291017-24291017 on Build GRCh38
rs147405102 Chr.14: 24291185-24291185 on Build GRCh38
rs776624581 Chr.14: 24291249-24291249 on Build GRCh38
rs145558204 Chr.14: 24290992-24290992 on Build GRCh38

View Full Product Details