Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24298878-24299389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS1
Entrez Gene ID
115817
Gene Name
dehydrogenase/reductase 1
Gene Aliases
SDR19C1
UniGene
Hs.348350
Gene Chromosome Location
Chr.14: 24290598-24299833 on Build GRCh38
Transcripts
NM_001136050.2 NM_138452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607558 c.130C>T p.L44F Chr.14: 24298977-24298977 on Build GRCh38
1237217 c.59G>A p.G20D Chr.14: 24299048-24299048 on Build GRCh38
6265736 c.105C>A p.Y35* Chr.14: 24299002-24299002 on Build GRCh38
6994616 c.142G>T p.A48S Chr.14: 24298965-24298965 on Build GRCh38
5847956 c.100G>A p.V34I Chr.14: 24299007-24299007 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746955860 Chr.14: 24298992-24298992 on Build GRCh38
rs200621886 Chr.14: 24299105-24299105 on Build GRCh38
rs774414811 Chr.14: 24298991-24298991 on Build GRCh38
rs140692443 Chr.14: 24298997-24298997 on Build GRCh38
rs932412919 Chr.14: 24299287-24299287 on Build GRCh38
rs115389572 Chr.14: 24298914-24298914 on Build GRCh38
rs17851364 Chr.14: 24299081-24299081 on Build GRCh38
rs765157232 Chr.14: 24298967-24298967 on Build GRCh38
rs943902240 Chr.14: 24299261-24299261 on Build GRCh38
rs745900117 Chr.14: 24299073-24299073 on Build GRCh38
rs536211046 Chr.14: 24299296-24299296 on Build GRCh38
rs369212415 Chr.14: 24299138-24299138 on Build GRCh38
rs925118981 Chr.14: 24298969-24298969 on Build GRCh38
rs757495915 Chr.14: 24299150-24299150 on Build GRCh38
rs377446942 Chr.14: 24299146-24299146 on Build GRCh38
rs748097310 Chr.14: 24299087-24299087 on Build GRCh38
rs762707146 Chr.14: 24299097-24299097 on Build GRCh38
rs771060650 Chr.14: 24298990-24298990 on Build GRCh38
rs752178395 Chr.14: 24298971-24298971 on Build GRCh38
rs1004380258 Chr.14: 24299351-24299351 on Build GRCh38
rs201442139 Chr.14: 24298945-24298945 on Build GRCh38
rs1053634005 Chr.14: 24299162-24299162 on Build GRCh38
rs906510740 Chr.14: 24299277-24299277 on Build GRCh38
rs753513434 Chr.14: 24298933-24298933 on Build GRCh38
rs567648980 Chr.14: 24299289-24299289 on Build GRCh38
rs764449798 Chr.14: 24299144-24299144 on Build GRCh38
rs778189762 Chr.14: 24299049-24299049 on Build GRCh38
rs1040746795 Chr.14: 24299280-24299280 on Build GRCh38
rs368336040 Chr.14: 24299036-24299036 on Build GRCh38
rs753198791 Chr.14: 24299139-24299139 on Build GRCh38
rs778710187 Chr.14: 24298933-24298933 on Build GRCh38
rs988503947 Chr.14: 24299230-24299230 on Build GRCh38
rs552374808 Chr.14: 24299168-24299168 on Build GRCh38
rs756482236 Chr.14: 24299048-24299048 on Build GRCh38
rs755528674 Chr.14: 24299037-24299037 on Build GRCh38
rs765050876 Chr.14: 24299001-24299001 on Build GRCh38
rs371494896 Chr.14: 24299147-24299147 on Build GRCh38
rs761541379 Chr.14: 24298941-24298941 on Build GRCh38
rs1040694859 Chr.14: 24299012-24299012 on Build GRCh38
rs115780613 Chr.14: 24299206-24299206 on Build GRCh38
rs374418159 Chr.14: 24299011-24299011 on Build GRCh38
rs913822383 Chr.14: 24299251-24299251 on Build GRCh38
rs768422626 Chr.14: 24298996-24298996 on Build GRCh38
rs760596590 Chr.14: 24298919-24298919 on Build GRCh38
rs12892419 Chr.14: 24299328-24299328 on Build GRCh38
rs200320943 Chr.14: 24299129-24299129 on Build GRCh38
rs369406238 Chr.14: 24298973-24298973 on Build GRCh38
rs61998510 Chr.14: 24299028-24299028 on Build GRCh38
rs749581359 Chr.14: 24299055-24299055 on Build GRCh38
rs1042762254 Chr.14: 24299239-24299239 on Build GRCh38
rs201787891 Chr.14: 24298959-24298959 on Build GRCh38
rs749980818 Chr.14: 24298962-24298962 on Build GRCh38
rs775459252 Chr.14: 24298917-24298917 on Build GRCh38
rs766511436 Chr.14: 24299161-24299161 on Build GRCh38
rs202001969 Chr.14: 24299090-24299090 on Build GRCh38
rs143906801 Chr.14: 24298916-24298916 on Build GRCh38
rs540228614 Chr.14: 24298977-24298977 on Build GRCh38
rs771718420 Chr.14: 24299042-24299054 on Build GRCh38
rs11550019 Chr.14: 24299029-24299029 on Build GRCh38
rs147277524 Chr.14: 24299194-24299194 on Build GRCh38
rs756643656 Chr.14: 24299142-24299142 on Build GRCh38
rs978799546 Chr.14: 24299180-24299180 on Build GRCh38
rs368510430 Chr.14: 24298985-24298985 on Build GRCh38
rs150092528 Chr.14: 24299014-24299014 on Build GRCh38
rs892161579 Chr.14: 24299008-24299008 on Build GRCh38
rs899475389 Chr.14: 24299285-24299285 on Build GRCh38
rs531622686 Chr.14: 24299196-24299196 on Build GRCh38
rs1027868017 Chr.14: 24299050-24299050 on Build GRCh38
rs753215367 Chr.14: 24299045-24299045 on Build GRCh38
rs764749437 Chr.14: 24298956-24298956 on Build GRCh38
rs758022297 Chr.14: 24298965-24298965 on Build GRCh38
rs775088595 Chr.14: 24299141-24299142 on Build GRCh38
rs774025250 Chr.14: 24299124-24299124 on Build GRCh38
rs764018793 Chr.14: 24298925-24298925 on Build GRCh38
rs761477205 Chr.14: 24298998-24298998 on Build GRCh38
rs751055899 Chr.14: 24299031-24299031 on Build GRCh38
rs906596572 Chr.14: 24298953-24298953 on Build GRCh38
rs201377735 Chr.14: 24298932-24298932 on Build GRCh38
rs768671081 Chr.14: 24299082-24299082 on Build GRCh38
rs757407969 Chr.14: 24299061-24299061 on Build GRCh38
rs74731195 Chr.14: 24298957-24298957 on Build GRCh38
rs193095681 Chr.14: 24299085-24299085 on Build GRCh38
rs908318108 Chr.14: 24299109-24299110 on Build GRCh38
rs779067737 Chr.14: 24299158-24299158 on Build GRCh38
rs200356670 Chr.14: 24299093-24299093 on Build GRCh38
rs758489787 Chr.14: 24299042-24299042 on Build GRCh38
rs565891970 Chr.14: 24298986-24298986 on Build GRCh38
rs745732042 Chr.14: 24299008-24299009 on Build GRCh38
rs34225269 Chr.14: 24298960-24298960 on Build GRCh38
rs778981660 Chr.14: 24299069-24299069 on Build GRCh38

View Full Product Details