Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31445919-31446458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTD2
Entrez Gene ID
112487
Gene Name
D-tyrosyl-tRNA deacylase 2 (putative)
Gene Aliases
C14orf126
UniGene
Hs.116014
Gene Chromosome Location
Chr.14: 31446036-31457474 on Build GRCh38
Transcripts
NM_080664.2 NR_110045.1

Gene Symbol
LOC101927124
Entrez Gene ID
101927124
Gene Name
uncharacterized LOC101927124
Gene Aliases
-
UniGene
Hs.647561
Gene Chromosome Location
Chr.14: 31420758-31452883 on Build GRCh38
Transcripts
NM_080664.2 NR_110045.1

SNP Detail

SNP ID Chromosome Location
rs888009814 Chr.14: 31446206-31446206 on Build GRCh38
rs917667714 Chr.14: 31446170-31446170 on Build GRCh38
rs892380915 Chr.14: 31446325-31446325 on Build GRCh38
rs1052255560 Chr.14: 31446311-31446311 on Build GRCh38
rs34607249 Chr.14: 31446298-31446298 on Build GRCh38
rs397852620 Chr.14: 31446343-31446343 on Build GRCh38
rs571979543 Chr.14: 31446350-31446350 on Build GRCh38
rs879137817 Chr.14: 31446193-31446193 on Build GRCh38
rs1022646311 Chr.14: 31446357-31446357 on Build GRCh38
rs566763892 Chr.14: 31446195-31446195 on Build GRCh38
rs904499470 Chr.14: 31446363-31446363 on Build GRCh38
rs753075011 Chr.14: 31446119-31446119 on Build GRCh38
rs997688861 Chr.14: 31446252-31446252 on Build GRCh38
rs5807623 Chr.14: 31446333-31446333 on Build GRCh38
rs954840364 Chr.14: 31446139-31446139 on Build GRCh38
rs942255803 Chr.14: 31446212-31446212 on Build GRCh38
rs564903660 Chr.14: 31446075-31446075 on Build GRCh38
rs968388144 Chr.14: 31446089-31446089 on Build GRCh38
rs529895624 Chr.14: 31446392-31446395 on Build GRCh38
rs151338217 Chr.14: 31446259-31446259 on Build GRCh38
rs1030858101 Chr.14: 31446002-31446002 on Build GRCh38
rs141841124 Chr.14: 31446427-31446427 on Build GRCh38
rs568747368 Chr.14: 31446027-31446027 on Build GRCh38
rs1003352582 Chr.14: 31445989-31445989 on Build GRCh38
rs1039323583 Chr.14: 31446228-31446228 on Build GRCh38
rs1021668046 Chr.14: 31446031-31446031 on Build GRCh38
rs546985564 Chr.14: 31446241-31446241 on Build GRCh38
rs533803017 Chr.14: 31446237-31446237 on Build GRCh38
rs996603297 Chr.14: 31446364-31446364 on Build GRCh38
rs767054786 Chr.14: 31446122-31446122 on Build GRCh38
rs979397083 Chr.14: 31446127-31446127 on Build GRCh38
rs575949108 Chr.14: 31446415-31446415 on Build GRCh38
rs17097892 Chr.14: 31446010-31446010 on Build GRCh38
rs947726136 Chr.14: 31446247-31446247 on Build GRCh38
rs181255940 Chr.14: 31446291-31446291 on Build GRCh38
rs188440604 Chr.14: 31446020-31446020 on Build GRCh38
rs1044799430 Chr.14: 31446251-31446251 on Build GRCh38
rs920242104 Chr.14: 31446239-31446239 on Build GRCh38
rs974064205 Chr.14: 31446213-31446213 on Build GRCh38
rs974824949 Chr.14: 31446069-31446069 on Build GRCh38
rs954808309 Chr.14: 31446404-31446404 on Build GRCh38
rs576245536 Chr.14: 31445949-31445949 on Build GRCh38
rs1020842300 Chr.14: 31446080-31446080 on Build GRCh38
rs532955371 Chr.14: 31446153-31446153 on Build GRCh38
rs908114513 Chr.14: 31446158-31446158 on Build GRCh38
rs969130105 Chr.14: 31446035-31446035 on Build GRCh38
rs551437227 Chr.14: 31446171-31446171 on Build GRCh38
rs891899167 Chr.14: 31446280-31446280 on Build GRCh38
rs183492535 Chr.14: 31446354-31446354 on Build GRCh38
rs372922681 Chr.14: 31446133-31446133 on Build GRCh38
rs940959455 Chr.14: 31446169-31446169 on Build GRCh38
rs1029067312 Chr.14: 31446383-31446383 on Build GRCh38

View Full Product Details