Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34761990-34762455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ1A
Entrez Gene ID
11177
Gene Name
bromodomain adjacent to zinc finger domain 1A
Gene Aliases
ACF1, WALp1, WCRF180, hACF1
UniGene
Hs.509140
Gene Chromosome Location
Chr.14: 34752731-34875647 on Build GRCh38
Transcripts
NM_013448.2 NM_182648.1 XM_011536374.1 XM_011536376.2 XM_017020942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4430024 c.3828G>A p.G1276G Chr.14: 34762172-34762172 on Build GRCh38
4995507 c.3856G>T p.G1286C Chr.14: 34762144-34762144 on Build GRCh38
7140218 c.3853C>A p.R1285S Chr.14: 34762147-34762147 on Build GRCh38
5784800 c.3947G>C p.R1316T Chr.14: 34762053-34762053 on Build GRCh38
6575468 c.3941G>C p.S1314T Chr.14: 34762059-34762059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764084799 Chr.14: 34762153-34762153 on Build GRCh38
rs770632374 Chr.14: 34762120-34762120 on Build GRCh38
rs148152677 Chr.14: 34762048-34762048 on Build GRCh38
rs750347699 Chr.14: 34762208-34762210 on Build GRCh38
rs570044580 Chr.14: 34762352-34762352 on Build GRCh38
rs560209924 Chr.14: 34762147-34762147 on Build GRCh38
rs773811202 Chr.14: 34762134-34762134 on Build GRCh38
rs748836448 Chr.14: 34762264-34762264 on Build GRCh38
rs370552302 Chr.14: 34762043-34762043 on Build GRCh38
rs767568666 Chr.14: 34762044-34762044 on Build GRCh38
rs370494713 Chr.14: 34762260-34762260 on Build GRCh38
rs527563396 Chr.14: 34762287-34762287 on Build GRCh38
rs756109196 Chr.14: 34762051-34762051 on Build GRCh38
rs953873684 Chr.14: 34762136-34762136 on Build GRCh38
rs567490116 Chr.14: 34762333-34762333 on Build GRCh38
rs941610267 Chr.14: 34762358-34762358 on Build GRCh38
rs745307605 Chr.14: 34762081-34762081 on Build GRCh38
rs112142111 Chr.14: 34762378-34762378 on Build GRCh38
rs1043213448 Chr.14: 34762194-34762194 on Build GRCh38
rs983007656 Chr.14: 34762395-34762395 on Build GRCh38
rs762715980 Chr.14: 34762033-34762033 on Build GRCh38
rs113405444 Chr.14: 34762037-34762037 on Build GRCh38
rs758349533 Chr.14: 34762173-34762173 on Build GRCh38
rs770252684 Chr.14: 34762292-34762292 on Build GRCh38
rs202209689 Chr.14: 34762099-34762099 on Build GRCh38
rs770263520 Chr.14: 34762106-34762106 on Build GRCh38
rs1046783872 Chr.14: 34762059-34762059 on Build GRCh38
rs755176637 Chr.14: 34762219-34762219 on Build GRCh38
rs1037126912 Chr.14: 34762364-34762364 on Build GRCh38
rs770985404 Chr.14: 34762371-34762371 on Build GRCh38
rs747411899 Chr.14: 34762083-34762083 on Build GRCh38
rs780086689 Chr.14: 34762082-34762082 on Build GRCh38
rs779522851 Chr.14: 34762192-34762192 on Build GRCh38
rs183081972 Chr.14: 34762227-34762227 on Build GRCh38
rs751791847 Chr.14: 34762212-34762212 on Build GRCh38
rs372560606 Chr.14: 34762268-34762268 on Build GRCh38
rs938184133 Chr.14: 34762329-34762329 on Build GRCh38
rs998234336 Chr.14: 34762406-34762406 on Build GRCh38
rs762328657 Chr.14: 34762112-34762112 on Build GRCh38
rs10150630 Chr.14: 34762321-34762321 on Build GRCh38
rs566400062 Chr.14: 34762159-34762159 on Build GRCh38
rs10150856 Chr.14: 34762374-34762374 on Build GRCh38
rs751400491 Chr.14: 34762041-34762041 on Build GRCh38
rs945481545 Chr.14: 34762278-34762278 on Build GRCh38
rs376498114 Chr.14: 34762098-34762098 on Build GRCh38
rs928436368 Chr.14: 34762424-34762424 on Build GRCh38
rs146357991 Chr.14: 34762036-34762036 on Build GRCh38
rs762918292 Chr.14: 34762245-34762246 on Build GRCh38
rs759222062 Chr.14: 34762135-34762135 on Build GRCh38
rs776972033 Chr.14: 34762095-34762095 on Build GRCh38
rs750577695 Chr.14: 34762179-34762180 on Build GRCh38
rs772695036 Chr.14: 34762030-34762030 on Build GRCh38
rs919772649 Chr.14: 34762365-34762368 on Build GRCh38
rs913902166 Chr.14: 34762273-34762273 on Build GRCh38
rs781584465 Chr.14: 34762221-34762221 on Build GRCh38
rs768831463 Chr.14: 34762092-34762092 on Build GRCh38
rs767478696 Chr.14: 34762138-34762138 on Build GRCh38
rs373464356 Chr.14: 34762064-34762064 on Build GRCh38
rs548999857 Chr.14: 34762133-34762135 on Build GRCh38
rs201288556 Chr.14: 34762038-34762038 on Build GRCh38
rs779187494 Chr.14: 34762056-34762056 on Build GRCh38
rs772747853 Chr.14: 34762110-34762110 on Build GRCh38
rs545004797 Chr.14: 34762146-34762146 on Build GRCh38
rs766121798 Chr.14: 34762272-34762273 on Build GRCh38

View Full Product Details