Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35076875-35077372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177A1
Entrez Gene ID
283635
Gene Name
family with sequence similarity 177 member A1
Gene Aliases
C14orf24
UniGene
Hs.446357
Gene Chromosome Location
Chr.14: 35044907-35083383 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1 NM_173607.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6194829 c.341C>T p.T114I Chr.14: 35077151-35077151 on Build GRCh38
4416732 c.357T>C p.G119G Chr.14: 35077167-35077167 on Build GRCh38
6348360 c.337+6G>T p.? Chr.14: 35077222-35077222 on Build GRCh38
4050632 c.390T>G p.A130A Chr.14: 35077200-35077200 on Build GRCh38
6194830 c.272C>T p.T91I Chr.14: 35077151-35077151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777218565 Chr.14: 35077192-35077192 on Build GRCh38
rs773841902 Chr.14: 35077219-35077219 on Build GRCh38
rs918495367 Chr.14: 35077278-35077278 on Build GRCh38
rs752662181 Chr.14: 35077236-35077236 on Build GRCh38
rs560063186 Chr.14: 35077166-35077166 on Build GRCh38
rs377404411 Chr.14: 35077232-35077232 on Build GRCh38
rs8011242 Chr.14: 35077292-35077292 on Build GRCh38
rs772793988 Chr.14: 35077207-35077207 on Build GRCh38
rs577477880 Chr.14: 35077059-35077059 on Build GRCh38
rs748629588 Chr.14: 35077193-35077193 on Build GRCh38
rs559437734 Chr.14: 35077058-35077059 on Build GRCh38
rs189896429 Chr.14: 35076956-35076956 on Build GRCh38
rs3211139 Chr.14: 35077167-35077167 on Build GRCh38
rs1011045531 Chr.14: 35076932-35076932 on Build GRCh38
rs762541879 Chr.14: 35077208-35077208 on Build GRCh38
rs760539102 Chr.14: 35077243-35077243 on Build GRCh38
rs967494588 Chr.14: 35077083-35077083 on Build GRCh38
rs376190555 Chr.14: 35077259-35077259 on Build GRCh38
rs757350892 Chr.14: 35077165-35077165 on Build GRCh38
rs769436856 Chr.14: 35077196-35077196 on Build GRCh38
rs17103013 Chr.14: 35077246-35077246 on Build GRCh38
rs201438951 Chr.14: 35077230-35077230 on Build GRCh38
rs534459741 Chr.14: 35077322-35077322 on Build GRCh38
rs770681604 Chr.14: 35077218-35077218 on Build GRCh38
rs776985971 Chr.14: 35077122-35077123 on Build GRCh38
rs914347924 Chr.14: 35077072-35077072 on Build GRCh38
rs377332877 Chr.14: 35077121-35077121 on Build GRCh38
rs201685337 Chr.14: 35077150-35077150 on Build GRCh38
rs891147829 Chr.14: 35077294-35077294 on Build GRCh38
rs769395170 Chr.14: 35076948-35076948 on Build GRCh38
rs773029820 Chr.14: 35077152-35077152 on Build GRCh38
rs370623753 Chr.14: 35077129-35077129 on Build GRCh38
rs927916231 Chr.14: 35077327-35077327 on Build GRCh38
rs747442742 Chr.14: 35077177-35077177 on Build GRCh38
rs760355342 Chr.14: 35077315-35077315 on Build GRCh38
rs199770102 Chr.14: 35077158-35077158 on Build GRCh38
rs750557918 Chr.14: 35077250-35077250 on Build GRCh38
rs768882167 Chr.14: 35077182-35077182 on Build GRCh38
rs1008168943 Chr.14: 35077310-35077310 on Build GRCh38
rs531735582 Chr.14: 35077271-35077271 on Build GRCh38
rs563744556 Chr.14: 35077057-35077057 on Build GRCh38
rs111255820 Chr.14: 35076906-35076906 on Build GRCh38
rs751431165 Chr.14: 35077156-35077156 on Build GRCh38
rs563039822 Chr.14: 35077255-35077255 on Build GRCh38
rs772454032 Chr.14: 35077099-35077099 on Build GRCh38
rs966960222 Chr.14: 35076969-35076969 on Build GRCh38
rs893933705 Chr.14: 35076927-35076927 on Build GRCh38
rs767530050 Chr.14: 35077161-35077161 on Build GRCh38
rs766102553 Chr.14: 35077155-35077155 on Build GRCh38
rs1011750842 Chr.14: 35077338-35077338 on Build GRCh38
rs551753207 Chr.14: 35077285-35077285 on Build GRCh38
rs762629142 Chr.14: 35077153-35077153 on Build GRCh38
rs752608811 Chr.14: 35077163-35077163 on Build GRCh38
rs775854255 Chr.14: 35077101-35077101 on Build GRCh38
rs1018493315 Chr.14: 35077314-35077314 on Build GRCh38
rs977095224 Chr.14: 35077108-35077108 on Build GRCh38
rs758586047 Chr.14: 35077169-35077169 on Build GRCh38
rs747482088 Chr.14: 35077103-35077103 on Build GRCh38
rs746091302 Chr.14: 35077168-35077168 on Build GRCh38
rs730882244 Chr.14: 35077176-35077177 on Build GRCh38
rs78042202 Chr.14: 35077210-35077210 on Build GRCh38

View Full Product Details