Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35826258-35826754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRMS1L
Entrez Gene ID
84312
Gene Name
breast cancer metastasis-suppressor 1-like
Gene Aliases
BRMS1
UniGene
Hs.525299
Gene Chromosome Location
Chr.14: 35825596-35871963 on Build GRCh38
Transcripts
NM_032352.3 XM_005268128.1 XM_006720274.2 XM_006720275.2 XM_017021706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7210730 c.19G>T p.G7W Chr.14: 35826535-35826535 on Build GRCh38
3356680 c.65A>C p.Y22S Chr.14: 35826581-35826581 on Build GRCh38
6250661 c.86G>T p.S29I Chr.14: 35826602-35826602 on Build GRCh38
6423039 c.128A>T p.D43V Chr.14: 35826644-35826644 on Build GRCh38
6890978 c.31G>A p.E11K Chr.14: 35826547-35826547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047044022 Chr.14: 35826418-35826418 on Build GRCh38
rs936703950 Chr.14: 35826308-35826308 on Build GRCh38
rs544927218 Chr.14: 35826481-35826481 on Build GRCh38
rs760476835 Chr.14: 35826691-35826691 on Build GRCh38
rs12891057 Chr.14: 35826662-35826662 on Build GRCh38
rs762973311 Chr.14: 35826701-35826701 on Build GRCh38
rs758444583 Chr.14: 35826651-35826651 on Build GRCh38
rs905194780 Chr.14: 35826303-35826303 on Build GRCh38
rs781580459 Chr.14: 35826554-35826554 on Build GRCh38
rs560661545 Chr.14: 35826699-35826699 on Build GRCh38
rs948190281 Chr.14: 35826363-35826363 on Build GRCh38
rs961372775 Chr.14: 35826730-35826730 on Build GRCh38
rs750513925 Chr.14: 35826530-35826530 on Build GRCh38
rs1010075683 Chr.14: 35826366-35826366 on Build GRCh38
rs750534314 Chr.14: 35826644-35826644 on Build GRCh38
rs752515545 Chr.14: 35826615-35826615 on Build GRCh38
rs542381901 Chr.14: 35826417-35826417 on Build GRCh38
rs766051846 Chr.14: 35826486-35826486 on Build GRCh38
rs758757238 Chr.14: 35826536-35826536 on Build GRCh38
rs759321989 Chr.14: 35826487-35826487 on Build GRCh38
rs980976081 Chr.14: 35826494-35826494 on Build GRCh38
rs992798474 Chr.14: 35826347-35826347 on Build GRCh38
rs899756945 Chr.14: 35826447-35826447 on Build GRCh38
rs764139522 Chr.14: 35826692-35826692 on Build GRCh38
rs200249198 Chr.14: 35826473-35826473 on Build GRCh38
rs779181407 Chr.14: 35826504-35826504 on Build GRCh38
rs928320662 Chr.14: 35826282-35826282 on Build GRCh38
rs920122245 Chr.14: 35826687-35826687 on Build GRCh38
rs371738680 Chr.14: 35826510-35826510 on Build GRCh38
rs767382919 Chr.14: 35826612-35826612 on Build GRCh38
rs762385693 Chr.14: 35826597-35826597 on Build GRCh38
rs767379707 Chr.14: 35826489-35826489 on Build GRCh38
rs930086784 Chr.14: 35826397-35826397 on Build GRCh38
rs553067663 Chr.14: 35826323-35826323 on Build GRCh38
rs771175832 Chr.14: 35826425-35826425 on Build GRCh38
rs770271932 Chr.14: 35826577-35826577 on Build GRCh38
rs770507295 Chr.14: 35826607-35826607 on Build GRCh38
rs1041562211 Chr.14: 35826633-35826633 on Build GRCh38
rs747347326 Chr.14: 35826543-35826543 on Build GRCh38
rs748704138 Chr.14: 35826566-35826566 on Build GRCh38
rs776658466 Chr.14: 35826694-35826694 on Build GRCh38
rs772688910 Chr.14: 35826470-35826470 on Build GRCh38
rs375537336 Chr.14: 35826665-35826665 on Build GRCh38
rs112776558 Chr.14: 35826670-35826670 on Build GRCh38
rs770291975 Chr.14: 35826672-35826672 on Build GRCh38
rs773706992 Chr.14: 35826684-35826684 on Build GRCh38
rs774009079 Chr.14: 35826608-35826608 on Build GRCh38
rs769014595 Chr.14: 35826550-35826550 on Build GRCh38
rs891091586 Chr.14: 35826421-35826421 on Build GRCh38
rs983761403 Chr.14: 35826458-35826458 on Build GRCh38
rs992988311 Chr.14: 35826475-35826475 on Build GRCh38
rs115346142 Chr.14: 35826468-35826468 on Build GRCh38
rs1038538852 Chr.14: 35826367-35826367 on Build GRCh38
rs775489681 Chr.14: 35826690-35826690 on Build GRCh38
rs780255594 Chr.14: 35826542-35826542 on Build GRCh38
rs1011405532 Chr.14: 35826453-35826453 on Build GRCh38
rs1039985972 Chr.14: 35826437-35826437 on Build GRCh38
rs764119299 Chr.14: 35826643-35826643 on Build GRCh38
rs916843507 Chr.14: 35826362-35826362 on Build GRCh38
rs772776654 Chr.14: 35826591-35826591 on Build GRCh38
rs778198664 Chr.14: 35826674-35826674 on Build GRCh38
rs767661917 Chr.14: 35826710-35826710 on Build GRCh38
rs900994995 Chr.14: 35826639-35826639 on Build GRCh38
rs1045764952 Chr.14: 35826297-35826298 on Build GRCh38
rs752644454 Chr.14: 35826495-35826495 on Build GRCh38
rs920009451 Chr.14: 35826394-35826394 on Build GRCh38
rs564470699 Chr.14: 35826503-35826503 on Build GRCh38
rs755294795 Chr.14: 35826705-35826705 on Build GRCh38
rs766520272 Chr.14: 35826702-35826702 on Build GRCh38
rs772139825 Chr.14: 35826613-35826613 on Build GRCh38
rs771606859 Chr.14: 35826689-35826689 on Build GRCh38
rs772053371 Chr.14: 35826541-35826543 on Build GRCh38
rs1028275532 Chr.14: 35826440-35826440 on Build GRCh38
rs538039642 Chr.14: 35826372-35826372 on Build GRCh38
rs562159278 Chr.14: 35826460-35826460 on Build GRCh38
rs143811082 Chr.14: 35826676-35826676 on Build GRCh38
rs377430778 Chr.14: 35826686-35826686 on Build GRCh38
rs545490172 Chr.14: 35826314-35826314 on Build GRCh38
rs780162080 Chr.14: 35826660-35826660 on Build GRCh38
rs892521620 Chr.14: 35826329-35826329 on Build GRCh38
rs949622611 Chr.14: 35826290-35826291 on Build GRCh38
rs760748723 Chr.14: 35826623-35826623 on Build GRCh38
rs751579225 Chr.14: 35826703-35826703 on Build GRCh38
rs529387837 Chr.14: 35826718-35826718 on Build GRCh38
rs548328894 Chr.14: 35826720-35826720 on Build GRCh38
rs1015278890 Chr.14: 35826467-35826467 on Build GRCh38
rs539484713 Chr.14: 35826280-35826280 on Build GRCh38
rs547958908 Chr.14: 35826378-35826378 on Build GRCh38
rs140117219 Chr.14: 35826609-35826609 on Build GRCh38
rs926900816 Chr.14: 35826499-35826499 on Build GRCh38
rs573335558 Chr.14: 35826389-35826389 on Build GRCh38
rs912650671 Chr.14: 35826723-35826723 on Build GRCh38
rs111937952 Chr.14: 35826652-35826652 on Build GRCh38

View Full Product Details