Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35834766-35835276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRMS1L
Entrez Gene ID
84312
Gene Name
breast cancer metastasis-suppressor 1-like
Gene Aliases
BRMS1
UniGene
Hs.525299
Gene Chromosome Location
Chr.14: 35825596-35871963 on Build GRCh38
Transcripts
NM_032352.3 XM_005268128.1 XM_006720274.2 XM_006720275.2 XM_017021703.1 XM_017021704.1 XM_017021705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666189 c.418C>A p.Q140K Chr.14: 35834900-35834900 on Build GRCh38
6521447 c.439G>A p.E147K Chr.14: 35834921-35834921 on Build GRCh38
127448 c.366C>T p.I122I Chr.14: 35834848-35834848 on Build GRCh38
7250047 c.441+9A>G p.? Chr.14: 35834932-35834932 on Build GRCh38
6140626 c.439G>T p.E147* Chr.14: 35834921-35834921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369627912 Chr.14: 35834963-35834963 on Build GRCh38
rs376921034 Chr.14: 35835063-35835064 on Build GRCh38
rs142693587 Chr.14: 35834902-35834902 on Build GRCh38
rs747111316 Chr.14: 35834938-35834938 on Build GRCh38
rs566413161 Chr.14: 35834868-35834868 on Build GRCh38
rs189288320 Chr.14: 35835190-35835190 on Build GRCh38
rs373486864 Chr.14: 35835145-35835145 on Build GRCh38
rs796931447 Chr.14: 35835051-35835051 on Build GRCh38
rs757187388 Chr.14: 35834932-35834932 on Build GRCh38
rs766926898 Chr.14: 35834873-35834873 on Build GRCh38
rs891812927 Chr.14: 35834890-35834890 on Build GRCh38
rs551052132 Chr.14: 35835052-35835053 on Build GRCh38
rs1027814603 Chr.14: 35835038-35835038 on Build GRCh38
rs985667232 Chr.14: 35835179-35835179 on Build GRCh38
rs750778447 Chr.14: 35834869-35834869 on Build GRCh38
rs1051814517 Chr.14: 35835047-35835047 on Build GRCh38
rs751127867 Chr.14: 35834884-35834884 on Build GRCh38
rs983211683 Chr.14: 35835078-35835078 on Build GRCh38
rs748367199 Chr.14: 35834943-35834943 on Build GRCh38
rs777023940 Chr.14: 35834910-35834910 on Build GRCh38
rs950063422 Chr.14: 35835242-35835242 on Build GRCh38
rs756632546 Chr.14: 35835248-35835248 on Build GRCh38
rs529306370 Chr.14: 35834805-35834805 on Build GRCh38
rs554151728 Chr.14: 35835210-35835210 on Build GRCh38
rs891760981 Chr.14: 35835227-35835227 on Build GRCh38
rs934225545 Chr.14: 35835044-35835044 on Build GRCh38
rs776665415 Chr.14: 35834832-35834832 on Build GRCh38
rs372667102 Chr.14: 35835236-35835236 on Build GRCh38
rs965703347 Chr.14: 35835121-35835121 on Build GRCh38
rs770845990 Chr.14: 35834799-35834799 on Build GRCh38
rs755773558 Chr.14: 35834909-35834909 on Build GRCh38
rs770043914 Chr.14: 35834961-35834961 on Build GRCh38
rs942665336 Chr.14: 35835097-35835097 on Build GRCh38
rs569522158 Chr.14: 35834970-35834970 on Build GRCh38
rs749074108 Chr.14: 35834928-35834928 on Build GRCh38
rs749277182 Chr.14: 35834797-35834797 on Build GRCh38
rs924148463 Chr.14: 35835043-35835043 on Build GRCh38
rs776740009 Chr.14: 35834942-35834942 on Build GRCh38
rs138114776 Chr.14: 35834899-35834899 on Build GRCh38
rs763611945 Chr.14: 35834872-35834872 on Build GRCh38
rs773716909 Chr.14: 35834962-35834962 on Build GRCh38
rs573755499 Chr.14: 35835221-35835221 on Build GRCh38
rs370454115 Chr.14: 35834824-35834824 on Build GRCh38
rs2415305 Chr.14: 35835052-35835052 on Build GRCh38
rs775610580 Chr.14: 35834800-35834800 on Build GRCh38
rs762268409 Chr.14: 35834865-35834865 on Build GRCh38
rs1046871968 Chr.14: 35835194-35835194 on Build GRCh38
rs4981309 Chr.14: 35835162-35835162 on Build GRCh38
rs3058641 Chr.14: 35835064-35835065 on Build GRCh38
rs371215575 Chr.14: 35835204-35835204 on Build GRCh38
rs386381117 Chr.14: 35835058-35835059 on Build GRCh38
rs1014574702 Chr.14: 35835111-35835111 on Build GRCh38
rs976966559 Chr.14: 35835128-35835128 on Build GRCh38
rs768577531 Chr.14: 35834939-35834939 on Build GRCh38
rs768176526 Chr.14: 35834810-35834811 on Build GRCh38
rs760676401 Chr.14: 35834820-35834820 on Build GRCh38
rs111903762 Chr.14: 35835040-35835040 on Build GRCh38

View Full Product Details