Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 37590782-37591309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA1
Entrez Gene ID
3169
Gene Name
forkhead box A1
Gene Aliases
HNF3A, TCF3A
UniGene
Hs.163484
Gene Chromosome Location
Chr.14: 37589552-37595120 on Build GRCh38
Transcripts
NM_004496.3 XM_017021246.1

SNP Detail

SNP ID Chromosome Location
rs560369071 Chr.14: 37590943-37590943 on Build GRCh38
rs929170662 Chr.14: 37591145-37591145 on Build GRCh38
rs370407762 Chr.14: 37591198-37591198 on Build GRCh38
rs553682929 Chr.14: 37591065-37591065 on Build GRCh38
rs899033517 Chr.14: 37590810-37590810 on Build GRCh38
rs34059222 Chr.14: 37590887-37590887 on Build GRCh38
rs771564852 Chr.14: 37590924-37590924 on Build GRCh38
rs1047525590 Chr.14: 37591219-37591219 on Build GRCh38
rs1017695073 Chr.14: 37591238-37591238 on Build GRCh38
rs768452041 Chr.14: 37591069-37591069 on Build GRCh38
rs35689225 Chr.14: 37591245-37591246 on Build GRCh38
rs1036762646 Chr.14: 37591067-37591067 on Build GRCh38
rs916552436 Chr.14: 37591073-37591073 on Build GRCh38
rs760722682 Chr.14: 37591246-37591246 on Build GRCh38
rs1010983929 Chr.14: 37591285-37591285 on Build GRCh38
rs555361506 Chr.14: 37591142-37591142 on Build GRCh38
rs570945930 Chr.14: 37591025-37591025 on Build GRCh38
rs1033989021 Chr.14: 37590882-37590882 on Build GRCh38
rs866341845 Chr.14: 37591099-37591099 on Build GRCh38
rs1008561395 Chr.14: 37591227-37591227 on Build GRCh38
rs28756127 Chr.14: 37591034-37591034 on Build GRCh38
rs575199708 Chr.14: 37591172-37591172 on Build GRCh38
rs1052935 Chr.14: 37591147-37591147 on Build GRCh38
rs577740419 Chr.14: 37591234-37591234 on Build GRCh38
rs976211371 Chr.14: 37591266-37591266 on Build GRCh38
rs965333236 Chr.14: 37590960-37590960 on Build GRCh38
rs941351305 Chr.14: 37591107-37591107 on Build GRCh38
rs761345795 Chr.14: 37591185-37591185 on Build GRCh38
rs568122813 Chr.14: 37590863-37590863 on Build GRCh38
rs577328157 Chr.14: 37590993-37590993 on Build GRCh38
rs573552437 Chr.14: 37591070-37591070 on Build GRCh38
rs897899940 Chr.14: 37590850-37590850 on Build GRCh38
rs1052936 Chr.14: 37591146-37591146 on Build GRCh38
rs771938444 Chr.14: 37591205-37591205 on Build GRCh38
rs373922372 Chr.14: 37591276-37591276 on Build GRCh38
rs540009140 Chr.14: 37591255-37591255 on Build GRCh38
rs971290223 Chr.14: 37590862-37590862 on Build GRCh38
rs774003130 Chr.14: 37591133-37591133 on Build GRCh38
rs923412470 Chr.14: 37591016-37591016 on Build GRCh38
rs75207901 Chr.14: 37591007-37591008 on Build GRCh38
rs1017247183 Chr.14: 37590939-37590939 on Build GRCh38
rs966905430 Chr.14: 37591015-37591015 on Build GRCh38
rs1014047573 Chr.14: 37591122-37591122 on Build GRCh38
rs957509140 Chr.14: 37591219-37591219 on Build GRCh38
rs1026044118 Chr.14: 37590872-37590872 on Build GRCh38
rs1024524056 Chr.14: 37590860-37590860 on Build GRCh38
rs979965527 Chr.14: 37590879-37590879 on Build GRCh38
rs1055133461 Chr.14: 37591072-37591072 on Build GRCh38
rs928049664 Chr.14: 37591055-37591055 on Build GRCh38
rs911822 Chr.14: 37591222-37591222 on Build GRCh38
rs888008996 Chr.14: 37591046-37591046 on Build GRCh38
rs993288925 Chr.14: 37590840-37590840 on Build GRCh38
rs994048587 Chr.14: 37590856-37590856 on Build GRCh38
rs543970164 Chr.14: 37591220-37591220 on Build GRCh38
rs190079848 Chr.14: 37591135-37591135 on Build GRCh38
rs1050692749 Chr.14: 37591040-37591040 on Build GRCh38
rs915469963 Chr.14: 37591002-37591002 on Build GRCh38
rs1001304639 Chr.14: 37591169-37591169 on Build GRCh38

View Full Product Details