Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 38255732-38256246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC14A
Entrez Gene ID
161198
Gene Name
C-type lectin domain family 14 member A
Gene Aliases
C14orf27, CEG1, EGFR-5
UniGene
Hs.525307
Gene Chromosome Location
Chr.14: 38254000-38256370 on Build GRCh38
Transcripts
NM_175060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5690206 c.208C>A p.L70M Chr.14: 38255815-38255815 on Build GRCh38
4963993 c.199G>A p.G67S Chr.14: 38255824-38255824 on Build GRCh38
6045987 c.249C>T p.P83P Chr.14: 38255774-38255774 on Build GRCh38
4949498 c.187A>C p.T63P Chr.14: 38255836-38255836 on Build GRCh38
4050772 c.97G>A p.A33T Chr.14: 38255926-38255926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989405122 Chr.14: 38256185-38256185 on Build GRCh38
rs774280343 Chr.14: 38256211-38256211 on Build GRCh38
rs776268464 Chr.14: 38255809-38255809 on Build GRCh38
rs768761837 Chr.14: 38255970-38255970 on Build GRCh38
rs971794168 Chr.14: 38255821-38255821 on Build GRCh38
rs758048393 Chr.14: 38255915-38255915 on Build GRCh38
rs779399763 Chr.14: 38255923-38255923 on Build GRCh38
rs971766257 Chr.14: 38256133-38256133 on Build GRCh38
rs915040001 Chr.14: 38256215-38256215 on Build GRCh38
rs750950940 Chr.14: 38255900-38255900 on Build GRCh38
rs779813119 Chr.14: 38255797-38255797 on Build GRCh38
rs538320294 Chr.14: 38256017-38256017 on Build GRCh38
rs959990074 Chr.14: 38255847-38255847 on Build GRCh38
rs959242025 Chr.14: 38255769-38255769 on Build GRCh38
rs763532442 Chr.14: 38256035-38256035 on Build GRCh38
rs566211254 Chr.14: 38255902-38255902 on Build GRCh38
rs1034864203 Chr.14: 38256064-38256064 on Build GRCh38
rs956159027 Chr.14: 38256166-38256166 on Build GRCh38
rs748747134 Chr.14: 38256032-38256032 on Build GRCh38
rs747943604 Chr.14: 38255946-38255946 on Build GRCh38
rs760765387 Chr.14: 38255964-38255964 on Build GRCh38
rs1002862941 Chr.14: 38256010-38256010 on Build GRCh38
rs987139137 Chr.14: 38256156-38256156 on Build GRCh38
rs772238929 Chr.14: 38255956-38255956 on Build GRCh38
rs933021078 Chr.14: 38255833-38255833 on Build GRCh38
rs2812035 Chr.14: 38256080-38256080 on Build GRCh38
rs775450370 Chr.14: 38255959-38255959 on Build GRCh38
rs764324310 Chr.14: 38255860-38255860 on Build GRCh38
rs532618414 Chr.14: 38255794-38255794 on Build GRCh38
rs992794040 Chr.14: 38255853-38255853 on Build GRCh38
rs775852376 Chr.14: 38255843-38255843 on Build GRCh38
rs745372944 Chr.14: 38255792-38255792 on Build GRCh38
rs930439282 Chr.14: 38255988-38255988 on Build GRCh38
rs1038758250 Chr.14: 38255978-38255978 on Build GRCh38
rs919029004 Chr.14: 38256135-38256135 on Build GRCh38
rs372040859 Chr.14: 38256024-38256024 on Build GRCh38
rs746724750 Chr.14: 38255927-38255927 on Build GRCh38
rs769756571 Chr.14: 38255823-38255823 on Build GRCh38
rs768536280 Chr.14: 38255800-38255800 on Build GRCh38
rs769523243 Chr.14: 38255947-38255947 on Build GRCh38
rs1014203987 Chr.14: 38256083-38256083 on Build GRCh38
rs760781143 Chr.14: 38255845-38255845 on Build GRCh38
rs756563478 Chr.14: 38255754-38255754 on Build GRCh38
rs1003512102 Chr.14: 38255762-38255762 on Build GRCh38
rs910250227 Chr.14: 38256179-38256179 on Build GRCh38
rs934844682 Chr.14: 38255936-38255936 on Build GRCh38
rs1022997060 Chr.14: 38256093-38256093 on Build GRCh38
rs912157226 Chr.14: 38255960-38255960 on Build GRCh38
rs552707013 Chr.14: 38255870-38255870 on Build GRCh38
rs775176720 Chr.14: 38255952-38255952 on Build GRCh38
rs550774538 Chr.14: 38255774-38255774 on Build GRCh38
rs754610181 Chr.14: 38255928-38255928 on Build GRCh38
rs570682344 Chr.14: 38255782-38255782 on Build GRCh38
rs963514202 Chr.14: 38256126-38256126 on Build GRCh38
rs534698776 Chr.14: 38255974-38255974 on Build GRCh38
rs754182776 Chr.14: 38255863-38255863 on Build GRCh38
rs762324985 Chr.14: 38255990-38255990 on Build GRCh38
rs779519677 Chr.14: 38255783-38255783 on Build GRCh38
rs954647232 Chr.14: 38256139-38256139 on Build GRCh38
rs767485639 Chr.14: 38255834-38255834 on Build GRCh38
rs943259323 Chr.14: 38256180-38256180 on Build GRCh38
rs542654188 Chr.14: 38256182-38256182 on Build GRCh38
rs747890154 Chr.14: 38255815-38255815 on Build GRCh38
rs757625739 Chr.14: 38255864-38255864 on Build GRCh38
rs764365650 Chr.14: 38255766-38255766 on Build GRCh38
rs568635618 Chr.14: 38256074-38256074 on Build GRCh38
rs372360218 Chr.14: 38255987-38255987 on Build GRCh38
rs780858742 Chr.14: 38255934-38255934 on Build GRCh38
rs903153060 Chr.14: 38256004-38256004 on Build GRCh38
rs970478265 Chr.14: 38256117-38256117 on Build GRCh38
rs759629465 Chr.14: 38255827-38255827 on Build GRCh38
rs369601289 Chr.14: 38256100-38256100 on Build GRCh38
rs749222346 Chr.14: 38255954-38255954 on Build GRCh38
rs878915973 Chr.14: 38256052-38256052 on Build GRCh38
rs773069926 Chr.14: 38255826-38255826 on Build GRCh38

View Full Product Details