Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39265611-39266106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE5
Entrez Gene ID
4253
Gene Name
CTAGE family member 5, ER export factor
Gene Aliases
MEA6, MGEA, MGEA11, MGEA6
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265264-39351193 on Build GRCh38
Transcripts
NM_203354.2 XM_011536778.2 XM_011536785.2 XM_017021316.1 XM_017021317.1 XM_017021322.1 XM_017021324.1 XM_017021327.1 NR_038935.1

Gene Symbol
LOC100288846
Entrez Gene ID
100288846
Gene Name
uncharacterized LOC100288846
Gene Aliases
-
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265703-39267061 on Build GRCh38
Transcripts
NM_203354.2 XM_011536778.2 XM_011536785.2 XM_017021316.1 XM_017021317.1 XM_017021322.1 XM_017021324.1 XM_017021327.1 NR_038935.1

SNP Detail

SNP ID Chromosome Location
rs904644844 Chr.14: 39265988-39265988 on Build GRCh38
rs1003248563 Chr.14: 39265809-39265809 on Build GRCh38
rs899989600 Chr.14: 39265688-39265688 on Build GRCh38
rs910563274 Chr.14: 39265956-39265956 on Build GRCh38
rs192872983 Chr.14: 39265905-39265905 on Build GRCh38
rs1014586727 Chr.14: 39265806-39265806 on Build GRCh38
rs12587713 Chr.14: 39265900-39265900 on Build GRCh38
rs778323673 Chr.14: 39265904-39265904 on Build GRCh38
rs771993929 Chr.14: 39265981-39265981 on Build GRCh38
rs950432643 Chr.14: 39265898-39265898 on Build GRCh38
rs762508696 Chr.14: 39265761-39265761 on Build GRCh38
rs932790357 Chr.14: 39265972-39265972 on Build GRCh38
rs910677678 Chr.14: 39265653-39265653 on Build GRCh38
rs960010026 Chr.14: 39265794-39265794 on Build GRCh38
rs1048017612 Chr.14: 39266047-39266047 on Build GRCh38
rs564796607 Chr.14: 39266059-39266059 on Build GRCh38
rs973469286 Chr.14: 39266006-39266006 on Build GRCh38
rs17109079 Chr.14: 39265896-39265896 on Build GRCh38
rs899019676 Chr.14: 39265744-39265744 on Build GRCh38
rs996947844 Chr.14: 39265696-39265696 on Build GRCh38
rs944336221 Chr.14: 39265913-39265913 on Build GRCh38
rs920665520 Chr.14: 39266011-39266011 on Build GRCh38
rs190212368 Chr.14: 39266044-39266044 on Build GRCh38
rs1043084267 Chr.14: 39265702-39265702 on Build GRCh38
rs1033754586 Chr.14: 39265836-39265838 on Build GRCh38
rs118132193 Chr.14: 39265923-39265923 on Build GRCh38
rs921446244 Chr.14: 39265965-39265965 on Build GRCh38
rs113056324 Chr.14: 39265862-39265862 on Build GRCh38
rs1041254512 Chr.14: 39265665-39265665 on Build GRCh38
rs757534014 Chr.14: 39265642-39265642 on Build GRCh38
rs973194122 Chr.14: 39265883-39265883 on Build GRCh38
rs929110320 Chr.14: 39265760-39265760 on Build GRCh38
rs78977030 Chr.14: 39265639-39265639 on Build GRCh38
rs944268921 Chr.14: 39265661-39265661 on Build GRCh38
rs943336409 Chr.14: 39266001-39266001 on Build GRCh38
rs764215667 Chr.14: 39265642-39265645 on Build GRCh38
rs968887655 Chr.14: 39265872-39265872 on Build GRCh38
rs542738434 Chr.14: 39265634-39265635 on Build GRCh38
rs150503222 Chr.14: 39266072-39266072 on Build GRCh38
rs115534011 Chr.14: 39266076-39266076 on Build GRCh38
rs775180950 Chr.14: 39265650-39265650 on Build GRCh38
rs748088561 Chr.14: 39265925-39265925 on Build GRCh38
rs543426586 Chr.14: 39265933-39265933 on Build GRCh38
rs998783129 Chr.14: 39265736-39265736 on Build GRCh38
rs904551566 Chr.14: 39265731-39265731 on Build GRCh38
rs1022030779 Chr.14: 39265865-39265865 on Build GRCh38
rs538717082 Chr.14: 39265754-39265754 on Build GRCh38
rs768218341 Chr.14: 39265901-39265901 on Build GRCh38
rs894779133 Chr.14: 39265838-39265838 on Build GRCh38
rs1031620956 Chr.14: 39265786-39265786 on Build GRCh38
rs772909580 Chr.14: 39266051-39266051 on Build GRCh38
rs141805016 Chr.14: 39265860-39265860 on Build GRCh38
rs943451073 Chr.14: 39265695-39265695 on Build GRCh38
rs184917759 Chr.14: 39265986-39265986 on Build GRCh38

View Full Product Details