Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39266764-39267266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE5
Entrez Gene ID
4253
Gene Name
CTAGE family member 5, ER export factor
Gene Aliases
MEA6, MGEA, MGEA11, MGEA6
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265264-39351193 on Build GRCh38
Transcripts
NM_001247988.1 NM_001247989.1 NM_001247990.1 NM_005930.3 NM_203354.2 NM_203355.2 NM_203356.2 XM_005267648.4 XM_006720148.2 XM_011536775.2 XM_011536778.2 XM_011536785.2 XM_017021314.1 XM_017021315.1 XM_017021316.1 XM_017021317.1 XM_017021318.1 XM_017021319.1 XM_017021320.1 XM_017021321.1 XM_017021322.1 XM_017021324.1 XM_017021325.1 XM_017021327.1 XM_017021328.1 XM_017021329.1 XM_017021330.1 NR_038935.1

Gene Symbol
LOC100288846
Entrez Gene ID
100288846
Gene Name
uncharacterized LOC100288846
Gene Aliases
-
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265703-39267061 on Build GRCh38
Transcripts
NM_001247988.1 NM_001247989.1 NM_001247990.1 NM_005930.3 NM_203354.2 NM_203355.2 NM_203356.2 XM_005267648.4 XM_006720148.2 XM_011536775.2 XM_011536778.2 XM_011536785.2 XM_017021314.1 XM_017021315.1 XM_017021316.1 XM_017021317.1 XM_017021318.1 XM_017021319.1 XM_017021320.1 XM_017021321.1 XM_017021322.1 XM_017021324.1 XM_017021325.1 XM_017021327.1 XM_017021328.1 XM_017021329.1 XM_017021330.1 NR_038935.1

SNP Detail

SNP ID Chromosome Location
rs1014943344 Chr.14: 39267079-39267079 on Build GRCh38
rs930836377 Chr.14: 39266955-39266955 on Build GRCh38
rs1023756579 Chr.14: 39267006-39267006 on Build GRCh38
rs931870685 Chr.14: 39267108-39267108 on Build GRCh38
rs757794583 Chr.14: 39267114-39267114 on Build GRCh38
rs1030743648 Chr.14: 39267221-39267221 on Build GRCh38
rs985963778 Chr.14: 39267118-39267118 on Build GRCh38
rs1019513592 Chr.14: 39267207-39267207 on Build GRCh38
rs1046474612 Chr.14: 39267156-39267156 on Build GRCh38
rs967889181 Chr.14: 39266885-39266885 on Build GRCh38
rs930606903 Chr.14: 39266784-39266784 on Build GRCh38
rs918162895 Chr.14: 39267140-39267140 on Build GRCh38
rs1045078174 Chr.14: 39267142-39267142 on Build GRCh38
rs956640232 Chr.14: 39266895-39266896 on Build GRCh38
rs936419508 Chr.14: 39266976-39266976 on Build GRCh38
rs971549249 Chr.14: 39266895-39266895 on Build GRCh38
rs1000514485 Chr.14: 39266864-39266864 on Build GRCh38
rs775826682 Chr.14: 39267209-39267209 on Build GRCh38
rs436691 Chr.14: 39267090-39267090 on Build GRCh38
rs114848241 Chr.14: 39266819-39266819 on Build GRCh38
rs975030450 Chr.14: 39267087-39267087 on Build GRCh38
rs996503602 Chr.14: 39267224-39267224 on Build GRCh38
rs1039387504 Chr.14: 39266791-39266791 on Build GRCh38
rs1002191221 Chr.14: 39267167-39267167 on Build GRCh38
rs879661632 Chr.14: 39267089-39267090 on Build GRCh38
rs548654464 Chr.14: 39267115-39267115 on Build GRCh38
rs1005333341 Chr.14: 39266856-39266856 on Build GRCh38
rs1024536857 Chr.14: 39267003-39267003 on Build GRCh38
rs1035021493 Chr.14: 39266858-39266858 on Build GRCh38
rs528503106 Chr.14: 39267109-39267109 on Build GRCh38
rs925108540 Chr.14: 39267122-39267122 on Build GRCh38
rs907476443 Chr.14: 39267159-39267159 on Build GRCh38
rs12896609 Chr.14: 39266925-39266925 on Build GRCh38
rs976140579 Chr.14: 39267211-39267211 on Build GRCh38
rs940833029 Chr.14: 39267131-39267131 on Build GRCh38
rs1050148324 Chr.14: 39266963-39266963 on Build GRCh38
rs779878505 Chr.14: 39266992-39266992 on Build GRCh38
rs570800366 Chr.14: 39266859-39266859 on Build GRCh38
rs186525189 Chr.14: 39266989-39266989 on Build GRCh38
rs41317316 Chr.14: 39266970-39266970 on Build GRCh38
rs530715774 Chr.14: 39267196-39267196 on Build GRCh38
rs923101383 Chr.14: 39266954-39266954 on Build GRCh38
rs927333408 Chr.14: 39266907-39266907 on Build GRCh38
rs1001740274 Chr.14: 39267026-39267026 on Build GRCh38
rs574648019 Chr.14: 39266931-39266931 on Build GRCh38
rs963927810 Chr.14: 39267219-39267219 on Build GRCh38
rs952096805 Chr.14: 39266921-39266921 on Build GRCh38
rs746907608 Chr.14: 39267053-39267053 on Build GRCh38
rs114049785 Chr.14: 39266805-39266805 on Build GRCh38
rs940961705 Chr.14: 39266986-39266986 on Build GRCh38
rs1052083232 Chr.14: 39266980-39266980 on Build GRCh38
rs1010478611 Chr.14: 39266890-39266890 on Build GRCh38
rs952457495 Chr.14: 39267119-39267119 on Build GRCh38
rs73277415 Chr.14: 39267205-39267205 on Build GRCh38
rs12897075 Chr.14: 39267166-39267166 on Build GRCh38
rs892143424 Chr.14: 39267143-39267143 on Build GRCh38
rs1006314339 Chr.14: 39267193-39267193 on Build GRCh38
rs754564543 Chr.14: 39266783-39266783 on Build GRCh38
rs1049021185 Chr.14: 39266790-39266790 on Build GRCh38
rs12588387 Chr.14: 39266892-39266892 on Build GRCh38
rs559461945 Chr.14: 39267103-39267103 on Build GRCh38
rs907821322 Chr.14: 39266798-39266798 on Build GRCh38
rs940638717 Chr.14: 39266933-39266933 on Build GRCh38
rs538951216 Chr.14: 39267175-39267175 on Build GRCh38
rs956705755 Chr.14: 39267063-39267063 on Build GRCh38
rs976425772 Chr.14: 39267097-39267097 on Build GRCh38
rs1000958607 Chr.14: 39267158-39267158 on Build GRCh38
rs887912445 Chr.14: 39267179-39267179 on Build GRCh38
rs114209396 Chr.14: 39266861-39266861 on Build GRCh38
rs930783892 Chr.14: 39266810-39266810 on Build GRCh38
rs552719171 Chr.14: 39266897-39266897 on Build GRCh38
rs562209657 Chr.14: 39267132-39267132 on Build GRCh38
rs931812937 Chr.14: 39266962-39266962 on Build GRCh38
rs112830177 Chr.14: 39266841-39266841 on Build GRCh38
rs1034499530 Chr.14: 39267037-39267037 on Build GRCh38
rs12896883 Chr.14: 39267061-39267061 on Build GRCh38
rs183229880 Chr.14: 39266924-39266924 on Build GRCh38
rs962033113 Chr.14: 39267084-39267084 on Build GRCh38
rs759861554 Chr.14: 39267127-39267127 on Build GRCh38
rs901670848 Chr.14: 39266850-39266850 on Build GRCh38
rs577503658 Chr.14: 39267022-39267022 on Build GRCh38
rs1031373045 Chr.14: 39266891-39266891 on Build GRCh38
rs114353705 Chr.14: 39267228-39267228 on Build GRCh38
rs539322352 Chr.14: 39267233-39267233 on Build GRCh38
rs770103410 Chr.14: 39267152-39267152 on Build GRCh38
rs1049226965 Chr.14: 39266966-39266966 on Build GRCh38
rs903549129 Chr.14: 39266862-39266862 on Build GRCh38
rs1012227345 Chr.14: 39267005-39267005 on Build GRCh38
rs1030603954 Chr.14: 39266886-39266886 on Build GRCh38
rs572814807 Chr.14: 39266905-39266905 on Build GRCh38
rs1001138835 Chr.14: 39267039-39267039 on Build GRCh38
rs12896340 Chr.14: 39266939-39266939 on Build GRCh38
rs41317314 Chr.14: 39267073-39267073 on Build GRCh38
rs907376932 Chr.14: 39267016-39267016 on Build GRCh38
rs913245122 Chr.14: 39266916-39266916 on Build GRCh38
rs956327656 Chr.14: 39266888-39266888 on Build GRCh38
rs1008163380 Chr.14: 39267068-39267068 on Build GRCh38

View Full Product Details