Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39267441-39267949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE5
Entrez Gene ID
4253
Gene Name
CTAGE family member 5, ER export factor
Gene Aliases
MEA6, MGEA, MGEA11, MGEA6
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265264-39351193 on Build GRCh38
Transcripts
NM_001247988.1 NM_001247989.1 NM_001247990.1 NM_005930.3 NM_203354.2 NM_203355.2 NM_203356.2 XM_005267648.4 XM_006720148.2 XM_011536775.2 XM_011536778.2 XM_011536784.2 XM_011536785.2 XM_017021314.1 XM_017021315.1 XM_017021316.1 XM_017021317.1 XM_017021318.1 XM_017021319.1 XM_017021320.1 XM_017021321.1 XM_017021322.1 XM_017021323.1 XM_017021324.1 XM_017021325.1 XM_017021326.1 XM_017021327.1 XM_017021328.1 XM_017021329.1 XM_017021330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4844283 c.45C>T p.L15L Chr.14: 39267504-39267504 on Build GRCh38
955688 c.51G>T p.E17D Chr.14: 39267510-39267510 on Build GRCh38
339611 c.49G>T p.E17* Chr.14: 39267508-39267508 on Build GRCh38
4844284 c.45C>T p.L15L Chr.14: 39267504-39267504 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777798508 Chr.14: 39267572-39267572 on Build GRCh38
rs7140561 Chr.14: 39267476-39267476 on Build GRCh38
rs751260971 Chr.14: 39267555-39267555 on Build GRCh38
rs905604405 Chr.14: 39267885-39267885 on Build GRCh38
rs533158189 Chr.14: 39267791-39267791 on Build GRCh38
rs770344379 Chr.14: 39267530-39267530 on Build GRCh38
rs773541449 Chr.14: 39267535-39267535 on Build GRCh38
rs1048329668 Chr.14: 39267575-39267575 on Build GRCh38
rs909538345 Chr.14: 39267772-39267772 on Build GRCh38
rs566806358 Chr.14: 39267822-39267822 on Build GRCh38
rs762013883 Chr.14: 39267528-39267528 on Build GRCh38
rs531054689 Chr.14: 39267735-39267735 on Build GRCh38
rs933454326 Chr.14: 39267821-39267821 on Build GRCh38
rs573151225 Chr.14: 39267615-39267615 on Build GRCh38
rs17855895 Chr.14: 39267490-39267490 on Build GRCh38
rs763145286 Chr.14: 39267468-39267468 on Build GRCh38
rs1033496750 Chr.14: 39267922-39267922 on Build GRCh38
rs752179503 Chr.14: 39267497-39267497 on Build GRCh38
rs537519427 Chr.14: 39267918-39267918 on Build GRCh38
rs1015680877 Chr.14: 39267614-39267614 on Build GRCh38
rs1020794983 Chr.14: 39267672-39267672 on Build GRCh38
rs988369996 Chr.14: 39267734-39267734 on Build GRCh38
rs932459866 Chr.14: 39267546-39267546 on Build GRCh38
rs781018209 Chr.14: 39267487-39267487 on Build GRCh38
rs772241512 Chr.14: 39267500-39267500 on Build GRCh38
rs972684337 Chr.14: 39267794-39267794 on Build GRCh38
rs754654507 Chr.14: 39267713-39267713 on Build GRCh38
rs568593534 Chr.14: 39267862-39267862 on Build GRCh38
rs959100775 Chr.14: 39267752-39267752 on Build GRCh38
rs762372630 Chr.14: 39267470-39267470 on Build GRCh38
rs755817221 Chr.14: 39267566-39267566 on Build GRCh38
rs1009489529 Chr.14: 39267888-39267888 on Build GRCh38
rs777515592 Chr.14: 39267493-39267493 on Build GRCh38
rs943454203 Chr.14: 39267842-39267842 on Build GRCh38
rs959151321 Chr.14: 39267924-39267924 on Build GRCh38
rs1007542706 Chr.14: 39267570-39267570 on Build GRCh38
rs532900848 Chr.14: 39267817-39267817 on Build GRCh38
rs764716632 Chr.14: 39267472-39267472 on Build GRCh38
rs747215161 Chr.14: 39267516-39267516 on Build GRCh38
rs762155247 Chr.14: 39267537-39267537 on Build GRCh38
rs1051924332 Chr.14: 39267826-39267826 on Build GRCh38
rs1032629745 Chr.14: 39267706-39267706 on Build GRCh38
rs906381541 Chr.14: 39267909-39267909 on Build GRCh38
rs377393410 Chr.14: 39267912-39267912 on Build GRCh38
rs369916190 Chr.14: 39267714-39267714 on Build GRCh38
rs969569654 Chr.14: 39267632-39267632 on Build GRCh38
rs1021268917 Chr.14: 39267899-39267899 on Build GRCh38
rs543128509 Chr.14: 39267652-39267652 on Build GRCh38
rs778574923 Chr.14: 39267736-39267736 on Build GRCh38
rs548606333 Chr.14: 39267851-39267851 on Build GRCh38
rs746353205 Chr.14: 39267608-39267608 on Build GRCh38
rs1052256828 Chr.14: 39267539-39267539 on Build GRCh38
rs759175723 Chr.14: 39267556-39267556 on Build GRCh38
rs754807691 Chr.14: 39267481-39267481 on Build GRCh38
rs376664088 Chr.14: 39267544-39267544 on Build GRCh38
rs113934008 Chr.14: 39267551-39267551 on Build GRCh38
rs757843486 Chr.14: 39267475-39267475 on Build GRCh38
rs753798108 Chr.14: 39267574-39267574 on Build GRCh38
rs779833957 Chr.14: 39267494-39267495 on Build GRCh38
rs780467159 Chr.14: 39267501-39267501 on Build GRCh38
rs752268603 Chr.14: 39267777-39267777 on Build GRCh38
rs767011478 Chr.14: 39267557-39267557 on Build GRCh38
rs751171248 Chr.14: 39267480-39267480 on Build GRCh38
rs562148288 Chr.14: 39267667-39267667 on Build GRCh38
rs777045235 Chr.14: 39267518-39267518 on Build GRCh38
rs942265410 Chr.14: 39267789-39267789 on Build GRCh38
rs34909359 Chr.14: 39267489-39267489 on Build GRCh38
rs750734999 Chr.14: 39267654-39267654 on Build GRCh38
rs554607340 Chr.14: 39267828-39267828 on Build GRCh38
rs752551102 Chr.14: 39267558-39267558 on Build GRCh38
rs980923807 Chr.14: 39267642-39267642 on Build GRCh38
rs369514422 Chr.14: 39267548-39267548 on Build GRCh38
rs889127210 Chr.14: 39267540-39267540 on Build GRCh38
rs542282969 Chr.14: 39267616-39267616 on Build GRCh38
rs751537355 Chr.14: 39267546-39267547 on Build GRCh38
rs958529230 Chr.14: 39267732-39267732 on Build GRCh38
rs983815004 Chr.14: 39267763-39267763 on Build GRCh38
rs942243918 Chr.14: 39267486-39267486 on Build GRCh38
rs1003923861 Chr.14: 39267612-39267612 on Build GRCh38
rs1044197949 Chr.14: 39267879-39267879 on Build GRCh38
rs147380961 Chr.14: 39267756-39267756 on Build GRCh38
rs560005506 Chr.14: 39267579-39267579 on Build GRCh38

View Full Product Details