Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39276844-39277243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE5
Entrez Gene ID
4253
Gene Name
CTAGE family member 5, ER export factor
Gene Aliases
MEA6, MGEA, MGEA11, MGEA6
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265264-39351193 on Build GRCh38
Transcripts
NM_001247988.1 NM_001247989.1 NM_001247990.1 NM_005930.3 NM_203354.2 NM_203355.2 NM_203356.2 XM_005267648.4 XM_006720148.2 XM_011536775.2 XM_011536778.2 XM_011536784.2 XM_011536785.2 XM_017021314.1 XM_017021315.1 XM_017021316.1 XM_017021317.1 XM_017021318.1 XM_017021319.1 XM_017021320.1 XM_017021321.1 XM_017021322.1 XM_017021323.1 XM_017021324.1 XM_017021325.1 XM_017021326.1 XM_017021327.1 XM_017021328.1 XM_017021329.1 XM_017021330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6039858 c.118_119insT p.A42fs*11 Chr.14: 39277024-39277025 on Build GRCh38
4675274 c.169T>C p.F57L Chr.14: 39277039-39277039 on Build GRCh38
7334444 c.127T>C p.L43L Chr.14: 39276997-39276997 on Build GRCh38
297750 c.194C>T p.S65L Chr.14: 39277064-39277064 on Build GRCh38
4675276 c.169T>C p.F57L Chr.14: 39277039-39277039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531885144 Chr.14: 39277134-39277134 on Build GRCh38
rs745417023 Chr.14: 39276998-39276998 on Build GRCh38
rs539741772 Chr.14: 39277171-39277172 on Build GRCh38
rs1009142674 Chr.14: 39277136-39277136 on Build GRCh38
rs956941577 Chr.14: 39276955-39276955 on Build GRCh38
rs998971529 Chr.14: 39277028-39277028 on Build GRCh38
rs774792407 Chr.14: 39276996-39276996 on Build GRCh38
rs751698567 Chr.14: 39276979-39276979 on Build GRCh38
rs746341500 Chr.14: 39277098-39277098 on Build GRCh38
rs572052017 Chr.14: 39277205-39277206 on Build GRCh38
rs1057356689 Chr.14: 39276899-39276899 on Build GRCh38
rs760234223 Chr.14: 39277007-39277007 on Build GRCh38
rs753888024 Chr.14: 39276972-39276972 on Build GRCh38
rs372943436 Chr.14: 39276911-39276911 on Build GRCh38
rs370296546 Chr.14: 39276964-39276964 on Build GRCh38
rs778096789 Chr.14: 39276989-39276989 on Build GRCh38
rs754685760 Chr.14: 39276911-39276914 on Build GRCh38
rs769553721 Chr.14: 39277039-39277039 on Build GRCh38
rs767223451 Chr.14: 39276944-39276944 on Build GRCh38
rs561013872 Chr.14: 39276990-39276990 on Build GRCh38
rs771453708 Chr.14: 39277002-39277002 on Build GRCh38
rs776576456 Chr.14: 39277009-39277009 on Build GRCh38
rs756285984 Chr.14: 39277045-39277045 on Build GRCh38
rs1047042210 Chr.14: 39277184-39277184 on Build GRCh38
rs748285391 Chr.14: 39276983-39276983 on Build GRCh38
rs747741991 Chr.14: 39277014-39277016 on Build GRCh38
rs775259334 Chr.14: 39277006-39277006 on Build GRCh38
rs749041023 Chr.14: 39277000-39277000 on Build GRCh38
rs199650703 Chr.14: 39277044-39277044 on Build GRCh38
rs535569584 Chr.14: 39276921-39276921 on Build GRCh38
rs771441132 Chr.14: 39276994-39276994 on Build GRCh38
rs750592264 Chr.14: 39277096-39277096 on Build GRCh38
rs781210219 Chr.14: 39276898-39276898 on Build GRCh38
rs192992134 Chr.14: 39276876-39276876 on Build GRCh38
rs781505966 Chr.14: 39276981-39276981 on Build GRCh38
rs757579796 Chr.14: 39277075-39277075 on Build GRCh38
rs760897982 Chr.14: 39277215-39277215 on Build GRCh38
rs377514735 Chr.14: 39277008-39277008 on Build GRCh38
rs1008112771 Chr.14: 39277207-39277207 on Build GRCh38
rs370813465 Chr.14: 39276884-39276884 on Build GRCh38
rs761638597 Chr.14: 39277010-39277010 on Build GRCh38
rs931342414 Chr.14: 39277121-39277121 on Build GRCh38
rs201398321 Chr.14: 39276977-39276977 on Build GRCh38
rs771673040 Chr.14: 39277099-39277099 on Build GRCh38
rs758914530 Chr.14: 39276942-39276942 on Build GRCh38
rs765284464 Chr.14: 39277025-39277025 on Build GRCh38
rs750344019 Chr.14: 39277032-39277032 on Build GRCh38
rs774861205 Chr.14: 39277108-39277108 on Build GRCh38
rs894734283 Chr.14: 39276945-39276945 on Build GRCh38
rs773855574 Chr.14: 39276934-39276934 on Build GRCh38
rs770486195 Chr.14: 39276925-39276925 on Build GRCh38
rs755123186 Chr.14: 39277037-39277037 on Build GRCh38
rs773746975 Chr.14: 39276916-39276916 on Build GRCh38
rs747306756 Chr.14: 39276893-39276893 on Build GRCh38
rs779437727 Chr.14: 39277080-39277080 on Build GRCh38
rs78536283 Chr.14: 39277038-39277038 on Build GRCh38
rs75842899 Chr.14: 39277039-39277039 on Build GRCh38
rs920282001 Chr.14: 39277187-39277187 on Build GRCh38
rs185291670 Chr.14: 39277064-39277064 on Build GRCh38
rs61740767 Chr.14: 39276988-39276988 on Build GRCh38
rs768922866 Chr.14: 39276897-39276897 on Build GRCh38
rs572080267 Chr.14: 39276892-39276892 on Build GRCh38
rs375950626 Chr.14: 39276923-39276923 on Build GRCh38
rs757259755 Chr.14: 39276976-39276976 on Build GRCh38
rs556016740 Chr.14: 39277186-39277192 on Build GRCh38
rs924225577 Chr.14: 39276924-39276924 on Build GRCh38
rs374472059 Chr.14: 39277205-39277211 on Build GRCh38
rs760560406 Chr.14: 39276965-39276965 on Build GRCh38
rs887063772 Chr.14: 39277192-39277192 on Build GRCh38
rs563175728 Chr.14: 39277125-39277125 on Build GRCh38
rs748564629 Chr.14: 39276909-39276909 on Build GRCh38
rs938840923 Chr.14: 39276869-39276869 on Build GRCh38

View Full Product Details