Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39279323-39279802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE5
Entrez Gene ID
4253
Gene Name
CTAGE family member 5, ER export factor
Gene Aliases
MEA6, MGEA, MGEA11, MGEA6
UniGene
Hs.509200
Gene Chromosome Location
Chr.14: 39265264-39351193 on Build GRCh38
Transcripts
NM_001247988.1 NM_001247989.1 NM_001247990.1 NM_005930.3 NM_203354.2 NM_203355.2 NM_203356.2 XM_005267648.4 XM_006720148.2 XM_011536775.2 XM_011536778.2 XM_011536784.2 XM_011536785.2 XM_017021314.1 XM_017021315.1 XM_017021316.1 XM_017021317.1 XM_017021318.1 XM_017021319.1 XM_017021320.1 XM_017021321.1 XM_017021322.1 XM_017021323.1 XM_017021324.1 XM_017021325.1 XM_017021326.1 XM_017021327.1 XM_017021328.1 XM_017021329.1 XM_017021330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4453391 c.285A>T p.K95N Chr.14: 39279516-39279516 on Build GRCh38
5705695 c.170G>A p.R57Q Chr.14: 39279347-39279347 on Build GRCh38
3419797 c.225G>T p.E75D Chr.14: 39279456-39279456 on Build GRCh38
7180563 c.212C>G p.S71C Chr.14: 39279479-39279479 on Build GRCh38
4453390 c.249A>T p.K83N Chr.14: 39279516-39279516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755538803 Chr.14: 39279572-39279572 on Build GRCh38
rs150107086 Chr.14: 39279618-39279618 on Build GRCh38
rs751002901 Chr.14: 39279359-39279359 on Build GRCh38
rs758862204 Chr.14: 39279446-39279446 on Build GRCh38
rs764135087 Chr.14: 39279426-39279426 on Build GRCh38
rs370129642 Chr.14: 39279524-39279524 on Build GRCh38
rs762742828 Chr.14: 39279405-39279405 on Build GRCh38
rs781035258 Chr.14: 39279396-39279396 on Build GRCh38
rs201029778 Chr.14: 39279390-39279390 on Build GRCh38
rs556297014 Chr.14: 39279536-39279536 on Build GRCh38
rs563609129 Chr.14: 39279771-39279771 on Build GRCh38
rs79293806 Chr.14: 39279585-39279585 on Build GRCh38
rs371643419 Chr.14: 39279442-39279442 on Build GRCh38
rs752078931 Chr.14: 39279463-39279463 on Build GRCh38
rs138671940 Chr.14: 39279484-39279484 on Build GRCh38
rs776886067 Chr.14: 39279558-39279566 on Build GRCh38
rs777425988 Chr.14: 39279471-39279471 on Build GRCh38
rs574732319 Chr.14: 39279725-39279725 on Build GRCh38
rs771122819 Chr.14: 39279388-39279388 on Build GRCh38
rs761095444 Chr.14: 39279514-39279515 on Build GRCh38
rs750762725 Chr.14: 39279452-39279452 on Build GRCh38
rs201213535 Chr.14: 39279411-39279411 on Build GRCh38
rs747878553 Chr.14: 39279469-39279469 on Build GRCh38
rs777931342 Chr.14: 39279635-39279635 on Build GRCh38
rs1024899070 Chr.14: 39279609-39279609 on Build GRCh38
rs543832787 Chr.14: 39279768-39279768 on Build GRCh38
rs555289302 Chr.14: 39279721-39279721 on Build GRCh38
rs932353978 Chr.14: 39279479-39279479 on Build GRCh38
rs922927374 Chr.14: 39279755-39279755 on Build GRCh38
rs572313254 Chr.14: 39279705-39279705 on Build GRCh38
rs746527532 Chr.14: 39279384-39279384 on Build GRCh38
rs775821116 Chr.14: 39279440-39279440 on Build GRCh38
rs143110568 Chr.14: 39279356-39279356 on Build GRCh38
rs867905550 Chr.14: 39279454-39279455 on Build GRCh38
rs951907020 Chr.14: 39279672-39279672 on Build GRCh38
rs909411963 Chr.14: 39279777-39279777 on Build GRCh38
rs765430399 Chr.14: 39279431-39279431 on Build GRCh38
rs541279704 Chr.14: 39279708-39279708 on Build GRCh38
rs913803666 Chr.14: 39279665-39279665 on Build GRCh38
rs1005994793 Chr.14: 39279685-39279685 on Build GRCh38
rs546451296 Chr.14: 39279382-39279383 on Build GRCh38
rs961817463 Chr.14: 39279722-39279722 on Build GRCh38
rs539789900 Chr.14: 39279383-39279383 on Build GRCh38
rs762015106 Chr.14: 39279429-39279429 on Build GRCh38
rs1013543303 Chr.14: 39279601-39279601 on Build GRCh38
rs754586323 Chr.14: 39279467-39279467 on Build GRCh38
rs772267235 Chr.14: 39279431-39279435 on Build GRCh38
rs537357810 Chr.14: 39279387-39279389 on Build GRCh38
rs772879462 Chr.14: 39279404-39279404 on Build GRCh38
rs373911136 Chr.14: 39279556-39279556 on Build GRCh38
rs757665840 Chr.14: 39279355-39279355 on Build GRCh38
rs749068286 Chr.14: 39279475-39279475 on Build GRCh38
rs747782667 Chr.14: 39279398-39279398 on Build GRCh38
rs774202058 Chr.14: 39279483-39279483 on Build GRCh38
rs780461402 Chr.14: 39279459-39279459 on Build GRCh38
rs766747239 Chr.14: 39279567-39279567 on Build GRCh38
rs1048601200 Chr.14: 39279761-39279761 on Build GRCh38
rs1049927921 Chr.14: 39279450-39279450 on Build GRCh38
rs201691215 Chr.14: 39279400-39279400 on Build GRCh38
rs750623613 Chr.14: 39279542-39279542 on Build GRCh38
rs988447518 Chr.14: 39279413-39279413 on Build GRCh38
rs780839007 Chr.14: 39279468-39279468 on Build GRCh38
rs754114577 Chr.14: 39279428-39279428 on Build GRCh38
rs976424664 Chr.14: 39279686-39279686 on Build GRCh38
rs764268145 Chr.14: 39279516-39279517 on Build GRCh38
rs1034167076 Chr.14: 39279662-39279662 on Build GRCh38
rs776693445 Chr.14: 39279523-39279523 on Build GRCh38
rs369990672 Chr.14: 39279526-39279526 on Build GRCh38
rs760782588 Chr.14: 39279422-39279422 on Build GRCh38
rs936088497 Chr.14: 39279512-39279512 on Build GRCh38
rs748786921 Chr.14: 39279376-39279377 on Build GRCh38
rs761750235 Chr.14: 39279575-39279576 on Build GRCh38
rs759453959 Chr.14: 39279489-39279489 on Build GRCh38
rs770726747 Chr.14: 39279476-39279476 on Build GRCh38
rs768886345 Chr.14: 39279718-39279718 on Build GRCh38
rs768787526 Chr.14: 39279521-39279521 on Build GRCh38
rs920904464 Chr.14: 39279699-39279699 on Build GRCh38
rs955826902 Chr.14: 39279770-39279770 on Build GRCh38
rs758745521 Chr.14: 39279458-39279458 on Build GRCh38
rs532779690 Chr.14: 39279408-39279410 on Build GRCh38
rs138478920 Chr.14: 39279624-39279624 on Build GRCh38
rs375402435 Chr.14: 39279408-39279408 on Build GRCh38
rs988111684 Chr.14: 39279663-39279663 on Build GRCh38
rs752172213 Chr.14: 39279569-39279569 on Build GRCh38
rs752270100 Chr.14: 39279588-39279588 on Build GRCh38

View Full Product Details