Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39396637-39397148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO33
Entrez Gene ID
254170
Gene Name
F-box protein 33
Gene Aliases
BMND12, Fbx33, c14_5247
UniGene
Hs.146632
Gene Chromosome Location
Chr.14: 39396373-39432500 on Build GRCh38
Transcripts
NM_203301.3

SNP Detail

SNP ID Chromosome Location
rs748319475 Chr.14: 39396835-39396835 on Build GRCh38
rs574504043 Chr.14: 39396905-39396905 on Build GRCh38
rs886143712 Chr.14: 39397021-39397021 on Build GRCh38
rs1014814202 Chr.14: 39397098-39397098 on Build GRCh38
rs541037446 Chr.14: 39396788-39396788 on Build GRCh38
rs994602358 Chr.14: 39396945-39396945 on Build GRCh38
rs554340874 Chr.14: 39396801-39396801 on Build GRCh38
rs888690242 Chr.14: 39396971-39396971 on Build GRCh38
rs948115674 Chr.14: 39396779-39396779 on Build GRCh38
rs562783826 Chr.14: 39397025-39397025 on Build GRCh38
rs773208946 Chr.14: 39396882-39396882 on Build GRCh38
rs890255375 Chr.14: 39397091-39397091 on Build GRCh38
rs868256382 Chr.14: 39397032-39397032 on Build GRCh38
rs146811633 Chr.14: 39397084-39397084 on Build GRCh38
rs761238093 Chr.14: 39397007-39397007 on Build GRCh38
rs928122368 Chr.14: 39396873-39396873 on Build GRCh38
rs1048719261 Chr.14: 39396980-39396980 on Build GRCh38
rs958018848 Chr.14: 39396757-39396757 on Build GRCh38
rs191116878 Chr.14: 39396786-39396786 on Build GRCh38
rs143657301 Chr.14: 39396773-39396773 on Build GRCh38
rs960216770 Chr.14: 39397072-39397072 on Build GRCh38
rs944568682 Chr.14: 39396872-39396872 on Build GRCh38
rs999762547 Chr.14: 39397104-39397104 on Build GRCh38
rs978001103 Chr.14: 39396671-39396671 on Build GRCh38
rs531882354 Chr.14: 39397068-39397068 on Build GRCh38
rs563185602 Chr.14: 39396847-39396847 on Build GRCh38
rs569949711 Chr.14: 39396707-39396707 on Build GRCh38
rs1030501925 Chr.14: 39397069-39397069 on Build GRCh38
rs140034930 Chr.14: 39396760-39396760 on Build GRCh38
rs900185300 Chr.14: 39397059-39397059 on Build GRCh38
rs368189159 Chr.14: 39396903-39396903 on Build GRCh38
rs779077927 Chr.14: 39396694-39396694 on Build GRCh38
rs1046479565 Chr.14: 39396822-39396822 on Build GRCh38
rs1047268527 Chr.14: 39396912-39396912 on Build GRCh38
rs954712439 Chr.14: 39396661-39396661 on Build GRCh38
rs565584099 Chr.14: 39397116-39397116 on Build GRCh38
rs542798918 Chr.14: 39397018-39397018 on Build GRCh38
rs772152104 Chr.14: 39396875-39396875 on Build GRCh38
rs769324637 Chr.14: 39396964-39396964 on Build GRCh38

View Full Product Details