Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39401387-39401897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO33
Entrez Gene ID
254170
Gene Name
F-box protein 33
Gene Aliases
BMND12, Fbx33, c14_5247
UniGene
Hs.146632
Gene Chromosome Location
Chr.14: 39396373-39432500 on Build GRCh38
Transcripts
NM_203301.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3968811 c.1137C>T p.I379I Chr.14: 39401435-39401435 on Build GRCh38
7222104 c.1072G>C p.E358Q Chr.14: 39401500-39401500 on Build GRCh38
5612588 c.1170C>T p.P390P Chr.14: 39401402-39401402 on Build GRCh38
5567901 c.802T>C p.S268P Chr.14: 39401770-39401770 on Build GRCh38
381927 c.947G>T p.C316F Chr.14: 39401625-39401625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772708113 Chr.14: 39401630-39401630 on Build GRCh38
rs542813352 Chr.14: 39401773-39401773 on Build GRCh38
rs778790009 Chr.14: 39401749-39401749 on Build GRCh38
rs987000598 Chr.14: 39401753-39401753 on Build GRCh38
rs755349496 Chr.14: 39401765-39401765 on Build GRCh38
rs980132796 Chr.14: 39401735-39401735 on Build GRCh38
rs777947116 Chr.14: 39401588-39401588 on Build GRCh38
rs367829584 Chr.14: 39401583-39401583 on Build GRCh38
rs1027797772 Chr.14: 39401818-39401818 on Build GRCh38
rs904377528 Chr.14: 39401754-39401754 on Build GRCh38
rs368690413 Chr.14: 39401864-39401864 on Build GRCh38
rs748313600 Chr.14: 39401471-39401471 on Build GRCh38
rs770181339 Chr.14: 39401862-39401862 on Build GRCh38
rs762817650 Chr.14: 39401654-39401654 on Build GRCh38
rs758603898 Chr.14: 39401760-39401760 on Build GRCh38
rs61754303 Chr.14: 39401528-39401528 on Build GRCh38
rs144987416 Chr.14: 39401435-39401435 on Build GRCh38
rs141687934 Chr.14: 39401742-39401742 on Build GRCh38
rs750279892 Chr.14: 39401555-39401555 on Build GRCh38
rs772786157 Chr.14: 39401684-39401684 on Build GRCh38
rs201684312 Chr.14: 39401800-39401800 on Build GRCh38
rs1039691239 Chr.14: 39401767-39401767 on Build GRCh38
rs776110676 Chr.14: 39401670-39401670 on Build GRCh38
rs897896140 Chr.14: 39401802-39401802 on Build GRCh38
rs557650803 Chr.14: 39401421-39401421 on Build GRCh38
rs758517059 Chr.14: 39401866-39401866 on Build GRCh38
rs758089884 Chr.14: 39401460-39401460 on Build GRCh38
rs766592457 Chr.14: 39401871-39401871 on Build GRCh38
rs143145457 Chr.14: 39401574-39401574 on Build GRCh38
rs751828118 Chr.14: 39401680-39401683 on Build GRCh38
rs761999715 Chr.14: 39401771-39401771 on Build GRCh38
rs775357641 Chr.14: 39401825-39401825 on Build GRCh38
rs114177972 Chr.14: 39401682-39401682 on Build GRCh38
rs779256618 Chr.14: 39401495-39401495 on Build GRCh38
rs747554293 Chr.14: 39401763-39401763 on Build GRCh38
rs761549687 Chr.14: 39401676-39401676 on Build GRCh38
rs757242515 Chr.14: 39401606-39401606 on Build GRCh38
rs370540187 Chr.14: 39401544-39401544 on Build GRCh38
rs769187449 Chr.14: 39401666-39401666 on Build GRCh38
rs149090376 Chr.14: 39401529-39401529 on Build GRCh38
rs910845249 Chr.14: 39401645-39401645 on Build GRCh38
rs968265106 Chr.14: 39401723-39401723 on Build GRCh38
rs942383934 Chr.14: 39401661-39401661 on Build GRCh38
rs772511993 Chr.14: 39401514-39401514 on Build GRCh38
rs371768048 Chr.14: 39401739-39401739 on Build GRCh38
rs7156962 Chr.14: 39401813-39401813 on Build GRCh38
rs771092232 Chr.14: 39401485-39401485 on Build GRCh38
rs1025466805 Chr.14: 39401543-39401543 on Build GRCh38
rs748773357 Chr.14: 39401784-39401784 on Build GRCh38
rs746414224 Chr.14: 39401511-39401511 on Build GRCh38
rs770466131 Chr.14: 39401798-39401798 on Build GRCh38
rs759490824 Chr.14: 39401807-39401807 on Build GRCh38
rs886931892 Chr.14: 39401804-39401804 on Build GRCh38
rs751545108 Chr.14: 39401726-39401726 on Build GRCh38
rs759294681 Chr.14: 39401734-39401734 on Build GRCh38
rs747689922 Chr.14: 39401775-39401775 on Build GRCh38
rs201517133 Chr.14: 39401564-39401564 on Build GRCh38
rs772799613 Chr.14: 39401781-39401781 on Build GRCh38
rs778035293 Chr.14: 39401478-39401478 on Build GRCh38
rs779846906 Chr.14: 39401461-39401461 on Build GRCh38
rs942764033 Chr.14: 39401755-39401755 on Build GRCh38
rs754129378 Chr.14: 39401599-39401599 on Build GRCh38
rs754824176 Chr.14: 39401586-39401586 on Build GRCh38
rs539795896 Chr.14: 39401526-39401526 on Build GRCh38
rs375157529 Chr.14: 39401552-39401552 on Build GRCh38
rs766097493 Chr.14: 39401579-39401579 on Build GRCh38
rs144152967 Chr.14: 39401417-39401417 on Build GRCh38
rs766288233 Chr.14: 39401713-39401713 on Build GRCh38
rs753286646 Chr.14: 39401439-39401439 on Build GRCh38
rs775239655 Chr.14: 39401434-39401434 on Build GRCh38
rs575056327 Chr.14: 39401834-39401834 on Build GRCh38
rs17852779 Chr.14: 39401581-39401581 on Build GRCh38
rs756852751 Chr.14: 39401447-39401447 on Build GRCh38
rs147025084 Chr.14: 39401743-39401743 on Build GRCh38
rs750418473 Chr.14: 39401454-39401454 on Build GRCh38
rs571077291 Chr.14: 39401430-39401430 on Build GRCh38
rs780767686 Chr.14: 39401641-39401641 on Build GRCh38
rs922362702 Chr.14: 39401733-39401733 on Build GRCh38
rs769484889 Chr.14: 39401776-39401776 on Build GRCh38
rs762518282 Chr.14: 39401689-39401689 on Build GRCh38
rs746300679 Chr.14: 39401618-39401618 on Build GRCh38
rs756335602 Chr.14: 39401472-39401472 on Build GRCh38
rs376006502 Chr.14: 39401762-39401762 on Build GRCh38
rs749313472 Chr.14: 39401483-39401483 on Build GRCh38
rs752055626 Chr.14: 39401873-39401873 on Build GRCh38
rs754970812 Chr.14: 39401689-39401691 on Build GRCh38
rs779068540 Chr.14: 39401616-39401616 on Build GRCh38
rs765068524 Chr.14: 39401452-39401452 on Build GRCh38

View Full Product Details