Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44906861-44907404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf28
Entrez Gene ID
122525
Gene Name
chromosome 14 open reading frame 28
Gene Aliases
DRIP-1, DRIP1, c14_5270
UniGene
Hs.82098
Gene Chromosome Location
Chr.14: 44897304-44907257 on Build GRCh38
Transcripts
NM_001017923.1 XM_011536408.1 NR_110050.1

Gene Symbol
LOC101927418
Entrez Gene ID
101927418
Gene Name
uncharacterized LOC101927418
Gene Aliases
-
UniGene
Hs.144809
Gene Chromosome Location
Chr.14: 44898908-44911917 on Build GRCh38
Transcripts
NM_001017923.1 XM_011536408.1 NR_110050.1

SNP Detail

SNP ID Chromosome Location
rs1028588506 Chr.14: 44907083-44907083 on Build GRCh38
rs142316797 Chr.14: 44907210-44907210 on Build GRCh38
rs1021802756 Chr.14: 44907174-44907174 on Build GRCh38
rs992923628 Chr.14: 44907218-44907218 on Build GRCh38
rs564301656 Chr.14: 44907140-44907140 on Build GRCh38
rs1013835247 Chr.14: 44907353-44907353 on Build GRCh38
rs575664856 Chr.14: 44907159-44907159 on Build GRCh38
rs766317579 Chr.14: 44906892-44906892 on Build GRCh38
rs1031933479 Chr.14: 44906977-44906977 on Build GRCh38
rs45590239 Chr.14: 44907203-44907203 on Build GRCh38
rs1055603985 Chr.14: 44907273-44907273 on Build GRCh38
rs763857217 Chr.14: 44907052-44907052 on Build GRCh38
rs940788567 Chr.14: 44907263-44907263 on Build GRCh38
rs1037549584 Chr.14: 44907063-44907063 on Build GRCh38
rs550453199 Chr.14: 44907078-44907078 on Build GRCh38
rs1008631331 Chr.14: 44907098-44907098 on Build GRCh38
rs929507285 Chr.14: 44907163-44907163 on Build GRCh38
rs927295987 Chr.14: 44906914-44906914 on Build GRCh38
rs200118802 Chr.14: 44906933-44906934 on Build GRCh38
rs562673536 Chr.14: 44907112-44907112 on Build GRCh38
rs558920366 Chr.14: 44907020-44907020 on Build GRCh38
rs965229800 Chr.14: 44907043-44907045 on Build GRCh38
rs1013446353 Chr.14: 44906911-44906914 on Build GRCh38
rs1037916174 Chr.14: 44907288-44907288 on Build GRCh38
rs890150270 Chr.14: 44907095-44907095 on Build GRCh38
rs879632027 Chr.14: 44906922-44906922 on Build GRCh38
rs895264446 Chr.14: 44907323-44907323 on Build GRCh38
rs758634715 Chr.14: 44907033-44907033 on Build GRCh38
rs577109621 Chr.14: 44907040-44907040 on Build GRCh38
rs1031210810 Chr.14: 44907130-44907130 on Build GRCh38
rs956223133 Chr.14: 44907147-44907147 on Build GRCh38
rs968933460 Chr.14: 44907198-44907198 on Build GRCh38
rs1010784383 Chr.14: 44907164-44907164 on Build GRCh38
rs867145651 Chr.14: 44907232-44907232 on Build GRCh38
rs910998154 Chr.14: 44907041-44907041 on Build GRCh38
rs957715490 Chr.14: 44907018-44907018 on Build GRCh38
rs899049095 Chr.14: 44907073-44907073 on Build GRCh38
rs547058027 Chr.14: 44907211-44907211 on Build GRCh38
rs752963387 Chr.14: 44906928-44906928 on Build GRCh38
rs980362325 Chr.14: 44907202-44907202 on Build GRCh38
rs1057469814 Chr.14: 44906933-44906933 on Build GRCh38

View Full Product Details