Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 45153939-45154488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCM
Entrez Gene ID
57697
Gene Name
Fanconi anemia complementation group M
Gene Aliases
FAAP250, KIAA1596
UniGene
Hs.509229
Gene Chromosome Location
Chr.14: 45135939-45200890 on Build GRCh38
Transcripts
NM_001308133.1 NM_001308134.1 NM_020937.3 XM_011537034.2 XM_011537035.2 XM_017021523.1 XM_017021524.1 XM_017021525.1 XM_017021526.1 XM_017021527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316447 c.1139G>T p.R380I Chr.14: 45154008-45154008 on Build GRCh38
4440363 c.1137G>A p.M379I Chr.14: 45154006-45154006 on Build GRCh38
7169781 c.1142C>T p.S381L Chr.14: 45154011-45154011 on Build GRCh38
2197450 c.1100T>C p.L367S Chr.14: 45153969-45153969 on Build GRCh38
3968819 c.1128A>G p.Q376Q Chr.14: 45153997-45153997 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs184932864 Chr.14: 45154309-45154309 on Build GRCh38
rs758870757 Chr.14: 45154015-45154015 on Build GRCh38
rs541414382 Chr.14: 45154290-45154290 on Build GRCh38
rs939252723 Chr.14: 45154169-45154169 on Build GRCh38
rs770726399 Chr.14: 45153989-45153989 on Build GRCh38
rs768830100 Chr.14: 45154104-45154104 on Build GRCh38
rs982873560 Chr.14: 45154174-45154174 on Build GRCh38
rs1009129820 Chr.14: 45154274-45154274 on Build GRCh38
rs908183681 Chr.14: 45154198-45154198 on Build GRCh38
rs373869932 Chr.14: 45154077-45154077 on Build GRCh38
rs150497733 Chr.14: 45154273-45154273 on Build GRCh38
rs1022389198 Chr.14: 45154358-45154358 on Build GRCh38
rs5808292 Chr.14: 45154399-45154399 on Build GRCh38
rs892461701 Chr.14: 45154316-45154316 on Build GRCh38
rs927688359 Chr.14: 45154248-45154248 on Build GRCh38
rs538172189 Chr.14: 45154060-45154060 on Build GRCh38
rs980558877 Chr.14: 45154107-45154107 on Build GRCh38
rs369985517 Chr.14: 45154075-45154075 on Build GRCh38
rs754592572 Chr.14: 45154024-45154024 on Build GRCh38
rs776630034 Chr.14: 45154003-45154003 on Build GRCh38
rs1050583328 Chr.14: 45154270-45154270 on Build GRCh38
rs961956621 Chr.14: 45154201-45154201 on Build GRCh38
rs761915062 Chr.14: 45154006-45154006 on Build GRCh38
rs1009322588 Chr.14: 45154061-45154061 on Build GRCh38
rs201627169 Chr.14: 45154155-45154155 on Build GRCh38
rs774077329 Chr.14: 45153994-45153994 on Build GRCh38
rs200621018 Chr.14: 45154084-45154084 on Build GRCh38
rs192890750 Chr.14: 45154211-45154211 on Build GRCh38
rs773303797 Chr.14: 45153974-45153974 on Build GRCh38
rs941090578 Chr.14: 45153966-45153966 on Build GRCh38
rs1002049330 Chr.14: 45154446-45154446 on Build GRCh38
rs774133782 Chr.14: 45154086-45154086 on Build GRCh38
rs755662028 Chr.14: 45154074-45154074 on Build GRCh38
rs747611390 Chr.14: 45154064-45154064 on Build GRCh38
rs1029198668 Chr.14: 45154047-45154047 on Build GRCh38
rs535598993 Chr.14: 45154182-45154182 on Build GRCh38
rs980599994 Chr.14: 45154237-45154237 on Build GRCh38
rs1043684485 Chr.14: 45154363-45154363 on Build GRCh38
rs1034835241 Chr.14: 45154455-45154455 on Build GRCh38
rs576821098 Chr.14: 45154452-45154452 on Build GRCh38
rs762845761 Chr.14: 45153984-45153984 on Build GRCh38
rs941142547 Chr.14: 45154200-45154200 on Build GRCh38
rs376481889 Chr.14: 45154001-45154001 on Build GRCh38
rs151005385 Chr.14: 45154379-45154379 on Build GRCh38
rs760935075 Chr.14: 45154000-45154000 on Build GRCh38
rs867620907 Chr.14: 45154355-45154355 on Build GRCh38
rs375950184 Chr.14: 45154341-45154341 on Build GRCh38
rs555334829 Chr.14: 45154325-45154325 on Build GRCh38
rs751927264 Chr.14: 45154020-45154020 on Build GRCh38
rs747666389 Chr.14: 45153967-45153967 on Build GRCh38
rs372836421 Chr.14: 45154319-45154319 on Build GRCh38
rs1027609694 Chr.14: 45154224-45154224 on Build GRCh38
rs370944110 Chr.14: 45154091-45154091 on Build GRCh38
rs766761905 Chr.14: 45154016-45154016 on Build GRCh38
rs765686839 Chr.14: 45154082-45154082 on Build GRCh38
rs927877063 Chr.14: 45154166-45154166 on Build GRCh38
rs371607057 Chr.14: 45154204-45154204 on Build GRCh38
rs932110355 Chr.14: 45154399-45154399 on Build GRCh38
rs969606692 Chr.14: 45154233-45154233 on Build GRCh38
rs754930910 Chr.14: 45154066-45154066 on Build GRCh38
rs769814213 Chr.14: 45153969-45153969 on Build GRCh38
rs1969713 Chr.14: 45154257-45154257 on Build GRCh38
rs765686151 Chr.14: 45154011-45154011 on Build GRCh38
rs899327482 Chr.14: 45153991-45153991 on Build GRCh38
rs750921743 Chr.14: 45154012-45154012 on Build GRCh38
rs1015343891 Chr.14: 45154186-45154186 on Build GRCh38
rs969426031 Chr.14: 45154364-45154364 on Build GRCh38

View Full Product Details