Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50312927-50313461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5S
Entrez Gene ID
27109
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B)
Gene Aliases
ATPW, FB, HSU79253
UniGene
Hs.438489
Gene Chromosome Location
Chr.14: 50311752-50326715 on Build GRCh38
Transcripts
NM_001003803.2 NM_001003805.2 NM_015684.3 NR_033761.1 XM_011536649.2 XM_011536650.2 XM_011536651.2 XM_011536653.2 XM_011536654.2 XM_011536655.2 XM_011536656.2 XM_017021220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027856 c.8G>C p.C3S Chr.14: 50313028-50313028 on Build GRCh38
7149258 c.15C>G p.V5V Chr.14: 50313035-50313035 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1036146198 Chr.14: 50313431-50313431 on Build GRCh38
rs955425607 Chr.14: 50313256-50313256 on Build GRCh38
rs553193121 Chr.14: 50313166-50313169 on Build GRCh38
rs937033807 Chr.14: 50313402-50313402 on Build GRCh38
rs4534749 Chr.14: 50313365-50313365 on Build GRCh38
rs755336408 Chr.14: 50312996-50312996 on Build GRCh38
rs770083077 Chr.14: 50312985-50312985 on Build GRCh38
rs990320497 Chr.14: 50313285-50313285 on Build GRCh38
rs113780995 Chr.14: 50312981-50312981 on Build GRCh38
rs765411242 Chr.14: 50313044-50313044 on Build GRCh38
rs73279414 Chr.14: 50313069-50313069 on Build GRCh38
rs766442781 Chr.14: 50313430-50313430 on Build GRCh38
rs751631376 Chr.14: 50313071-50313071 on Build GRCh38
rs774436343 Chr.14: 50313026-50313026 on Build GRCh38
rs568146374 Chr.14: 50313249-50313249 on Build GRCh38
rs892774612 Chr.14: 50313286-50313286 on Build GRCh38
rs1055503476 Chr.14: 50313280-50313280 on Build GRCh38
rs891571054 Chr.14: 50313066-50313066 on Build GRCh38
rs747976627 Chr.14: 50313407-50313407 on Build GRCh38
rs149992117 Chr.14: 50313406-50313406 on Build GRCh38
rs201035813 Chr.14: 50312998-50312998 on Build GRCh38
rs772122956 Chr.14: 50313029-50313029 on Build GRCh38
rs143698262 Chr.14: 50313271-50313271 on Build GRCh38
rs1045529921 Chr.14: 50313360-50313360 on Build GRCh38
rs1008030523 Chr.14: 50313351-50313351 on Build GRCh38
rs760490827 Chr.14: 50313042-50313042 on Build GRCh38
rs796932525 Chr.14: 50313133-50313133 on Build GRCh38
rs1055742582 Chr.14: 50313116-50313116 on Build GRCh38
rs371438000 Chr.14: 50313008-50313008 on Build GRCh38
rs879511703 Chr.14: 50313278-50313278 on Build GRCh38
rs1029406906 Chr.14: 50313087-50313087 on Build GRCh38
rs756459355 Chr.14: 50313104-50313104 on Build GRCh38
rs777311761 Chr.14: 50313105-50313105 on Build GRCh38
rs985269966 Chr.14: 50313223-50313223 on Build GRCh38
rs4534748 Chr.14: 50313305-50313305 on Build GRCh38
rs1020139041 Chr.14: 50312955-50312955 on Build GRCh38
rs887704508 Chr.14: 50313410-50313410 on Build GRCh38
rs748777968 Chr.14: 50313014-50313014 on Build GRCh38
rs543498519 Chr.14: 50313024-50313024 on Build GRCh38
rs1016934250 Chr.14: 50313164-50313164 on Build GRCh38
rs903036944 Chr.14: 50313334-50313334 on Build GRCh38
rs534223735 Chr.14: 50313265-50313265 on Build GRCh38
rs139241423 Chr.14: 50313399-50313399 on Build GRCh38
rs192355817 Chr.14: 50313046-50313046 on Build GRCh38
rs768241376 Chr.14: 50313398-50313398 on Build GRCh38
rs767989707 Chr.14: 50313078-50313078 on Build GRCh38
rs947185799 Chr.14: 50313307-50313307 on Build GRCh38
rs147225013 Chr.14: 50313275-50313275 on Build GRCh38
rs41301465 Chr.14: 50312971-50312971 on Build GRCh38
rs1045415559 Chr.14: 50313070-50313070 on Build GRCh38
rs773555240 Chr.14: 50313050-50313051 on Build GRCh38
rs113906741 Chr.14: 50313212-50313212 on Build GRCh38
rs985770183 Chr.14: 50312986-50312986 on Build GRCh38
rs139674067 Chr.14: 50313143-50313143 on Build GRCh38
rs201504821 Chr.14: 50312973-50312973 on Build GRCh38
rs759615653 Chr.14: 50313028-50313028 on Build GRCh38
rs545366715 Chr.14: 50312969-50312969 on Build GRCh38
rs753099725 Chr.14: 50313102-50313102 on Build GRCh38
rs1006027763 Chr.14: 50313152-50313152 on Build GRCh38
rs775479249 Chr.14: 50313040-50313040 on Build GRCh38
rs1011738042 Chr.14: 50312948-50312948 on Build GRCh38
rs41313503 Chr.14: 50313172-50313172 on Build GRCh38
rs908050082 Chr.14: 50313230-50313230 on Build GRCh38
rs200399707 Chr.14: 50313077-50313077 on Build GRCh38
rs535405079 Chr.14: 50313154-50313154 on Build GRCh38
rs1041829588 Chr.14: 50313323-50313323 on Build GRCh38
rs570547930 Chr.14: 50313064-50313064 on Build GRCh38
rs997150865 Chr.14: 50313435-50313435 on Build GRCh38
rs748499367 Chr.14: 50313001-50313001 on Build GRCh38
rs748526988 Chr.14: 50312975-50312976 on Build GRCh38
rs994247225 Chr.14: 50313225-50313225 on Build GRCh38
rs928273504 Chr.14: 50313391-50313391 on Build GRCh38
rs769380866 Chr.14: 50313004-50313004 on Build GRCh38

View Full Product Details