Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50334471-50335021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL1
Entrez Gene ID
8814
Gene Name
cyclin dependent kinase like 1
Gene Aliases
KKIALRE, P42
UniGene
Hs.280881
Gene Chromosome Location
Chr.14: 50329404-50397404 on Build GRCh38
Transcripts
NM_001282236.1 NM_004196.4 XM_005268157.2 XM_005268159.2 XM_005268160.4 XM_011537276.1 XM_017021729.1 XM_017021730.1 XM_017021731.1 XM_017021732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6075745 c.774T>C p.S258S Chr.14: 50334589-50334589 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754522505 Chr.14: 50334627-50334631 on Build GRCh38
rs74049603 Chr.14: 50334811-50334811 on Build GRCh38
rs750532769 Chr.14: 50334711-50334711 on Build GRCh38
rs374989440 Chr.14: 50334514-50334514 on Build GRCh38
rs1018703166 Chr.14: 50334939-50334939 on Build GRCh38
rs764291672 Chr.14: 50334607-50334607 on Build GRCh38
rs778876488 Chr.14: 50334698-50334698 on Build GRCh38
rs754943128 Chr.14: 50334899-50334899 on Build GRCh38
rs758522157 Chr.14: 50334725-50334725 on Build GRCh38
rs775996011 Chr.14: 50334545-50334545 on Build GRCh38
rs1014489072 Chr.14: 50334689-50334689 on Build GRCh38
rs11570857 Chr.14: 50334548-50334548 on Build GRCh38
rs879604336 Chr.14: 50334724-50334724 on Build GRCh38
rs896012207 Chr.14: 50334989-50334989 on Build GRCh38
rs556971870 Chr.14: 50334733-50334733 on Build GRCh38
rs11570856 Chr.14: 50334831-50334831 on Build GRCh38
rs762798952 Chr.14: 50334570-50334570 on Build GRCh38
rs929353764 Chr.14: 50334971-50334971 on Build GRCh38
rs746511334 Chr.14: 50334537-50334537 on Build GRCh38
rs560147185 Chr.14: 50334830-50334830 on Build GRCh38
rs753838614 Chr.14: 50334612-50334612 on Build GRCh38
rs746198224 Chr.14: 50334517-50334517 on Build GRCh38
rs533320571 Chr.14: 50334837-50334837 on Build GRCh38
rs1004772440 Chr.14: 50334939-50334939 on Build GRCh38
rs542227429 Chr.14: 50334755-50334755 on Build GRCh38
rs779631482 Chr.14: 50334536-50334536 on Build GRCh38
rs985396743 Chr.14: 50334812-50334812 on Build GRCh38
rs921375699 Chr.14: 50334657-50334657 on Build GRCh38
rs544043579 Chr.14: 50334955-50334955 on Build GRCh38
rs376796436 Chr.14: 50334520-50334520 on Build GRCh38
rs949755181 Chr.14: 50334796-50334796 on Build GRCh38
rs555824391 Chr.14: 50334787-50334787 on Build GRCh38
rs367644400 Chr.14: 50334530-50334530 on Build GRCh38
rs965407066 Chr.14: 50334600-50334600 on Build GRCh38
rs993687400 Chr.14: 50334584-50334584 on Build GRCh38
rs896006015 Chr.14: 50334791-50334791 on Build GRCh38
rs983213870 Chr.14: 50334585-50334585 on Build GRCh38
rs750975246 Chr.14: 50334555-50334555 on Build GRCh38
rs555628844 Chr.14: 50334630-50334630 on Build GRCh38
rs780916746 Chr.14: 50334641-50334641 on Build GRCh38
rs537253695 Chr.14: 50334672-50334672 on Build GRCh38
rs1038071558 Chr.14: 50334860-50334860 on Build GRCh38
rs202053157 Chr.14: 50334931-50334931 on Build GRCh38
rs911728883 Chr.14: 50334593-50334594 on Build GRCh38
rs886317910 Chr.14: 50334933-50334933 on Build GRCh38
rs767875741 Chr.14: 50334574-50334574 on Build GRCh38
rs757820882 Chr.14: 50334618-50334618 on Build GRCh38
rs753634767 Chr.14: 50334630-50334632 on Build GRCh38
rs758076506 Chr.14: 50334528-50334528 on Build GRCh38
rs769836681 Chr.14: 50334551-50334551 on Build GRCh38
rs1028407076 Chr.14: 50334990-50334990 on Build GRCh38
rs752194321 Chr.14: 50334776-50334776 on Build GRCh38
rs114181799 Chr.14: 50334977-50334977 on Build GRCh38
rs758701523 Chr.14: 50334656-50334656 on Build GRCh38
rs779029480 Chr.14: 50334970-50334970 on Build GRCh38
rs969735363 Chr.14: 50334714-50334714 on Build GRCh38
rs11445321 Chr.14: 50334920-50334921 on Build GRCh38
rs766058038 Chr.14: 50334524-50334530 on Build GRCh38
rs200110007 Chr.14: 50334930-50334931 on Build GRCh38
rs189283267 Chr.14: 50334752-50334752 on Build GRCh38
rs988681191 Chr.14: 50334777-50334777 on Build GRCh38
rs896083867 Chr.14: 50334688-50334688 on Build GRCh38
rs1027741364 Chr.14: 50334909-50334909 on Build GRCh38
rs760675659 Chr.14: 50334601-50334601 on Build GRCh38
rs899249859 Chr.14: 50334882-50334882 on Build GRCh38
rs765690338 Chr.14: 50334623-50334623 on Build GRCh38
rs11570858 Chr.14: 50334515-50334515 on Build GRCh38
rs373022131 Chr.14: 50334541-50334541 on Build GRCh38
rs779681516 Chr.14: 50334658-50334658 on Build GRCh38
rs773184833 Chr.14: 50334555-50334555 on Build GRCh38
rs932649161 Chr.14: 50334675-50334675 on Build GRCh38

View Full Product Details