Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50621451-50621960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL1
Entrez Gene ID
51062
Gene Name
atlastin GTPase 1
Gene Aliases
AD-FSP, FSP1, GBP3, HSN1D, SPG3, SPG3A, atlastin1
UniGene
Hs.584905
Gene Chromosome Location
Chr.14: 50533082-50633068 on Build GRCh38
Transcripts
NM_001127713.1 NM_015915.4 NM_181598.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6606801 c.1024C>T p.P342S Chr.14: 50621876-50621876 on Build GRCh38
4887753 c.991G>A p.A331T Chr.14: 50621843-50621843 on Build GRCh38
6408461 c.1006T>C p.Y336H Chr.14: 50621858-50621858 on Build GRCh38
5362361 c.991-11_991-10delTT p.? Chr.14: 50621832-50621833 on Build GRCh38
1369968 c.1047G>T p.Q349H Chr.14: 50621899-50621899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918986156 Chr.14: 50621672-50621672 on Build GRCh38
rs967763110 Chr.14: 50621495-50621495 on Build GRCh38
rs896712898 Chr.14: 50621862-50621862 on Build GRCh38
rs745982210 Chr.14: 50621934-50621934 on Build GRCh38
rs923234900 Chr.14: 50621593-50621593 on Build GRCh38
rs760671844 Chr.14: 50621825-50621826 on Build GRCh38
rs375882898 Chr.14: 50621911-50621911 on Build GRCh38
rs749993404 Chr.14: 50621849-50621849 on Build GRCh38
rs753092464 Chr.14: 50621799-50621799 on Build GRCh38
rs545276695 Chr.14: 50621937-50621937 on Build GRCh38
rs957515046 Chr.14: 50621493-50621493 on Build GRCh38
rs746881198 Chr.14: 50621917-50621917 on Build GRCh38
rs747931277 Chr.14: 50621928-50621928 on Build GRCh38
rs1035908263 Chr.14: 50621566-50621566 on Build GRCh38
rs542431760 Chr.14: 50621786-50621786 on Build GRCh38
rs943276816 Chr.14: 50621811-50621811 on Build GRCh38
rs537173230 Chr.14: 50621567-50621567 on Build GRCh38
rs766814368 Chr.14: 50621576-50621576 on Build GRCh38
rs979185754 Chr.14: 50621525-50621525 on Build GRCh38
rs371714758 Chr.14: 50621831-50621831 on Build GRCh38
rs192428744 Chr.14: 50621837-50621837 on Build GRCh38
rs754774903 Chr.14: 50621919-50621919 on Build GRCh38
rs150218601 Chr.14: 50621848-50621848 on Build GRCh38
rs761128367 Chr.14: 50621807-50621807 on Build GRCh38
rs763860692 Chr.14: 50621830-50621831 on Build GRCh38
rs889069270 Chr.14: 50621790-50621790 on Build GRCh38
rs573010386 Chr.14: 50621844-50621844 on Build GRCh38
rs931908235 Chr.14: 50621599-50621599 on Build GRCh38
rs1038456368 Chr.14: 50621842-50621842 on Build GRCh38
rs557079314 Chr.14: 50621582-50621582 on Build GRCh38
rs932974610 Chr.14: 50621724-50621724 on Build GRCh38
rs375741565 Chr.14: 50621812-50621812 on Build GRCh38
rs887614168 Chr.14: 50621818-50621818 on Build GRCh38
rs774414333 Chr.14: 50621933-50621933 on Build GRCh38
rs113299495 Chr.14: 50621758-50621758 on Build GRCh38
rs753551265 Chr.14: 50621836-50621836 on Build GRCh38
rs35103726 Chr.14: 50621724-50621725 on Build GRCh38
rs1035663395 Chr.14: 50621850-50621850 on Build GRCh38
rs557509259 Chr.14: 50621541-50621541 on Build GRCh38
rs764970555 Chr.14: 50621841-50621844 on Build GRCh38
rs753642042 Chr.14: 50621832-50621832 on Build GRCh38
rs972212166 Chr.14: 50621655-50621655 on Build GRCh38
rs573898926 Chr.14: 50621689-50621689 on Build GRCh38
rs780908334 Chr.14: 50621922-50621922 on Build GRCh38
rs577709661 Chr.14: 50621558-50621558 on Build GRCh38
rs369950441 Chr.14: 50621931-50621931 on Build GRCh38
rs761300786 Chr.14: 50621831-50621832 on Build GRCh38
rs1050239377 Chr.14: 50621792-50621792 on Build GRCh38
rs780064369 Chr.14: 50621912-50621912 on Build GRCh38
rs750873169 Chr.14: 50621515-50621515 on Build GRCh38
rs941814426 Chr.14: 50621624-50621624 on Build GRCh38
rs909370240 Chr.14: 50621604-50621604 on Build GRCh38
rs757828342 Chr.14: 50621853-50621853 on Build GRCh38

View Full Product Details