Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50881504-50881937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD12B
Entrez Gene ID
145447
Gene Name
abhydrolase domain containing 12B
Gene Aliases
BEM46L3, C14orf29, c14_5314
UniGene
Hs.271896
Gene Chromosome Location
Chr.14: 50872090-50904970 on Build GRCh38
Transcripts
NM_001206673.1 NM_181533.3 NM_181814.1 XM_006720052.1 XM_011536472.1 XM_011536474.2 XM_017021031.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5778139 c.475A>C p.K159Q Chr.14: 50881615-50881615 on Build GRCh38
5778140 c.244A>C p.K82Q Chr.14: 50881615-50881615 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1018745545 Chr.14: 50881773-50881773 on Build GRCh38
rs973371447 Chr.14: 50881758-50881758 on Build GRCh38
rs571015478 Chr.14: 50881723-50881723 on Build GRCh38
rs932939194 Chr.14: 50881841-50881841 on Build GRCh38
rs1017286340 Chr.14: 50881733-50881733 on Build GRCh38
rs78896478 Chr.14: 50881777-50881777 on Build GRCh38
rs367703780 Chr.14: 50881864-50881864 on Build GRCh38
rs767696604 Chr.14: 50881548-50881548 on Build GRCh38
rs760747465 Chr.14: 50881658-50881658 on Build GRCh38
rs982896985 Chr.14: 50881541-50881541 on Build GRCh38
rs775596474 Chr.14: 50881549-50881549 on Build GRCh38
rs200135016 Chr.14: 50881594-50881594 on Build GRCh38
rs1009031320 Chr.14: 50881726-50881726 on Build GRCh38
rs781294981 Chr.14: 50881626-50881626 on Build GRCh38
rs149280376 Chr.14: 50881667-50881667 on Build GRCh38
rs949407000 Chr.14: 50881620-50881620 on Build GRCh38
rs750799428 Chr.14: 50881675-50881675 on Build GRCh38
rs894059959 Chr.14: 50881854-50881854 on Build GRCh38
rs929109518 Chr.14: 50881540-50881540 on Build GRCh38
rs1053038896 Chr.14: 50881860-50881860 on Build GRCh38
rs964709244 Chr.14: 50881756-50881756 on Build GRCh38
rs879238723 Chr.14: 50881562-50881562 on Build GRCh38
rs750117835 Chr.14: 50881604-50881604 on Build GRCh38
rs908622770 Chr.14: 50881605-50881605 on Build GRCh38
rs764597534 Chr.14: 50881870-50881870 on Build GRCh38
rs548833648 Chr.14: 50881696-50881696 on Build GRCh38
rs555172041 Chr.14: 50881844-50881844 on Build GRCh38
rs756756786 Chr.14: 50881564-50881564 on Build GRCh38
rs141717882 Chr.14: 50881781-50881781 on Build GRCh38
rs760731459 Chr.14: 50881584-50881584 on Build GRCh38
rs797009959 Chr.14: 50881564-50881565 on Build GRCh38
rs777216766 Chr.14: 50881665-50881665 on Build GRCh38
rs201392341 Chr.14: 50881581-50881581 on Build GRCh38
rs758981266 Chr.14: 50881581-50881584 on Build GRCh38
rs201966468 Chr.14: 50881661-50881661 on Build GRCh38
rs375805491 Chr.14: 50881580-50881581 on Build GRCh38
rs17794038 Chr.14: 50881703-50881703 on Build GRCh38
rs983224252 Chr.14: 50881730-50881730 on Build GRCh38
rs747404778 Chr.14: 50881648-50881650 on Build GRCh38
rs371987623 Chr.14: 50881653-50881653 on Build GRCh38
rs750950771 Chr.14: 50881564-50881568 on Build GRCh38
rs1027563487 Chr.14: 50881772-50881772 on Build GRCh38
rs777468877 Chr.14: 50881642-50881642 on Build GRCh38
rs369385880 Chr.14: 50881563-50881564 on Build GRCh38
rs34965747 Chr.14: 50881564-50881565 on Build GRCh38
rs138523238 Chr.14: 50881627-50881627 on Build GRCh38
rs982581109 Chr.14: 50881797-50881797 on Build GRCh38
rs775686452 Chr.14: 50881654-50881654 on Build GRCh38
rs1014074723 Chr.14: 50881693-50881693 on Build GRCh38
rs569749239 Chr.14: 50881816-50881816 on Build GRCh38
rs768955764 Chr.14: 50881669-50881669 on Build GRCh38
rs780528780 Chr.14: 50881594-50881595 on Build GRCh38
rs764650254 Chr.14: 50881587-50881587 on Build GRCh38
rs746573105 Chr.14: 50881618-50881618 on Build GRCh38
rs917053364 Chr.14: 50881780-50881780 on Build GRCh38
rs754473643 Chr.14: 50881621-50881621 on Build GRCh38
rs894006340 Chr.14: 50881700-50881700 on Build GRCh38
rs765740125 Chr.14: 50881673-50881673 on Build GRCh38
rs529975272 Chr.14: 50881827-50881827 on Build GRCh38
rs548100930 Chr.14: 50881720-50881720 on Build GRCh38
rs890596785 Chr.14: 50881866-50881866 on Build GRCh38
rs937998819 Chr.14: 50881829-50881829 on Build GRCh38
rs557642834 Chr.14: 50881890-50881890 on Build GRCh38
rs765470166 Chr.14: 50881597-50881597 on Build GRCh38
rs754230019 Chr.14: 50881588-50881588 on Build GRCh38
rs944149189 Chr.14: 50881532-50881532 on Build GRCh38
rs561217093 Chr.14: 50881817-50881817 on Build GRCh38
rs749088583 Chr.14: 50881645-50881645 on Build GRCh38
rs12588230 Chr.14: 50881856-50881856 on Build GRCh38
rs983284272 Chr.14: 50881778-50881778 on Build GRCh38
rs376109019 Chr.14: 50881685-50881685 on Build GRCh38
rs779827106 Chr.14: 50881610-50881610 on Build GRCh38
rs890208830 Chr.14: 50881713-50881713 on Build GRCh38
rs1027333935 Chr.14: 50881705-50881705 on Build GRCh38
rs920446046 Chr.14: 50881526-50881526 on Build GRCh38
rs144483430 Chr.14: 50881702-50881702 on Build GRCh38
rs114700216 Chr.14: 50881563-50881563 on Build GRCh38
rs895930618 Chr.14: 50881878-50881878 on Build GRCh38
rs979396094 Chr.14: 50881843-50881843 on Build GRCh38

View Full Product Details