Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50886467-50886935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD12B
Entrez Gene ID
145447
Gene Name
abhydrolase domain containing 12B
Gene Aliases
BEM46L3, C14orf29, c14_5314
UniGene
Hs.271896
Gene Chromosome Location
Chr.14: 50872090-50904970 on Build GRCh38
Transcripts
NM_001206673.1 NM_181533.3 NM_181814.1 XM_006720052.1 XM_011536472.1 XM_011536474.2 XM_017021031.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5991975 c.469+8delA p.? Chr.14: 50886692-50886692 on Build GRCh38
1707336 c.469G>A p.G157R Chr.14: 50886684-50886684 on Build GRCh38
5991974 c.700+8delA p.? Chr.14: 50886692-50886692 on Build GRCh38
1707335 c.700G>A p.G234R Chr.14: 50886684-50886684 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761299453 Chr.14: 50886678-50886678 on Build GRCh38
rs375283388 Chr.14: 50886691-50886691 on Build GRCh38
rs545641122 Chr.14: 50886523-50886523 on Build GRCh38
rs371044930 Chr.14: 50886611-50886611 on Build GRCh38
rs898490414 Chr.14: 50886908-50886908 on Build GRCh38
rs948274690 Chr.14: 50886693-50886693 on Build GRCh38
rs376790794 Chr.14: 50886705-50886705 on Build GRCh38
rs765120667 Chr.14: 50886531-50886531 on Build GRCh38
rs377360310 Chr.14: 50886869-50886870 on Build GRCh38
rs562818072 Chr.14: 50886566-50886566 on Build GRCh38
rs891350038 Chr.14: 50886887-50886887 on Build GRCh38
rs750292661 Chr.14: 50886702-50886702 on Build GRCh38
rs898705428 Chr.14: 50886517-50886517 on Build GRCh38
rs544227405 Chr.14: 50886763-50886763 on Build GRCh38
rs547068199 Chr.14: 50886847-50886847 on Build GRCh38
rs936376930 Chr.14: 50886706-50886706 on Build GRCh38
rs575904586 Chr.14: 50886762-50886762 on Build GRCh38
rs544097165 Chr.14: 50886906-50886906 on Build GRCh38
rs530173354 Chr.14: 50886837-50886837 on Build GRCh38
rs763618687 Chr.14: 50886671-50886671 on Build GRCh38
rs756408419 Chr.14: 50886727-50886727 on Build GRCh38
rs879173994 Chr.14: 50886599-50886599 on Build GRCh38
rs980367794 Chr.14: 50886698-50886698 on Build GRCh38
rs1057015963 Chr.14: 50886551-50886551 on Build GRCh38
rs540007532 Chr.14: 50886808-50886808 on Build GRCh38
rs113033362 Chr.14: 50886518-50886518 on Build GRCh38
rs539000555 Chr.14: 50886863-50886864 on Build GRCh38
rs776162686 Chr.14: 50886674-50886674 on Build GRCh38
rs760440999 Chr.14: 50886678-50886679 on Build GRCh38
rs765225358 Chr.14: 50886681-50886681 on Build GRCh38
rs951231447 Chr.14: 50886586-50886586 on Build GRCh38
rs553732583 Chr.14: 50886496-50886496 on Build GRCh38
rs753700919 Chr.14: 50886509-50886509 on Build GRCh38
rs372830872 Chr.14: 50886734-50886734 on Build GRCh38
rs942932507 Chr.14: 50886899-50886899 on Build GRCh38
rs760333055 Chr.14: 50886666-50886666 on Build GRCh38
rs995596302 Chr.14: 50886730-50886730 on Build GRCh38
rs987896395 Chr.14: 50886773-50886773 on Build GRCh38
rs1024866983 Chr.14: 50886628-50886628 on Build GRCh38
rs775215776 Chr.14: 50886665-50886665 on Build GRCh38
rs182765699 Chr.14: 50886881-50886881 on Build GRCh38
rs7158183 Chr.14: 50886775-50886775 on Build GRCh38
rs578256505 Chr.14: 50886637-50886637 on Build GRCh38
rs751282403 Chr.14: 50886720-50886720 on Build GRCh38
rs992423380 Chr.14: 50886592-50886592 on Build GRCh38
rs766160091 Chr.14: 50886709-50886709 on Build GRCh38
rs969288124 Chr.14: 50886638-50886638 on Build GRCh38
rs1026200599 Chr.14: 50886521-50886521 on Build GRCh38

View Full Product Details