Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51651686-51652201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD6
Entrez Gene ID
122786
Gene Name
FERM domain containing 6
Gene Aliases
C14orf31, EX1, Willin, c14_5320
UniGene
Hs.434914
Gene Chromosome Location
Chr.14: 51489100-51730727 on Build GRCh38
Transcripts
NM_001042481.2 NM_001267046.1 NM_152330.3 XM_006720029.1 XM_006720030.1 XM_011536423.1 XM_011536424.1 XM_017020967.1 XM_017020968.1 NR_037676.1

Gene Symbol
FRMD6-AS1
Entrez Gene ID
145438
Gene Name
FRMD6 antisense RNA 1
Gene Aliases
C14orf82
UniGene
Hs.645410
Gene Chromosome Location
Chr.14: 51649516-51651744 on Build GRCh38
Transcripts
NM_001042481.2 NM_001267046.1 NM_152330.3 XM_006720029.1 XM_006720030.1 XM_011536423.1 XM_011536424.1 XM_017020967.1 XM_017020968.1 NR_037676.1

SNP Detail

SNP ID Chromosome Location
rs886794462 Chr.14: 51651755-51651755 on Build GRCh38
rs998633617 Chr.14: 51652036-51652036 on Build GRCh38
rs965774676 Chr.14: 51651893-51651893 on Build GRCh38
rs927724874 Chr.14: 51652069-51652069 on Build GRCh38
rs1028127466 Chr.14: 51652003-51652003 on Build GRCh38
rs529251249 Chr.14: 51652176-51652176 on Build GRCh38
rs545179871 Chr.14: 51652073-51652073 on Build GRCh38
rs955224651 Chr.14: 51652008-51652008 on Build GRCh38
rs997005943 Chr.14: 51652002-51652002 on Build GRCh38
rs904720264 Chr.14: 51651985-51651985 on Build GRCh38
rs1016715551 Chr.14: 51651767-51651767 on Build GRCh38
rs1031733732 Chr.14: 51651883-51651883 on Build GRCh38
rs537090661 Chr.14: 51651726-51651726 on Build GRCh38
rs1051200830 Chr.14: 51651796-51651810 on Build GRCh38
rs984040434 Chr.14: 51652050-51652050 on Build GRCh38
rs971358956 Chr.14: 51651855-51651855 on Build GRCh38
rs914702252 Chr.14: 51651901-51651901 on Build GRCh38
rs778228786 Chr.14: 51652153-51652153 on Build GRCh38
rs886763422 Chr.14: 51651959-51651959 on Build GRCh38
rs955255646 Chr.14: 51651831-51651832 on Build GRCh38
rs866839865 Chr.14: 51651827-51651827 on Build GRCh38
rs867458388 Chr.14: 51651733-51651733 on Build GRCh38
rs908162116 Chr.14: 51652138-51652138 on Build GRCh38
rs977480323 Chr.14: 51651750-51651750 on Build GRCh38
rs1032203741 Chr.14: 51652016-51652016 on Build GRCh38
rs79346288 Chr.14: 51652129-51652129 on Build GRCh38
rs953940788 Chr.14: 51651906-51651906 on Build GRCh38
rs972155857 Chr.14: 51651862-51651862 on Build GRCh38
rs553271582 Chr.14: 51651879-51651879 on Build GRCh38
rs1012813915 Chr.14: 51651832-51651832 on Build GRCh38
rs939042840 Chr.14: 51651935-51651935 on Build GRCh38
rs1003186398 Chr.14: 51651719-51651719 on Build GRCh38
rs998951543 Chr.14: 51652005-51652005 on Build GRCh38
rs546629700 Chr.14: 51651852-51651852 on Build GRCh38
rs970140155 Chr.14: 51652068-51652068 on Build GRCh38
rs893276004 Chr.14: 51651716-51651716 on Build GRCh38
rs1013112661 Chr.14: 51651951-51651951 on Build GRCh38
rs1053288725 Chr.14: 51652139-51652139 on Build GRCh38
rs1038005184 Chr.14: 51652152-51652152 on Build GRCh38
rs976768848 Chr.14: 51651894-51651894 on Build GRCh38
rs898203063 Chr.14: 51652000-51652000 on Build GRCh38
rs578030068 Chr.14: 51652094-51652094 on Build GRCh38
rs139014790 Chr.14: 51652082-51652082 on Build GRCh38
rs907414259 Chr.14: 51651728-51651728 on Build GRCh38
rs965474280 Chr.14: 51652017-51652017 on Build GRCh38
rs1003949310 Chr.14: 51651766-51651766 on Build GRCh38
rs1054778585 Chr.14: 51651938-51651938 on Build GRCh38
rs1013059633 Chr.14: 51651735-51651735 on Build GRCh38
rs1021744865 Chr.14: 51651739-51651739 on Build GRCh38
rs1024750782 Chr.14: 51651836-51651836 on Build GRCh38
rs111922964 Chr.14: 51651997-51651997 on Build GRCh38
rs563515552 Chr.14: 51652141-51652141 on Build GRCh38
rs998289439 Chr.14: 51652146-51652146 on Build GRCh38
rs924591215 Chr.14: 51651872-51651872 on Build GRCh38
rs894711765 Chr.14: 51651730-51651730 on Build GRCh38
rs767968458 Chr.14: 51652011-51652011 on Build GRCh38
rs894756667 Chr.14: 51651946-51651946 on Build GRCh38
rs958886732 Chr.14: 51651806-51651806 on Build GRCh38
rs939795055 Chr.14: 51651924-51651924 on Build GRCh38
rs999455068 Chr.14: 51652133-51652133 on Build GRCh38
rs985371471 Chr.14: 51652118-51652118 on Build GRCh38
rs556679887 Chr.14: 51651783-51651783 on Build GRCh38
rs927706273 Chr.14: 51651934-51651934 on Build GRCh38
rs909726283 Chr.14: 51651922-51651922 on Build GRCh38
rs961212375 Chr.14: 51652059-51652059 on Build GRCh38
rs921203931 Chr.14: 51651897-51651897 on Build GRCh38
rs954044838 Chr.14: 51652170-51652170 on Build GRCh38
rs906192384 Chr.14: 51651707-51651707 on Build GRCh38
rs12879510 Chr.14: 51651795-51651795 on Build GRCh38
rs909775695 Chr.14: 51652054-51652054 on Build GRCh38
rs1037205398 Chr.14: 51651927-51651927 on Build GRCh38
rs916254400 Chr.14: 51652088-51652088 on Build GRCh38
rs549173350 Chr.14: 51651917-51651917 on Build GRCh38
rs946363907 Chr.14: 51652100-51652100 on Build GRCh38
rs759467705 Chr.14: 51652066-51652066 on Build GRCh38
rs956037682 Chr.14: 51652048-51652048 on Build GRCh38
rs968806201 Chr.14: 51651749-51651749 on Build GRCh38
rs1043253468 Chr.14: 51651962-51651962 on Build GRCh38
rs986691713 Chr.14: 51651839-51651839 on Build GRCh38
rs1043304964 Chr.14: 51652110-51652110 on Build GRCh38
rs953994234 Chr.14: 51652046-51652046 on Build GRCh38
rs1028318703 Chr.14: 51652042-51652042 on Build GRCh38
rs1038522382 Chr.14: 51651987-51651987 on Build GRCh38
rs573907865 Chr.14: 51651794-51651794 on Build GRCh38

View Full Product Details