Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51967656-51968150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG2
Entrez Gene ID
54331
Gene Name
G protein subunit gamma 2
Gene Aliases
-
UniGene
Hs.187772
Gene Chromosome Location
Chr.14: 51847131-51969800 on Build GRCh38
Transcripts
NM_001243773.1 NM_001243774.1 NM_053064.4 XM_006720173.2 XM_011536846.2 XM_017021376.1 XM_017021377.1

SNP Detail

SNP ID Chromosome Location
rs750564884 Chr.14: 51967860-51967860 on Build GRCh38
rs3832939 Chr.14: 51967817-51967817 on Build GRCh38
rs113599001 Chr.14: 51967997-51967997 on Build GRCh38
rs35178030 Chr.14: 51968053-51968053 on Build GRCh38
rs549701999 Chr.14: 51967967-51967967 on Build GRCh38
rs1056211040 Chr.14: 51967831-51967831 on Build GRCh38
rs376713821 Chr.14: 51968014-51968014 on Build GRCh38
rs185099343 Chr.14: 51967943-51967943 on Build GRCh38
rs980413495 Chr.14: 51968017-51968017 on Build GRCh38
rs1022662946 Chr.14: 51967884-51967884 on Build GRCh38
rs527849179 Chr.14: 51967952-51967952 on Build GRCh38
rs1044689830 Chr.14: 51967739-51967739 on Build GRCh38
rs925212223 Chr.14: 51967936-51967936 on Build GRCh38
rs966330646 Chr.14: 51968121-51968121 on Build GRCh38
rs913566342 Chr.14: 51967896-51967896 on Build GRCh38
rs901388605 Chr.14: 51968070-51968073 on Build GRCh38
rs1055340191 Chr.14: 51967957-51967957 on Build GRCh38
rs3742537 Chr.14: 51967920-51967920 on Build GRCh38
rs8010602 Chr.14: 51967953-51967953 on Build GRCh38
rs748450528 Chr.14: 51968092-51968092 on Build GRCh38
rs1056642877 Chr.14: 51968101-51968101 on Build GRCh38
rs10144829 Chr.14: 51968107-51968107 on Build GRCh38
rs765507660 Chr.14: 51967748-51967748 on Build GRCh38
rs192596578 Chr.14: 51968064-51968064 on Build GRCh38
rs975806537 Chr.14: 51967892-51967892 on Build GRCh38
rs1027684264 Chr.14: 51967811-51967811 on Build GRCh38
rs979358468 Chr.14: 51967935-51967935 on Build GRCh38
rs897163672 Chr.14: 51967767-51967767 on Build GRCh38
rs913640533 Chr.14: 51968105-51968105 on Build GRCh38
rs144364053 Chr.14: 51967966-51967966 on Build GRCh38
rs762141000 Chr.14: 51967709-51967709 on Build GRCh38
rs188764948 Chr.14: 51967963-51967963 on Build GRCh38
rs1037158450 Chr.14: 51967976-51967976 on Build GRCh38
rs554960688 Chr.14: 51967996-51967996 on Build GRCh38
rs75661067 Chr.14: 51967711-51967711 on Build GRCh38
rs531772844 Chr.14: 51967834-51967834 on Build GRCh38
rs752273411 Chr.14: 51967898-51967898 on Build GRCh38
rs904955857 Chr.14: 51967794-51967794 on Build GRCh38
rs562861677 Chr.14: 51967775-51967775 on Build GRCh38
rs558322327 Chr.14: 51968065-51968065 on Build GRCh38
rs397853142 Chr.14: 51967823-51967823 on Build GRCh38
rs577407874 Chr.14: 51968108-51968108 on Build GRCh38
rs535057269 Chr.14: 51967984-51967984 on Build GRCh38
rs931331382 Chr.14: 51967990-51967990 on Build GRCh38
rs2000006 Chr.14: 51967869-51967869 on Build GRCh38
rs995756144 Chr.14: 51967777-51967778 on Build GRCh38
rs939065869 Chr.14: 51967824-51967824 on Build GRCh38
rs903744134 Chr.14: 51967903-51967903 on Build GRCh38
rs577414781 Chr.14: 51967690-51967690 on Build GRCh38
rs1037001279 Chr.14: 51967743-51967743 on Build GRCh38
rs369231553 Chr.14: 51967975-51967975 on Build GRCh38

View Full Product Details