Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51991255-51991738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf166
Entrez Gene ID
51637
Gene Name
chromosome 14 open reading frame 166
Gene Aliases
CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE1
UniGene
Hs.534457
Gene Chromosome Location
Chr.14: 51989510-52004702 on Build GRCh38
Transcripts
NM_016039.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
956165 c.162C>T p.S54S Chr.14: 51991417-51991417 on Build GRCh38
6298978 c.75T>C p.F25F Chr.14: 51991330-51991330 on Build GRCh38
4830107 c.120C>G p.Y40* Chr.14: 51991375-51991375 on Build GRCh38
6140421 c.101A>G p.D34G Chr.14: 51991356-51991356 on Build GRCh38
3886104 c.84C>T p.F28F Chr.14: 51991339-51991339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988720492 Chr.14: 51991513-51991513 on Build GRCh38
rs757410193 Chr.14: 51991493-51991493 on Build GRCh38
rs779550281 Chr.14: 51991391-51991391 on Build GRCh38
rs111849085 Chr.14: 51991523-51991523 on Build GRCh38
rs765090529 Chr.14: 51991340-51991340 on Build GRCh38
rs746660717 Chr.14: 51991417-51991417 on Build GRCh38
rs192922427 Chr.14: 51991653-51991653 on Build GRCh38
rs879264907 Chr.14: 51991597-51991597 on Build GRCh38
rs762719676 Chr.14: 51991342-51991342 on Build GRCh38
rs1014103647 Chr.14: 51991404-51991404 on Build GRCh38
rs776006355 Chr.14: 51991434-51991434 on Build GRCh38
rs775147416 Chr.14: 51991472-51991473 on Build GRCh38
rs967124674 Chr.14: 51991698-51991698 on Build GRCh38
rs774861619 Chr.14: 51991286-51991286 on Build GRCh38
rs976256388 Chr.14: 51991668-51991668 on Build GRCh38
rs73301033 Chr.14: 51991713-51991713 on Build GRCh38
rs766112608 Chr.14: 51991453-51991453 on Build GRCh38
rs538236457 Chr.14: 51991521-51991521 on Build GRCh38
rs979945917 Chr.14: 51991469-51991469 on Build GRCh38
rs746178007 Chr.14: 51991296-51991296 on Build GRCh38
rs769777264 Chr.14: 51991443-51991443 on Build GRCh38
rs372075744 Chr.14: 51991371-51991371 on Build GRCh38
rs771794428 Chr.14: 51991388-51991389 on Build GRCh38
rs753924231 Chr.14: 51991483-51991483 on Build GRCh38
rs368830857 Chr.14: 51991345-51991345 on Build GRCh38
rs1027681799 Chr.14: 51991514-51991514 on Build GRCh38
rs745861121 Chr.14: 51991303-51991303 on Build GRCh38
rs767770402 Chr.14: 51991343-51991343 on Build GRCh38
rs113738821 Chr.14: 51991676-51991676 on Build GRCh38
rs370113138 Chr.14: 51991479-51991479 on Build GRCh38
rs771526327 Chr.14: 51991284-51991284 on Build GRCh38
rs574970809 Chr.14: 51991563-51991563 on Build GRCh38
rs113114137 Chr.14: 51991440-51991440 on Build GRCh38
rs555042731 Chr.14: 51991528-51991528 on Build GRCh38
rs188526168 Chr.14: 51991396-51991396 on Build GRCh38
rs1007788854 Chr.14: 51991567-51991567 on Build GRCh38
rs369538342 Chr.14: 51991305-51991305 on Build GRCh38
rs760331257 Chr.14: 51991480-51991483 on Build GRCh38
rs757854431 Chr.14: 51991379-51991379 on Build GRCh38
rs371351236 Chr.14: 51991311-51991311 on Build GRCh38
rs138646269 Chr.14: 51991335-51991335 on Build GRCh38
rs183965924 Chr.14: 51991317-51991317 on Build GRCh38
rs772532579 Chr.14: 51991408-51991408 on Build GRCh38
rs938599204 Chr.14: 51991504-51991504 on Build GRCh38
rs894268446 Chr.14: 51991393-51991393 on Build GRCh38
rs779804988 Chr.14: 51991415-51991415 on Build GRCh38
rs577567827 Chr.14: 51991577-51991577 on Build GRCh38
rs771908599 Chr.14: 51991300-51991300 on Build GRCh38
rs879212579 Chr.14: 51991613-51991613 on Build GRCh38
rs540400088 Chr.14: 51991603-51991603 on Build GRCh38
rs200174268 Chr.14: 51991313-51991313 on Build GRCh38
rs764322132 Chr.14: 51991477-51991477 on Build GRCh38
rs773106709 Chr.14: 51991447-51991447 on Build GRCh38
rs760813595 Chr.14: 51991464-51991464 on Build GRCh38
rs994397991 Chr.14: 51991339-51991339 on Build GRCh38
rs889911712 Chr.14: 51991623-51991623 on Build GRCh38
rs913237539 Chr.14: 51991556-51991556 on Build GRCh38
rs952156954 Chr.14: 51991534-51991539 on Build GRCh38
rs375132194 Chr.14: 51991462-51991462 on Build GRCh38
rs750227458 Chr.14: 51991341-51991341 on Build GRCh38
rs768313510 Chr.14: 51991418-51991418 on Build GRCh38
rs753889283 Chr.14: 51991377-51991377 on Build GRCh38
rs200015040 Chr.14: 51991323-51991323 on Build GRCh38
rs756314038 Chr.14: 51991358-51991358 on Build GRCh38
rs140351303 Chr.14: 51991664-51991664 on Build GRCh38

View Full Product Details