Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52001715-52002211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf166
Entrez Gene ID
51637
Gene Name
chromosome 14 open reading frame 166
Gene Aliases
CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE1
UniGene
Hs.534457
Gene Chromosome Location
Chr.14: 51989510-52004702 on Build GRCh38
Transcripts
NM_016039.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4051239 c.521A>G p.Q174R Chr.14: 52001856-52001856 on Build GRCh38
956170 c.487C>T p.R163C Chr.14: 52001822-52001822 on Build GRCh38
6267191 c.505G>T p.V169F Chr.14: 52001840-52001840 on Build GRCh38
2199321 c.469C>T p.R157W Chr.14: 52001804-52001804 on Build GRCh38
3727952 c.470G>A p.R157Q Chr.14: 52001805-52001805 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1055480308 Chr.14: 52001811-52001811 on Build GRCh38
rs962619459 Chr.14: 52002121-52002121 on Build GRCh38
rs1019035681 Chr.14: 52002101-52002101 on Build GRCh38
rs1011311436 Chr.14: 52002021-52002021 on Build GRCh38
rs374136328 Chr.14: 52001781-52001781 on Build GRCh38
rs763827648 Chr.14: 52001909-52001909 on Build GRCh38
rs972316504 Chr.14: 52002125-52002125 on Build GRCh38
rs1029067568 Chr.14: 52002039-52002039 on Build GRCh38
rs747031503 Chr.14: 52001845-52001845 on Build GRCh38
rs550346976 Chr.14: 52001747-52001747 on Build GRCh38
rs367945451 Chr.14: 52001794-52001794 on Build GRCh38
rs889195812 Chr.14: 52002056-52002056 on Build GRCh38
rs111910035 Chr.14: 52002009-52002009 on Build GRCh38
rs201192759 Chr.14: 52001855-52001855 on Build GRCh38
rs749260099 Chr.14: 52001844-52001844 on Build GRCh38
rs550530083 Chr.14: 52002141-52002141 on Build GRCh38
rs372766786 Chr.14: 52001870-52001870 on Build GRCh38
rs371515575 Chr.14: 52001829-52001829 on Build GRCh38
rs779245204 Chr.14: 52001833-52001833 on Build GRCh38
rs753562341 Chr.14: 52001917-52001917 on Build GRCh38
rs937698006 Chr.14: 52002086-52002086 on Build GRCh38
rs866657230 Chr.14: 52001836-52001836 on Build GRCh38
rs1741124 Chr.14: 52002044-52002044 on Build GRCh38
rs559806869 Chr.14: 52002052-52002052 on Build GRCh38
rs1151573 Chr.14: 52001923-52001923 on Build GRCh38
rs758580985 Chr.14: 52001843-52001843 on Build GRCh38
rs983787355 Chr.14: 52002162-52002162 on Build GRCh38
rs752608305 Chr.14: 52001823-52001823 on Build GRCh38
rs766007004 Chr.14: 52001804-52001804 on Build GRCh38
rs777080619 Chr.14: 52001822-52001822 on Build GRCh38
rs912847312 Chr.14: 52001968-52001968 on Build GRCh38
rs577955875 Chr.14: 52001929-52001929 on Build GRCh38
rs762723183 Chr.14: 52001871-52001871 on Build GRCh38
rs1013463096 Chr.14: 52002140-52002140 on Build GRCh38
rs139420074 Chr.14: 52001857-52001857 on Build GRCh38
rs565169840 Chr.14: 52002082-52002082 on Build GRCh38
rs567125956 Chr.14: 52002185-52002185 on Build GRCh38
rs202053353 Chr.14: 52001842-52001842 on Build GRCh38
rs187974089 Chr.14: 52001942-52001942 on Build GRCh38
rs924960073 Chr.14: 52002085-52002085 on Build GRCh38
rs776999049 Chr.14: 52001856-52001856 on Build GRCh38
rs1043880137 Chr.14: 52002065-52002065 on Build GRCh38
rs897010503 Chr.14: 52002180-52002180 on Build GRCh38
rs1036784762 Chr.14: 52002150-52002150 on Build GRCh38
rs563559785 Chr.14: 52001967-52001967 on Build GRCh38
rs766073596 Chr.14: 52001886-52001886 on Build GRCh38
rs745629594 Chr.14: 52002007-52002007 on Build GRCh38
rs762598622 Chr.14: 52001801-52001801 on Build GRCh38
rs567340442 Chr.14: 52001812-52001812 on Build GRCh38
rs767532071 Chr.14: 52001897-52001897 on Build GRCh38
rs528249148 Chr.14: 52002064-52002064 on Build GRCh38
rs781244788 Chr.14: 52001779-52001780 on Build GRCh38
rs965530078 Chr.14: 52002000-52002000 on Build GRCh38
rs751049926 Chr.14: 52001805-52001805 on Build GRCh38
rs184971107 Chr.14: 52002139-52002139 on Build GRCh38
rs755423596 Chr.14: 52001755-52001756 on Build GRCh38
rs1009021141 Chr.14: 52002096-52002096 on Build GRCh38
rs765003822 Chr.14: 52001788-52001788 on Build GRCh38
rs756031775 Chr.14: 52001901-52001901 on Build GRCh38
rs770028493 Chr.14: 52001864-52001864 on Build GRCh38
rs947909241 Chr.14: 52001922-52001922 on Build GRCh38
rs573999255 Chr.14: 52001971-52001971 on Build GRCh38
rs368844832 Chr.14: 52001888-52001888 on Build GRCh38
rs1028311106 Chr.14: 52002130-52002130 on Build GRCh38
rs775580692 Chr.14: 52001745-52001745 on Build GRCh38
rs906452324 Chr.14: 52001934-52001934 on Build GRCh38
rs754054934 Chr.14: 52001961-52001961 on Build GRCh38
rs193045053 Chr.14: 52002072-52002072 on Build GRCh38
rs542676370 Chr.14: 52001778-52001778 on Build GRCh38
rs377176106 Chr.14: 52001900-52001900 on Build GRCh38
rs555079066 Chr.14: 52001890-52001890 on Build GRCh38
rs534888333 Chr.14: 52001892-52001892 on Build GRCh38

View Full Product Details