Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52858377-52858909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
UniGene
Hs.509343
Gene Chromosome Location
Chr.14: 52857271-52951192 on Build GRCh38
Transcripts
NM_001134999.1 NM_006832.2 XM_005267285.2 XM_006720008.2 XM_006720009.2 XM_006720010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4452614 c.1983A>G p.K661K Chr.14: 52858437-52858437 on Build GRCh38
5504046 c.1902G>A p.L634L Chr.14: 52858518-52858518 on Build GRCh38
123154 c.1889A>T p.D630V Chr.14: 52858531-52858531 on Build GRCh38
433137 c.1879G>C p.E627Q Chr.14: 52858541-52858541 on Build GRCh38
6140896 c.2037G>T p.W679C Chr.14: 52858383-52858383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889390219 Chr.14: 52858738-52858738 on Build GRCh38
rs1006699168 Chr.14: 52858652-52858652 on Build GRCh38
rs927834367 Chr.14: 52858846-52858846 on Build GRCh38
rs542628897 Chr.14: 52858825-52858828 on Build GRCh38
rs753838462 Chr.14: 52858522-52858522 on Build GRCh38
rs962468300 Chr.14: 52858501-52858501 on Build GRCh38
rs774241384 Chr.14: 52858548-52858548 on Build GRCh38
rs1050582913 Chr.14: 52858878-52858878 on Build GRCh38
rs377458375 Chr.14: 52858708-52858709 on Build GRCh38
rs747415162 Chr.14: 52858439-52858439 on Build GRCh38
rs376050043 Chr.14: 52858413-52858413 on Build GRCh38
rs188956047 Chr.14: 52858494-52858494 on Build GRCh38
rs569399329 Chr.14: 52858594-52858594 on Build GRCh38
rs889652107 Chr.14: 52858639-52858639 on Build GRCh38
rs993865991 Chr.14: 52858778-52858778 on Build GRCh38
rs763018889 Chr.14: 52858563-52858563 on Build GRCh38
rs897846115 Chr.14: 52858756-52858756 on Build GRCh38
rs952101283 Chr.14: 52858795-52858795 on Build GRCh38
rs145708259 Chr.14: 52858801-52858801 on Build GRCh38
rs1036329984 Chr.14: 52858567-52858567 on Build GRCh38
rs988207012 Chr.14: 52858857-52858857 on Build GRCh38
rs762102846 Chr.14: 52858588-52858588 on Build GRCh38
rs776834850 Chr.14: 52858489-52858489 on Build GRCh38
rs184193185 Chr.14: 52858828-52858828 on Build GRCh38
rs1009091225 Chr.14: 52858741-52858741 on Build GRCh38
rs751371377 Chr.14: 52858692-52858692 on Build GRCh38
rs780675333 Chr.14: 52858719-52858719 on Build GRCh38
rs545019827 Chr.14: 52858791-52858791 on Build GRCh38
rs755987270 Chr.14: 52858866-52858866 on Build GRCh38
rs769262576 Chr.14: 52858527-52858527 on Build GRCh38
rs934961035 Chr.14: 52858638-52858638 on Build GRCh38
rs140069493 Chr.14: 52858761-52858761 on Build GRCh38
rs1036850903 Chr.14: 52858790-52858790 on Build GRCh38
rs777222342 Chr.14: 52858581-52858582 on Build GRCh38
rs374847495 Chr.14: 52858583-52858583 on Build GRCh38
rs995460547 Chr.14: 52858595-52858595 on Build GRCh38
rs1052194767 Chr.14: 52858668-52858668 on Build GRCh38
rs779050016 Chr.14: 52858427-52858427 on Build GRCh38
rs1050553080 Chr.14: 52858694-52858694 on Build GRCh38
rs956408481 Chr.14: 52858856-52858856 on Build GRCh38
rs574996852 Chr.14: 52858747-52858748 on Build GRCh38
rs777386057 Chr.14: 52858884-52858884 on Build GRCh38
rs946450738 Chr.14: 52858877-52858877 on Build GRCh38
rs371846367 Chr.14: 52858533-52858533 on Build GRCh38
rs200392394 Chr.14: 52858532-52858532 on Build GRCh38
rs1026883602 Chr.14: 52858601-52858601 on Build GRCh38
rs765397116 Chr.14: 52858671-52858671 on Build GRCh38
rs750406753 Chr.14: 52858428-52858428 on Build GRCh38
rs759376440 Chr.14: 52858559-52858559 on Build GRCh38
rs774494448 Chr.14: 52858748-52858749 on Build GRCh38
rs369867313 Chr.14: 52858584-52858584 on Build GRCh38
rs191900200 Chr.14: 52858603-52858603 on Build GRCh38
rs74322721 Chr.14: 52858834-52858834 on Build GRCh38
rs765322347 Chr.14: 52858593-52858593 on Build GRCh38
rs968337105 Chr.14: 52858704-52858705 on Build GRCh38
rs78244020 Chr.14: 52858747-52858747 on Build GRCh38
rs767332144 Chr.14: 52858569-52858569 on Build GRCh38
rs1019800006 Chr.14: 52858655-52858655 on Build GRCh38
rs770886964 Chr.14: 52858544-52858544 on Build GRCh38
rs113834568 Chr.14: 52858703-52858704 on Build GRCh38
rs1023453707 Chr.14: 52858816-52858816 on Build GRCh38
rs1018928615 Chr.14: 52858496-52858496 on Build GRCh38
rs143432898 Chr.14: 52858438-52858438 on Build GRCh38
rs930615102 Chr.14: 52858796-52858796 on Build GRCh38
rs758991496 Chr.14: 52858434-52858434 on Build GRCh38
rs937531501 Chr.14: 52858740-52858740 on Build GRCh38
rs979011973 Chr.14: 52858717-52858717 on Build GRCh38
rs762386974 Chr.14: 52858584-52858585 on Build GRCh38
rs367840082 Chr.14: 52858518-52858518 on Build GRCh38
rs764289666 Chr.14: 52858411-52858411 on Build GRCh38

View Full Product Details