Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52860230-52860750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
UniGene
Hs.509343
Gene Chromosome Location
Chr.14: 52857271-52951192 on Build GRCh38
Transcripts
NM_001134999.1 NM_001135000.1 NM_006832.2 XM_005267285.2 XM_006720008.2 XM_006720009.2 XM_006720010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
119872 c.1624G>T p.A542S Chr.14: 52860444-52860444 on Build GRCh38
6538099 c.1679A>C p.Q560P Chr.14: 52860389-52860389 on Build GRCh38
1607743 c.1708A>G p.I570V Chr.14: 52860360-52860360 on Build GRCh38
6140894 c.1706G>A p.G569D Chr.14: 52860362-52860362 on Build GRCh38
6573445 c.1657G>A p.E553K Chr.14: 52860411-52860411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032339179 Chr.14: 52860512-52860512 on Build GRCh38
rs764302852 Chr.14: 52860330-52860330 on Build GRCh38
rs766910241 Chr.14: 52860290-52860290 on Build GRCh38
rs536508728 Chr.14: 52860275-52860275 on Build GRCh38
rs73300775 Chr.14: 52860525-52860525 on Build GRCh38
rs760001288 Chr.14: 52860310-52860310 on Build GRCh38
rs532320305 Chr.14: 52860597-52860597 on Build GRCh38
rs376113224 Chr.14: 52860457-52860457 on Build GRCh38
rs559118793 Chr.14: 52860495-52860495 on Build GRCh38
rs779563079 Chr.14: 52860388-52860388 on Build GRCh38
rs750202366 Chr.14: 52860484-52860484 on Build GRCh38
rs757030080 Chr.14: 52860334-52860334 on Build GRCh38
rs1035842607 Chr.14: 52860557-52860557 on Build GRCh38
rs1010498492 Chr.14: 52860562-52860562 on Build GRCh38
rs754515190 Chr.14: 52860513-52860513 on Build GRCh38
rs767593715 Chr.14: 52860320-52860320 on Build GRCh38
rs764214654 Chr.14: 52860465-52860465 on Build GRCh38
rs538894781 Chr.14: 52860458-52860458 on Build GRCh38
rs766474778 Chr.14: 52860704-52860704 on Build GRCh38
rs202005528 Chr.14: 52860317-52860317 on Build GRCh38
rs144315461 Chr.14: 52860623-52860623 on Build GRCh38
rs760039004 Chr.14: 52860451-52860451 on Build GRCh38
rs886124095 Chr.14: 52860537-52860537 on Build GRCh38
rs780980333 Chr.14: 52860418-52860418 on Build GRCh38
rs557524522 Chr.14: 52860646-52860646 on Build GRCh38
rs978830830 Chr.14: 52860390-52860390 on Build GRCh38
rs754034679 Chr.14: 52860471-52860471 on Build GRCh38
rs750325446 Chr.14: 52860373-52860373 on Build GRCh38
rs984539400 Chr.14: 52860607-52860607 on Build GRCh38
rs943039013 Chr.14: 52860657-52860658 on Build GRCh38
rs567723478 Chr.14: 52860267-52860267 on Build GRCh38
rs779843175 Chr.14: 52860419-52860420 on Build GRCh38
rs745649592 Chr.14: 52860416-52860417 on Build GRCh38
rs74957625 Chr.14: 52860352-52860352 on Build GRCh38
rs17694337 Chr.14: 52860551-52860551 on Build GRCh38
rs763207056 Chr.14: 52860574-52860574 on Build GRCh38
rs534778199 Chr.14: 52860487-52860487 on Build GRCh38
rs377669901 Chr.14: 52860481-52860481 on Build GRCh38
rs758338635 Chr.14: 52860418-52860419 on Build GRCh38
rs531261672 Chr.14: 52860637-52860637 on Build GRCh38
rs768281043 Chr.14: 52860414-52860414 on Build GRCh38
rs191166807 Chr.14: 52860538-52860538 on Build GRCh38
rs989241070 Chr.14: 52860296-52860296 on Build GRCh38
rs371119593 Chr.14: 52860501-52860501 on Build GRCh38
rs748650959 Chr.14: 52860539-52860539 on Build GRCh38
rs758294456 Chr.14: 52860379-52860379 on Build GRCh38
rs754124470 Chr.14: 52860333-52860333 on Build GRCh38
rs760665291 Chr.14: 52860461-52860461 on Build GRCh38
rs746675103 Chr.14: 52860421-52860422 on Build GRCh38
rs762058174 Chr.14: 52860474-52860474 on Build GRCh38
rs143992413 Chr.14: 52860590-52860590 on Build GRCh38
rs7160292 Chr.14: 52860302-52860302 on Build GRCh38
rs72686362 Chr.14: 52860450-52860450 on Build GRCh38
rs148800969 Chr.14: 52860650-52860650 on Build GRCh38
rs778767876 Chr.14: 52860413-52860414 on Build GRCh38
rs138631913 Chr.14: 52860346-52860346 on Build GRCh38
rs141263595 Chr.14: 52860445-52860445 on Build GRCh38
rs138507326 Chr.14: 52860558-52860558 on Build GRCh38
rs552052633 Chr.14: 52860606-52860606 on Build GRCh38
rs746509189 Chr.14: 52860400-52860400 on Build GRCh38
rs184228351 Chr.14: 52860666-52860666 on Build GRCh38
rs756300173 Chr.14: 52860328-52860328 on Build GRCh38
rs752631647 Chr.14: 52860492-52860492 on Build GRCh38
rs777284765 Chr.14: 52860580-52860580 on Build GRCh38
rs769922426 Chr.14: 52860550-52860550 on Build GRCh38
rs143148232 Chr.14: 52860533-52860533 on Build GRCh38
rs750606214 Chr.14: 52860506-52860507 on Build GRCh38
rs752741036 Chr.14: 52860323-52860323 on Build GRCh38
rs964093472 Chr.14: 52860723-52860723 on Build GRCh38

View Full Product Details