Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 53040993-53041508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD1
Entrez Gene ID
80821
Gene Name
DDHD domain containing 1
Gene Aliases
PA-PLA1, PAPLA1, SPG28
UniGene
Hs.125525
Gene Chromosome Location
Chr.14: 53036740-53153328 on Build GRCh38
Transcripts
NM_001160147.1 NM_001160148.1 NM_030637.2 XM_005268102.2 XM_005268103.2 XM_005268105.2 XM_011537188.2 XM_011537189.2 XM_017021668.1 XM_017021669.1

SNP Detail

SNP ID Chromosome Location
rs915711635 Chr.14: 53041279-53041279 on Build GRCh38
rs778761619 Chr.14: 53041117-53041117 on Build GRCh38
rs889485249 Chr.14: 53041263-53041263 on Build GRCh38
rs1055656388 Chr.14: 53041036-53041036 on Build GRCh38
rs1037244250 Chr.14: 53041150-53041150 on Build GRCh38
rs549142967 Chr.14: 53041224-53041224 on Build GRCh38
rs1026692378 Chr.14: 53041035-53041035 on Build GRCh38
rs1054153522 Chr.14: 53041385-53041385 on Build GRCh38
rs760267399 Chr.14: 53041435-53041436 on Build GRCh38
rs764927485 Chr.14: 53041090-53041090 on Build GRCh38
rs949802014 Chr.14: 53041050-53041050 on Build GRCh38
rs915748310 Chr.14: 53041037-53041037 on Build GRCh38
rs779525616 Chr.14: 53041295-53041295 on Build GRCh38
rs139713840 Chr.14: 53041458-53041458 on Build GRCh38
rs924389006 Chr.14: 53041335-53041335 on Build GRCh38
rs371424001 Chr.14: 53041319-53041319 on Build GRCh38
rs541685941 Chr.14: 53041051-53041051 on Build GRCh38
rs1015624342 Chr.14: 53041455-53041455 on Build GRCh38
rs879124225 Chr.14: 53041148-53041148 on Build GRCh38
rs146838722 Chr.14: 53041389-53041389 on Build GRCh38
rs1019441754 Chr.14: 53041068-53041068 on Build GRCh38
rs969025452 Chr.14: 53041315-53041315 on Build GRCh38
rs566029746 Chr.14: 53041292-53041292 on Build GRCh38
rs143930334 Chr.14: 53041124-53041124 on Build GRCh38
rs571116841 Chr.14: 53041419-53041419 on Build GRCh38
rs17126073 Chr.14: 53041301-53041301 on Build GRCh38
rs902813944 Chr.14: 53041053-53041053 on Build GRCh38
rs979832384 Chr.14: 53041101-53041101 on Build GRCh38
rs533524457 Chr.14: 53041075-53041075 on Build GRCh38
rs978725148 Chr.14: 53041361-53041361 on Build GRCh38
rs934299167 Chr.14: 53041375-53041375 on Build GRCh38
rs762134825 Chr.14: 53041463-53041463 on Build GRCh38
rs111393779 Chr.14: 53041071-53041071 on Build GRCh38
rs1008233092 Chr.14: 53041125-53041125 on Build GRCh38
rs925335997 Chr.14: 53041104-53041104 on Build GRCh38
rs988699746 Chr.14: 53041074-53041074 on Build GRCh38
rs1006621841 Chr.14: 53041265-53041265 on Build GRCh38
rs191760699 Chr.14: 53041214-53041214 on Build GRCh38
rs997403666 Chr.14: 53041106-53041106 on Build GRCh38
rs949562060 Chr.14: 53041140-53041140 on Build GRCh38
rs932891884 Chr.14: 53041393-53041393 on Build GRCh38
rs529366173 Chr.14: 53041207-53041207 on Build GRCh38
rs550389369 Chr.14: 53041091-53041091 on Build GRCh38
rs74356329 Chr.14: 53041424-53041424 on Build GRCh38
rs951973274 Chr.14: 53041254-53041254 on Build GRCh38
rs1050441149 Chr.14: 53041414-53041414 on Build GRCh38
rs1020150230 Chr.14: 53041307-53041307 on Build GRCh38
rs974079282 Chr.14: 53041190-53041190 on Build GRCh38
rs12435568 Chr.14: 53041275-53041275 on Build GRCh38
rs973306180 Chr.14: 53041463-53041463 on Build GRCh38
rs771545480 Chr.14: 53041258-53041258 on Build GRCh38
rs745782826 Chr.14: 53041178-53041178 on Build GRCh38
rs374569014 Chr.14: 53041456-53041456 on Build GRCh38
rs750108010 Chr.14: 53041153-53041153 on Build GRCh38

View Full Product Details