Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 53044173-53044686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD1
Entrez Gene ID
80821
Gene Name
DDHD domain containing 1
Gene Aliases
PA-PLA1, PAPLA1, SPG28
UniGene
Hs.125525
Gene Chromosome Location
Chr.14: 53036740-53153328 on Build GRCh38
Transcripts
NM_001160147.1 NM_001160148.1 NM_030637.2 XM_005268102.2 XM_005268103.2 XM_005268105.2 XM_011537188.2 XM_011537189.2 XM_017021668.1 XM_017021669.1

SNP Detail

SNP ID Chromosome Location
rs983119155 Chr.14: 53044272-53044272 on Build GRCh38
rs536956335 Chr.14: 53044549-53044549 on Build GRCh38
rs926388853 Chr.14: 53044335-53044335 on Build GRCh38
rs1030205034 Chr.14: 53044309-53044312 on Build GRCh38
rs1006354341 Chr.14: 53044227-53044227 on Build GRCh38
rs1037410749 Chr.14: 53044581-53044581 on Build GRCh38
rs754550541 Chr.14: 53044326-53044326 on Build GRCh38
rs569016565 Chr.14: 53044467-53044467 on Build GRCh38
rs567927109 Chr.14: 53044548-53044548 on Build GRCh38
rs1044370065 Chr.14: 53044425-53044425 on Build GRCh38
rs866759777 Chr.14: 53044625-53044625 on Build GRCh38
rs962560744 Chr.14: 53044619-53044619 on Build GRCh38
rs945612769 Chr.14: 53044424-53044424 on Build GRCh38
rs552391021 Chr.14: 53044459-53044459 on Build GRCh38
rs35117986 Chr.14: 53044506-53044507 on Build GRCh38
rs1029518419 Chr.14: 53044244-53044244 on Build GRCh38
rs780667184 Chr.14: 53044587-53044587 on Build GRCh38
rs978060178 Chr.14: 53044379-53044379 on Build GRCh38
rs954776713 Chr.14: 53044330-53044330 on Build GRCh38
rs541778167 Chr.14: 53044480-53044480 on Build GRCh38
rs886123564 Chr.14: 53044220-53044220 on Build GRCh38
rs892148059 Chr.14: 53044417-53044417 on Build GRCh38
rs1016030239 Chr.14: 53044253-53044253 on Build GRCh38
rs548200340 Chr.14: 53044547-53044547 on Build GRCh38
rs970148014 Chr.14: 53044484-53044485 on Build GRCh38
rs753183178 Chr.14: 53044294-53044294 on Build GRCh38
rs901961286 Chr.14: 53044515-53044515 on Build GRCh38
rs553350679 Chr.14: 53044550-53044550 on Build GRCh38
rs540126993 Chr.14: 53044366-53044366 on Build GRCh38
rs997575308 Chr.14: 53044521-53044521 on Build GRCh38
rs980227260 Chr.14: 53044493-53044493 on Build GRCh38
rs747769747 Chr.14: 53044596-53044596 on Build GRCh38
rs78819079 Chr.14: 53044408-53044408 on Build GRCh38
rs1047829923 Chr.14: 53044217-53044217 on Build GRCh38
rs928842962 Chr.14: 53044502-53044502 on Build GRCh38
rs909683383 Chr.14: 53044305-53044305 on Build GRCh38
rs116995768 Chr.14: 53044266-53044266 on Build GRCh38
rs755730498 Chr.14: 53044646-53044646 on Build GRCh38
rs926461365 Chr.14: 53044641-53044641 on Build GRCh38
rs980508180 Chr.14: 53044328-53044328 on Build GRCh38
rs980624118 Chr.14: 53044639-53044639 on Build GRCh38
rs146396409 Chr.14: 53044509-53044509 on Build GRCh38
rs530779149 Chr.14: 53044546-53044547 on Build GRCh38
rs1026244511 Chr.14: 53044531-53044531 on Build GRCh38
rs976084753 Chr.14: 53044202-53044202 on Build GRCh38
rs180733328 Chr.14: 53044381-53044381 on Build GRCh38
rs879050267 Chr.14: 53044269-53044269 on Build GRCh38
rs566815335 Chr.14: 53044571-53044571 on Build GRCh38
rs900245699 Chr.14: 53044584-53044584 on Build GRCh38
rs938979837 Chr.14: 53044400-53044400 on Build GRCh38

View Full Product Details