Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 53045191-53045729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD1
Entrez Gene ID
80821
Gene Name
DDHD domain containing 1
Gene Aliases
PA-PLA1, PAPLA1, SPG28
UniGene
Hs.125525
Gene Chromosome Location
Chr.14: 53036740-53153328 on Build GRCh38
Transcripts
NM_001160147.1 NM_001160148.1 NM_030637.2 XM_005268102.2 XM_005268103.2 XM_005268105.2 XM_011537188.2 XM_011537189.2 XM_017021668.1 XM_017021669.1

SNP Detail

SNP ID Chromosome Location
rs149699505 Chr.14: 53045267-53045267 on Build GRCh38
rs902353091 Chr.14: 53045617-53045617 on Build GRCh38
rs776486985 Chr.14: 53045634-53045634 on Build GRCh38
rs530777217 Chr.14: 53045357-53045357 on Build GRCh38
rs963554690 Chr.14: 53045551-53045551 on Build GRCh38
rs979220764 Chr.14: 53045399-53045399 on Build GRCh38
rs1000984215 Chr.14: 53045620-53045620 on Build GRCh38
rs954929268 Chr.14: 53045675-53045675 on Build GRCh38
rs751557416 Chr.14: 53045489-53045489 on Build GRCh38
rs921571402 Chr.14: 53045364-53045364 on Build GRCh38
rs752214225 Chr.14: 53045567-53045567 on Build GRCh38
rs911636867 Chr.14: 53045449-53045449 on Build GRCh38
rs1013455923 Chr.14: 53045349-53045349 on Build GRCh38
rs7157751 Chr.14: 53045232-53045232 on Build GRCh38
rs937357048 Chr.14: 53045504-53045504 on Build GRCh38
rs538933825 Chr.14: 53045454-53045454 on Build GRCh38
rs373441630 Chr.14: 53045257-53045257 on Build GRCh38
rs964339750 Chr.14: 53045642-53045642 on Build GRCh38
rs1013524937 Chr.14: 53045540-53045540 on Build GRCh38
rs527337690 Chr.14: 53045383-53045383 on Build GRCh38
rs966840663 Chr.14: 53045380-53045380 on Build GRCh38
rs754852661 Chr.14: 53045503-53045503 on Build GRCh38
rs1033634124 Chr.14: 53045306-53045306 on Build GRCh38
rs577244748 Chr.14: 53045652-53045652 on Build GRCh38
rs987580758 Chr.14: 53045432-53045432 on Build GRCh38
rs905858246 Chr.14: 53045502-53045502 on Build GRCh38
rs896310194 Chr.14: 53045310-53045310 on Build GRCh38
rs547367116 Chr.14: 53045405-53045405 on Build GRCh38
rs964349672 Chr.14: 53045450-53045450 on Build GRCh38
rs974402950 Chr.14: 53045650-53045650 on Build GRCh38
rs1015071754 Chr.14: 53045549-53045549 on Build GRCh38
rs867841284 Chr.14: 53045499-53045499 on Build GRCh38
rs915040764 Chr.14: 53045694-53045694 on Build GRCh38
rs963606572 Chr.14: 53045321-53045321 on Build GRCh38
rs561734562 Chr.14: 53045381-53045381 on Build GRCh38
rs546084634 Chr.14: 53045701-53045701 on Build GRCh38
rs532500830 Chr.14: 53045632-53045632 on Build GRCh38
rs570731673 Chr.14: 53045522-53045522 on Build GRCh38
rs1029970516 Chr.14: 53045667-53045667 on Build GRCh38
rs533982192 Chr.14: 53045627-53045627 on Build GRCh38
rs190839290 Chr.14: 53045484-53045484 on Build GRCh38
rs147641046 Chr.14: 53045490-53045490 on Build GRCh38
rs1038077973 Chr.14: 53045274-53045274 on Build GRCh38
rs149123093 Chr.14: 53045512-53045512 on Build GRCh38
rs1001539452 Chr.14: 53045247-53045247 on Build GRCh38
rs988308599 Chr.14: 53045679-53045679 on Build GRCh38
rs147763267 Chr.14: 53045268-53045268 on Build GRCh38
rs935071123 Chr.14: 53045656-53045656 on Build GRCh38
rs185346159 Chr.14: 53045298-53045298 on Build GRCh38
rs574864626 Chr.14: 53045554-53045554 on Build GRCh38
rs953642768 Chr.14: 53045618-53045621 on Build GRCh38
rs550590044 Chr.14: 53045407-53045407 on Build GRCh38
rs141141679 Chr.14: 53045280-53045280 on Build GRCh38
rs1002080118 Chr.14: 53045305-53045305 on Build GRCh38
rs1029253705 Chr.14: 53045601-53045601 on Build GRCh38
rs1032132095 Chr.14: 53045623-53045623 on Build GRCh38
rs554186925 Chr.14: 53045631-53045631 on Build GRCh38
rs933471440 Chr.14: 53045483-53045483 on Build GRCh38
rs970044375 Chr.14: 53045242-53045242 on Build GRCh38
rs1005400766 Chr.14: 53045537-53045537 on Build GRCh38
rs933530470 Chr.14: 53045248-53045248 on Build GRCh38
rs146998828 Chr.14: 53045469-53045469 on Build GRCh38

View Full Product Details